Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202872954-202873500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC148709
Entrez Gene ID
148709
Gene Name
actin pseudogene
Gene Aliases
-
UniGene
Hs.162880
Gene Chromosome Location
Chr.1: 202861754-202875241 on Build GRCh38
Transcripts
NR_002929.2

SNP Detail

SNP ID Chromosome Location
rs745581491 Chr.1: 202873171-202873171 on Build GRCh38
rs541859657 Chr.1: 202873170-202873170 on Build GRCh38
rs190952071 Chr.1: 202873476-202873476 on Build GRCh38
rs1014753103 Chr.1: 202873022-202873022 on Build GRCh38
rs1008766677 Chr.1: 202873019-202873019 on Build GRCh38
rs186037984 Chr.1: 202873405-202873405 on Build GRCh38
rs72278447 Chr.1: 202873031-202873031 on Build GRCh38
rs560812887 Chr.1: 202873384-202873384 on Build GRCh38
rs549319263 Chr.1: 202873451-202873451 on Build GRCh38
rs869131966 Chr.1: 202873046-202873049 on Build GRCh38
rs991096650 Chr.1: 202873096-202873096 on Build GRCh38
rs945096457 Chr.1: 202873270-202873270 on Build GRCh38
rs796890024 Chr.1: 202873408-202873408 on Build GRCh38
rs539315559 Chr.1: 202873083-202873083 on Build GRCh38
rs116134781 Chr.1: 202873197-202873197 on Build GRCh38
rs749669181 Chr.1: 202873282-202873282 on Build GRCh38
rs12074513 Chr.1: 202873213-202873213 on Build GRCh38
rs897625231 Chr.1: 202873397-202873397 on Build GRCh38
rs1004397752 Chr.1: 202873168-202873168 on Build GRCh38
rs762346276 Chr.1: 202873427-202873427 on Build GRCh38
rs1028433933 Chr.1: 202873201-202873201 on Build GRCh38
rs867184353 Chr.1: 202873030-202873030 on Build GRCh38
rs931995711 Chr.1: 202873269-202873269 on Build GRCh38
rs543823408 Chr.1: 202873031-202873032 on Build GRCh38
rs1046116447 Chr.1: 202873478-202873478 on Build GRCh38
rs572730903 Chr.1: 202873120-202873120 on Build GRCh38
rs35546709 Chr.1: 202873048-202873048 on Build GRCh38
rs747535540 Chr.1: 202873032-202873033 on Build GRCh38
rs774204908 Chr.1: 202873087-202873087 on Build GRCh38
rs949794127 Chr.1: 202873144-202873144 on Build GRCh38
rs995179019 Chr.1: 202873398-202873398 on Build GRCh38
rs752924991 Chr.1: 202873031-202873033 on Build GRCh38
rs796385377 Chr.1: 202873048-202873049 on Build GRCh38
rs913664344 Chr.1: 202873243-202873243 on Build GRCh38
rs35355954 Chr.1: 202873049-202873049 on Build GRCh38
rs1055039233 Chr.1: 202873450-202873450 on Build GRCh38
rs768957085 Chr.1: 202873373-202873373 on Build GRCh38
rs890528553 Chr.1: 202872989-202872989 on Build GRCh38
rs139132356 Chr.1: 202873094-202873094 on Build GRCh38
rs12094889 Chr.1: 202873205-202873205 on Build GRCh38
rs1012569717 Chr.1: 202873471-202873471 on Build GRCh38
rs887168001 Chr.1: 202873153-202873153 on Build GRCh38
rs774748568 Chr.1: 202873374-202873374 on Build GRCh38
rs540906218 Chr.1: 202873377-202873377 on Build GRCh38
rs982724851 Chr.1: 202873146-202873146 on Build GRCh38
rs181816976 Chr.1: 202873279-202873279 on Build GRCh38
rs962310485 Chr.1: 202873040-202873040 on Build GRCh38
rs191070759 Chr.1: 202873196-202873196 on Build GRCh38
rs1056240757 Chr.1: 202873002-202873002 on Build GRCh38
rs531413612 Chr.1: 202873191-202873191 on Build GRCh38
rs986846288 Chr.1: 202873224-202873224 on Build GRCh38
rs747778578 Chr.1: 202873132-202873132 on Build GRCh38
rs937893086 Chr.1: 202872990-202872990 on Build GRCh38
rs555436842 Chr.1: 202873173-202873173 on Build GRCh38

View Full Product Details