Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2395610-2395933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA17P
Entrez Gene ID
650655
Gene Name
ATP binding cassette subfamily A member 17, pseudogene
Gene Aliases
ABCA17
UniGene
Hs.376281
Gene Chromosome Location
Chr.16: 2340922-2426699 on Build GRCh38
Transcripts
NR_003574.1

SNP Detail

SNP ID Chromosome Location
rs188857231 Chr.16: 2395777-2395777 on Build GRCh38
rs550593151 Chr.16: 2395905-2395905 on Build GRCh38
rs564318539 Chr.16: 2395653-2395653 on Build GRCh38
rs951105062 Chr.16: 2395732-2395732 on Build GRCh38
rs371535143 Chr.16: 2395739-2395739 on Build GRCh38
rs1023725726 Chr.16: 2395667-2395667 on Build GRCh38
rs540712567 Chr.16: 2395661-2395661 on Build GRCh38
rs534914843 Chr.16: 2395700-2395700 on Build GRCh38
rs73486173 Chr.16: 2395666-2395666 on Build GRCh38
rs1022268737 Chr.16: 2395738-2395738 on Build GRCh38
rs147435209 Chr.16: 2395660-2395660 on Build GRCh38
rs982492143 Chr.16: 2395765-2395765 on Build GRCh38
rs933878417 Chr.16: 2395833-2395833 on Build GRCh38
rs1038055485 Chr.16: 2395632-2395632 on Build GRCh38
rs191699205 Chr.16: 2395791-2395791 on Build GRCh38
rs922502801 Chr.16: 2395785-2395785 on Build GRCh38
rs767101419 Chr.16: 2395762-2395762 on Build GRCh38
rs943800686 Chr.16: 2395875-2395875 on Build GRCh38
rs911029288 Chr.16: 2395899-2395899 on Build GRCh38
rs969531756 Chr.16: 2395677-2395677 on Build GRCh38
rs903825045 Chr.16: 2395645-2395645 on Build GRCh38
rs896802507 Chr.16: 2395634-2395634 on Build GRCh38
rs79014681 Chr.16: 2395731-2395731 on Build GRCh38
rs73486175 Chr.16: 2395894-2395894 on Build GRCh38
rs999563369 Chr.16: 2395651-2395651 on Build GRCh38
rs959726971 Chr.16: 2395709-2395709 on Build GRCh38
rs966662277 Chr.16: 2395756-2395756 on Build GRCh38
rs544546207 Chr.16: 2395643-2395643 on Build GRCh38
rs896616818 Chr.16: 2395664-2395664 on Build GRCh38

View Full Product Details