Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 64972645-64973201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMZ2P1
Entrez Gene ID
201283
Gene Name
archaelysin family metallopeptidase 2 pseudogene 1
Gene Aliases
-
UniGene
Hs.396447
Gene Chromosome Location
Chr.17: 64966550-64975585 on Build GRCh38
Transcripts
NR_026903.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6283942 c.306G>T p.V102V Chr.17: 64972898-64972898 on Build GRCh38
1479876 c.302C>A p.S101* Chr.17: 64972902-64972902 on Build GRCh38
1630345 c.326C>T p.S109L Chr.17: 64972878-64972878 on Build GRCh38
7152871 c.359G>C p.*120S Chr.17: 64972845-64972845 on Build GRCh38
3820273 c.278A>T p.D93V Chr.17: 64972926-64972926 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs940392584 Chr.17: 64973069-64973069 on Build GRCh38
rs62072157 Chr.17: 64972711-64972711 on Build GRCh38
rs868764724 Chr.17: 64973044-64973044 on Build GRCh38
rs1036321770 Chr.17: 64972785-64972785 on Build GRCh38
rs533567070 Chr.17: 64972984-64972984 on Build GRCh38
rs568017881 Chr.17: 64972894-64972894 on Build GRCh38
rs562569302 Chr.17: 64972852-64972852 on Build GRCh38
rs781183625 Chr.17: 64972911-64972911 on Build GRCh38
rs878900629 Chr.17: 64972810-64972810 on Build GRCh38
rs547980045 Chr.17: 64972886-64972886 on Build GRCh38
rs765908509 Chr.17: 64972744-64972744 on Build GRCh38
rs540894334 Chr.17: 64972760-64972760 on Build GRCh38
rs746094966 Chr.17: 64972938-64972938 on Build GRCh38
rs56168156 Chr.17: 64972671-64972671 on Build GRCh38
rs547272711 Chr.17: 64973017-64973017 on Build GRCh38
rs111823317 Chr.17: 64972957-64972957 on Build GRCh38
rs912841583 Chr.17: 64972890-64972890 on Build GRCh38
rs896130405 Chr.17: 64973132-64973132 on Build GRCh38
rs554170237 Chr.17: 64972783-64972783 on Build GRCh38
rs931440377 Chr.17: 64972972-64972972 on Build GRCh38
rs756092903 Chr.17: 64973028-64973028 on Build GRCh38
rs373794080 Chr.17: 64972809-64972809 on Build GRCh38
rs988526908 Chr.17: 64972884-64972884 on Build GRCh38
rs921437433 Chr.17: 64972966-64972966 on Build GRCh38
rs539244961 Chr.17: 64973139-64973139 on Build GRCh38
rs905059580 Chr.17: 64973175-64973175 on Build GRCh38
rs1000964855 Chr.17: 64972850-64972850 on Build GRCh38
rs532739276 Chr.17: 64972824-64972824 on Build GRCh38
rs1044968323 Chr.17: 64973157-64973157 on Build GRCh38
rs975615148 Chr.17: 64972936-64972936 on Build GRCh38
rs1013511380 Chr.17: 64973137-64973137 on Build GRCh38
rs1049250568 Chr.17: 64972752-64972752 on Build GRCh38
rs767041869 Chr.17: 64972812-64972812 on Build GRCh38
rs1036424618 Chr.17: 64973099-64973099 on Build GRCh38
rs764033617 Chr.17: 64973059-64973059 on Build GRCh38
rs190679015 Chr.17: 64972832-64972832 on Build GRCh38
rs564983256 Chr.17: 64972725-64972725 on Build GRCh38
rs1013480326 Chr.17: 64972800-64972800 on Build GRCh38
rs887459774 Chr.17: 64972775-64972775 on Build GRCh38
rs769948836 Chr.17: 64973142-64973142 on Build GRCh38
rs561294796 Chr.17: 64972708-64972708 on Build GRCh38
rs537671951 Chr.17: 64972895-64972895 on Build GRCh38
rs61741348 Chr.17: 64972875-64972875 on Build GRCh38
rs1023491405 Chr.17: 64972849-64972849 on Build GRCh38
rs570439629 Chr.17: 64973025-64973025 on Build GRCh38
rs572337800 Chr.17: 64972790-64972790 on Build GRCh38

View Full Product Details