Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53601214-53601802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPRX
Entrez Gene ID
503834
Gene Name
divergent-paired related homeobox
Gene Aliases
-
UniGene
Hs.579941
Gene Chromosome Location
Chr.19: 53601257-53637057 on Build GRCh38
Transcripts
XM_011527011.2 NR_002938.3

Gene Symbol
LOC284379
Entrez Gene ID
284379
Gene Name
solute carrier family 7 member 3 pseudogene
Gene Aliases
-
UniGene
Hs.631571
Gene Chromosome Location
Chr.19: 53599631-53603497 on Build GRCh38
Transcripts
XM_011527011.2 NR_002938.3

SNP Detail

SNP ID Chromosome Location
rs888113867 Chr.19: 53601624-53601624 on Build GRCh38
rs895472910 Chr.19: 53601367-53601367 on Build GRCh38
rs192602119 Chr.19: 53601704-53601704 on Build GRCh38
rs539555905 Chr.19: 53601539-53601539 on Build GRCh38
rs900390312 Chr.19: 53601754-53601754 on Build GRCh38
rs756788450 Chr.19: 53601310-53601310 on Build GRCh38
rs1008719127 Chr.19: 53601735-53601735 on Build GRCh38
rs189246996 Chr.19: 53601686-53601686 on Build GRCh38
rs902928052 Chr.19: 53601320-53601320 on Build GRCh38
rs376492844 Chr.19: 53601772-53601772 on Build GRCh38
rs1031994453 Chr.19: 53601679-53601679 on Build GRCh38
rs918268975 Chr.19: 53601767-53601767 on Build GRCh38
rs113970094 Chr.19: 53601405-53601405 on Build GRCh38
rs369156091 Chr.19: 53601483-53601483 on Build GRCh38
rs947342776 Chr.19: 53601451-53601451 on Build GRCh38
rs139356746 Chr.19: 53601702-53601702 on Build GRCh38
rs138198250 Chr.19: 53601757-53601757 on Build GRCh38
rs994122833 Chr.19: 53601297-53601297 on Build GRCh38
rs1031710882 Chr.19: 53601725-53601725 on Build GRCh38
rs746348794 Chr.19: 53601745-53601745 on Build GRCh38
rs574873003 Chr.19: 53601627-53601627 on Build GRCh38
rs1030817290 Chr.19: 53601545-53601545 on Build GRCh38
rs890334215 Chr.19: 53601733-53601733 on Build GRCh38
rs746838991 Chr.19: 53601409-53601409 on Build GRCh38
rs898104606 Chr.19: 53601635-53601635 on Build GRCh38
rs371473572 Chr.19: 53601687-53601687 on Build GRCh38
rs149948328 Chr.19: 53601372-53601372 on Build GRCh38
rs979091155 Chr.19: 53601574-53601574 on Build GRCh38
rs7250277 Chr.19: 53601689-53601689 on Build GRCh38
rs909518597 Chr.19: 53601691-53601691 on Build GRCh38
rs555621977 Chr.19: 53601619-53601619 on Build GRCh38
rs77360407 Chr.19: 53601498-53601498 on Build GRCh38
rs747322910 Chr.19: 53601249-53601249 on Build GRCh38
rs767068928 Chr.19: 53601354-53601354 on Build GRCh38
rs946950355 Chr.19: 53601771-53601771 on Build GRCh38
rs534267964 Chr.19: 53601271-53601271 on Build GRCh38
rs940990602 Chr.19: 53601692-53601692 on Build GRCh38
rs754796103 Chr.19: 53601764-53601764 on Build GRCh38
rs746404711 Chr.19: 53601262-53601262 on Build GRCh38
rs566131741 Chr.19: 53601538-53601538 on Build GRCh38
rs527576802 Chr.19: 53601410-53601410 on Build GRCh38
rs999042851 Chr.19: 53601346-53601346 on Build GRCh38
rs549428416 Chr.19: 53601492-53601492 on Build GRCh38
rs968709731 Chr.19: 53601670-53601670 on Build GRCh38
rs894232191 Chr.19: 53601534-53601534 on Build GRCh38
rs34873138 Chr.19: 53601483-53601484 on Build GRCh38
rs929495655 Chr.19: 53601625-53601625 on Build GRCh38
rs148149947 Chr.19: 53601642-53601642 on Build GRCh38
rs1056765084 Chr.19: 53601350-53601350 on Build GRCh38
rs183510697 Chr.19: 53601710-53601710 on Build GRCh38
rs142986755 Chr.19: 53601727-53601727 on Build GRCh38
rs140752568 Chr.19: 53601393-53601393 on Build GRCh38
rs1016879494 Chr.19: 53601403-53601403 on Build GRCh38
rs910511200 Chr.19: 53601606-53601606 on Build GRCh38
rs114079196 Chr.19: 53601774-53601774 on Build GRCh38
rs559254411 Chr.19: 53601699-53601699 on Build GRCh38
rs1057302140 Chr.19: 53601473-53601473 on Build GRCh38
rs139191262 Chr.19: 53601256-53601256 on Build GRCh38
rs995267207 Chr.19: 53601755-53601755 on Build GRCh38
rs1038371287 Chr.19: 53601628-53601628 on Build GRCh38
rs929510309 Chr.19: 53601769-53601769 on Build GRCh38
rs577522497 Chr.19: 53601368-53601368 on Build GRCh38
rs531426648 Chr.19: 53601489-53601489 on Build GRCh38
rs575925123 Chr.19: 53601762-53601762 on Build GRCh38
rs768832275 Chr.19: 53601315-53601315 on Build GRCh38
rs934386776 Chr.19: 53601471-53601471 on Build GRCh38
rs1002509878 Chr.19: 53601529-53601529 on Build GRCh38
rs776933852 Chr.19: 53601258-53601258 on Build GRCh38
rs550486258 Chr.19: 53601513-53601513 on Build GRCh38
rs986125747 Chr.19: 53601419-53601419 on Build GRCh38
rs199847683 Chr.19: 53601276-53601276 on Build GRCh38
rs559186040 Chr.19: 53601352-53601352 on Build GRCh38
rs1043226135 Chr.19: 53601643-53601643 on Build GRCh38
rs1036721103 Chr.19: 53601693-53601693 on Build GRCh38
rs7250397 Chr.19: 53601761-53601761 on Build GRCh38
rs73053649 Chr.19: 53601459-53601459 on Build GRCh38
rs564057528 Chr.19: 53601683-53601683 on Build GRCh38
rs772934352 Chr.19: 53601357-53601357 on Build GRCh38
rs968573537 Chr.19: 53601572-53601572 on Build GRCh38
rs1007608098 Chr.19: 53601658-53601658 on Build GRCh38
rs74175790 Chr.19: 53601499-53601500 on Build GRCh38
rs954814196 Chr.19: 53601548-53601548 on Build GRCh38
rs568694681 Chr.19: 53601516-53601516 on Build GRCh38
rs547184438 Chr.19: 53601697-53601697 on Build GRCh38
rs377373250 Chr.19: 53601720-53601720 on Build GRCh38
rs530564796 Chr.19: 53601505-53601505 on Build GRCh38
rs562377011 Chr.19: 53601776-53601776 on Build GRCh38
rs573882723 Chr.19: 53601623-53601623 on Build GRCh38
rs146897299 Chr.19: 53601701-53601701 on Build GRCh38
rs560331990 Chr.19: 53601406-53601406 on Build GRCh38
rs924446430 Chr.19: 53601464-53601464 on Build GRCh38
rs534465400 Chr.19: 53601584-53601584 on Build GRCh38
rs141053575 Chr.19: 53601676-53601676 on Build GRCh38
rs1000587023 Chr.19: 53601724-53601724 on Build GRCh38
rs1017521546 Chr.19: 53601669-53601669 on Build GRCh38
rs571028631 Chr.19: 53601608-53601608 on Build GRCh38
rs545118597 Chr.19: 53601700-53601700 on Build GRCh38
rs1017969299 Chr.19: 53601752-53601752 on Build GRCh38
rs924503249 Chr.19: 53601439-53601439 on Build GRCh38
rs536023727 Chr.19: 53601528-53601528 on Build GRCh38
rs970836205 Chr.19: 53601532-53601532 on Build GRCh38
rs146523642 Chr.19: 53601392-53601392 on Build GRCh38
rs1031469917 Chr.19: 53601681-53601681 on Build GRCh38
rs752515887 Chr.19: 53601766-53601766 on Build GRCh38
rs973418655 Chr.19: 53601688-53601688 on Build GRCh38
rs772309569 Chr.19: 53601638-53601638 on Build GRCh38
rs934502223 Chr.19: 53601585-53601585 on Build GRCh38
rs939569439 Chr.19: 53601632-53601632 on Build GRCh38
rs556715906 Chr.19: 53601385-53601385 on Build GRCh38
rs550389238 Chr.19: 53601713-53601713 on Build GRCh38
rs557683977 Chr.19: 53601543-53601543 on Build GRCh38
rs956006178 Chr.19: 53601682-53601682 on Build GRCh38
rs982387246 Chr.19: 53601542-53601542 on Build GRCh38
rs73589316 Chr.19: 53601544-53601544 on Build GRCh38
rs920796201 Chr.19: 53601476-53601476 on Build GRCh38
rs1035047102 Chr.19: 53601768-53601768 on Build GRCh38
rs541999629 Chr.19: 53601629-53601629 on Build GRCh38
rs752970284 Chr.19: 53601365-53601365 on Build GRCh38
rs753379270 Chr.19: 53601306-53601306 on Build GRCh38
rs761350594 Chr.19: 53601286-53601286 on Build GRCh38
rs559295879 Chr.19: 53601586-53601586 on Build GRCh38
rs899418386 Chr.19: 53601287-53601287 on Build GRCh38
rs781054953 Chr.19: 53601267-53601267 on Build GRCh38
rs7249437 Chr.19: 53601728-53601728 on Build GRCh38
rs781044646 Chr.19: 53601765-53601765 on Build GRCh38
rs1038251955 Chr.19: 53601270-53601270 on Build GRCh38
rs71363354 Chr.19: 53601531-53601531 on Build GRCh38
rs1012575251 Chr.19: 53601518-53601518 on Build GRCh38
rs537091444 Chr.19: 53601567-53601567 on Build GRCh38
rs943409264 Chr.19: 53601466-53601466 on Build GRCh38
rs558204445 Chr.19: 53601241-53601242 on Build GRCh38
rs906783943 Chr.19: 53601517-53601517 on Build GRCh38
rs528971717 Chr.19: 53601694-53601694 on Build GRCh38
rs746698946 Chr.19: 53601235-53601235 on Build GRCh38
rs553219435 Chr.19: 53601770-53601770 on Build GRCh38
rs146692562 Chr.19: 53601450-53601450 on Build GRCh38
rs1030536363 Chr.19: 53601639-53601639 on Build GRCh38
rs565838466 Chr.19: 53601564-53601564 on Build GRCh38
rs572373469 Chr.19: 53601737-53601737 on Build GRCh38
rs928169339 Chr.19: 53601695-53601695 on Build GRCh38
rs750039586 Chr.19: 53601386-53601386 on Build GRCh38
rs758093714 Chr.19: 53601729-53601729 on Build GRCh38

View Full Product Details