Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200744112-200744670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOX3P-AOX2P
Entrez Gene ID
107161151
Gene Name
AOX3P-AOX2P readthrough
Gene Aliases
AOX3/AOX3L1
UniGene
Hs.148269
Gene Chromosome Location
Chr.2: 200695723-200794915 on Build GRCh38
Transcripts
NR_135011.1 NR_135012.1

SNP Detail

SNP ID Chromosome Location
rs974798642 Chr.2: 200744280-200744280 on Build GRCh38
rs370909402 Chr.2: 200744343-200744343 on Build GRCh38
rs943807912 Chr.2: 200744291-200744291 on Build GRCh38
rs557719443 Chr.2: 200744289-200744289 on Build GRCh38
rs1012625596 Chr.2: 200744386-200744386 on Build GRCh38
rs923256055 Chr.2: 200744545-200744545 on Build GRCh38
rs546834839 Chr.2: 200744603-200744603 on Build GRCh38
rs930192315 Chr.2: 200744338-200744338 on Build GRCh38
rs1048562254 Chr.2: 200744534-200744534 on Build GRCh38
rs1003612534 Chr.2: 200744410-200744410 on Build GRCh38
rs989121977 Chr.2: 200744254-200744254 on Build GRCh38
rs1045367477 Chr.2: 200744637-200744637 on Build GRCh38
rs977091285 Chr.2: 200744532-200744532 on Build GRCh38
rs888669087 Chr.2: 200744542-200744542 on Build GRCh38
rs190601982 Chr.2: 200744344-200744344 on Build GRCh38
rs186228520 Chr.2: 200744322-200744322 on Build GRCh38
rs942211444 Chr.2: 200744327-200744327 on Build GRCh38
rs769251797 Chr.2: 200744281-200744281 on Build GRCh38
rs976461061 Chr.2: 200744503-200744503 on Build GRCh38
rs539324722 Chr.2: 200744182-200744182 on Build GRCh38
rs963964704 Chr.2: 200744525-200744525 on Build GRCh38
rs541209037 Chr.2: 200744495-200744495 on Build GRCh38
rs575747201 Chr.2: 200744290-200744290 on Build GRCh38
rs906521596 Chr.2: 200744647-200744647 on Build GRCh38
rs1030769117 Chr.2: 200744230-200744230 on Build GRCh38
rs201801486 Chr.2: 200744454-200744454 on Build GRCh38
rs943345450 Chr.2: 200744269-200744269 on Build GRCh38
rs906593597 Chr.2: 200744400-200744400 on Build GRCh38
rs186590285 Chr.2: 200744524-200744524 on Build GRCh38
rs550661916 Chr.2: 200744207-200744207 on Build GRCh38
rs146937939 Chr.2: 200744424-200744424 on Build GRCh38
rs534555749 Chr.2: 200744318-200744318 on Build GRCh38
rs933313201 Chr.2: 200744555-200744555 on Build GRCh38
rs914529069 Chr.2: 200744259-200744259 on Build GRCh38
rs10931918 Chr.2: 200744397-200744397 on Build GRCh38
rs182061571 Chr.2: 200744484-200744484 on Build GRCh38
rs955850437 Chr.2: 200744438-200744438 on Build GRCh38
rs561546615 Chr.2: 200744406-200744406 on Build GRCh38
rs970811459 Chr.2: 200744148-200744148 on Build GRCh38
rs374617552 Chr.2: 200744354-200744354 on Build GRCh38
rs879849843 Chr.2: 200744473-200744473 on Build GRCh38
rs981244732 Chr.2: 200744156-200744156 on Build GRCh38
rs957873632 Chr.2: 200744235-200744235 on Build GRCh38
rs762643723 Chr.2: 200744466-200744466 on Build GRCh38
rs918373034 Chr.2: 200744514-200744514 on Build GRCh38
rs141221664 Chr.2: 200744170-200744170 on Build GRCh38
rs547260139 Chr.2: 200744140-200744140 on Build GRCh38
rs955884142 Chr.2: 200744200-200744200 on Build GRCh38
rs554672587 Chr.2: 200744275-200744275 on Build GRCh38
rs948349692 Chr.2: 200744567-200744567 on Build GRCh38
rs908000744 Chr.2: 200744613-200744613 on Build GRCh38
rs911179392 Chr.2: 200744251-200744251 on Build GRCh38
rs747057934 Chr.2: 200744246-200744246 on Build GRCh38
rs910869225 Chr.2: 200744478-200744478 on Build GRCh38
rs528679061 Chr.2: 200744546-200744546 on Build GRCh38
rs529010798 Chr.2: 200744443-200744443 on Build GRCh38
rs536320136 Chr.2: 200744258-200744258 on Build GRCh38
rs983529848 Chr.2: 200744591-200744591 on Build GRCh38
rs968676517 Chr.2: 200744461-200744461 on Build GRCh38
rs568928818 Chr.2: 200744245-200744245 on Build GRCh38

View Full Product Details