Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 184798188-184798749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL1
Entrez Gene ID
2180
Gene Name
acyl-CoA synthetase long-chain family member 1
Gene Aliases
ACS1, FACL1, FACL2, LACS, LACS1, LACS2
UniGene
Hs.406678
Gene Chromosome Location
Chr.4: 184755595-184826593 on Build GRCh38
Transcripts
NM_001286708.1 NM_001286710.1 NM_001286711.1 NM_001286712.1 NM_001995.3 XM_005262828.1 XM_005262829.1 XM_005262831.1 XM_011531742.1 XM_017007887.1 XM_017007888.1 XM_017007889.1 NR_003542.1

Gene Symbol
SLED1
Entrez Gene ID
643036
Gene Name
proteoglycan 3 pseudogene
Gene Aliases
-
UniGene
Hs.689630
Gene Chromosome Location
Chr.4: 184798296-184799046 on Build GRCh38
Transcripts
NM_001286708.1 NM_001286710.1 NM_001286711.1 NM_001286712.1 NM_001995.3 XM_005262828.1 XM_005262829.1 XM_005262831.1 XM_011531742.1 XM_017007887.1 XM_017007888.1 XM_017007889.1 NR_003542.1

SNP Detail

SNP ID Chromosome Location
rs894088109 Chr.4: 184798691-184798691 on Build GRCh38
rs146146237 Chr.4: 184798306-184798306 on Build GRCh38
rs547531144 Chr.4: 184798727-184798727 on Build GRCh38
rs934488907 Chr.4: 184798676-184798676 on Build GRCh38
rs933909528 Chr.4: 184798388-184798388 on Build GRCh38
rs576645709 Chr.4: 184798301-184798301 on Build GRCh38
rs955237807 Chr.4: 184798659-184798659 on Build GRCh38
rs1035167592 Chr.4: 184798278-184798278 on Build GRCh38
rs767374852 Chr.4: 184798391-184798391 on Build GRCh38
rs1031264708 Chr.4: 184798393-184798393 on Build GRCh38
rs560282102 Chr.4: 184798454-184798454 on Build GRCh38
rs921170609 Chr.4: 184798352-184798352 on Build GRCh38
rs545461573 Chr.4: 184798539-184798539 on Build GRCh38
rs192713580 Chr.4: 184798550-184798550 on Build GRCh38
rs951122709 Chr.4: 184798279-184798279 on Build GRCh38
rs891458465 Chr.4: 184798405-184798405 on Build GRCh38
rs975130704 Chr.4: 184798332-184798332 on Build GRCh38
rs749925623 Chr.4: 184798363-184798363 on Build GRCh38
rs1052627647 Chr.4: 184798679-184798679 on Build GRCh38
rs1004423203 Chr.4: 184798582-184798582 on Build GRCh38
rs975047861 Chr.4: 184798614-184798614 on Build GRCh38
rs1017077146 Chr.4: 184798610-184798610 on Build GRCh38
rs983371071 Chr.4: 184798506-184798506 on Build GRCh38
rs1045056928 Chr.4: 184798518-184798518 on Build GRCh38
rs752988612 Chr.4: 184798706-184798706 on Build GRCh38
rs932105683 Chr.4: 184798567-184798567 on Build GRCh38
rs1050894711 Chr.4: 184798412-184798412 on Build GRCh38
rs548404861 Chr.4: 184798545-184798545 on Build GRCh38
rs750118307 Chr.4: 184798413-184798413 on Build GRCh38
rs556016660 Chr.4: 184798387-184798387 on Build GRCh38
rs951622778 Chr.4: 184798562-184798562 on Build GRCh38
rs900619372 Chr.4: 184798288-184798288 on Build GRCh38
rs116577687 Chr.4: 184798453-184798453 on Build GRCh38
rs1011308354 Chr.4: 184798497-184798497 on Build GRCh38
rs140034172 Chr.4: 184798664-184798664 on Build GRCh38
rs920949961 Chr.4: 184798645-184798645 on Build GRCh38
rs1052942704 Chr.4: 184798466-184798466 on Build GRCh38
rs867205262 Chr.4: 184798530-184798530 on Build GRCh38
rs779676976 Chr.4: 184798611-184798611 on Build GRCh38
rs982947205 Chr.4: 184798285-184798285 on Build GRCh38
rs1035540672 Chr.4: 184798489-184798489 on Build GRCh38
rs371313526 Chr.4: 184798461-184798461 on Build GRCh38
rs867444367 Chr.4: 184798448-184798448 on Build GRCh38
rs541695498 Chr.4: 184798663-184798663 on Build GRCh38
rs560725345 Chr.4: 184798551-184798551 on Build GRCh38
rs778833260 Chr.4: 184798603-184798603 on Build GRCh38
rs921995469 Chr.4: 184798554-184798554 on Build GRCh38
rs576819793 Chr.4: 184798469-184798469 on Build GRCh38
rs541028642 Chr.4: 184798482-184798482 on Build GRCh38
rs527472064 Chr.4: 184798699-184798699 on Build GRCh38
rs187925649 Chr.4: 184798417-184798417 on Build GRCh38

View Full Product Details