Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 4983425-4983984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF216P1
Entrez Gene ID
441191
Gene Name
ring finger protein 216 pseudogene 1
Gene Aliases
RNF216L, hCG2040556
UniGene
Hs.520636
Gene Chromosome Location
Chr.7: 4973985-4998169 on Build GRCh38
Transcripts
NR_015449.1 NR_023384.1 NR_023385.1

SNP Detail

SNP ID Chromosome Location
rs902208761 Chr.7: 4983709-4983709 on Build GRCh38
rs914764720 Chr.7: 4983913-4983913 on Build GRCh38
rs550525689 Chr.7: 4983535-4983535 on Build GRCh38
rs1045993397 Chr.7: 4983607-4983607 on Build GRCh38
rs71529316 Chr.7: 4983868-4983868 on Build GRCh38
rs10231660 Chr.7: 4983940-4983940 on Build GRCh38
rs894853263 Chr.7: 4983675-4983675 on Build GRCh38
rs999465696 Chr.7: 4983542-4983542 on Build GRCh38
rs533698516 Chr.7: 4983598-4983598 on Build GRCh38
rs116203269 Chr.7: 4983872-4983872 on Build GRCh38
rs10156118 Chr.7: 4983571-4983571 on Build GRCh38
rs191130621 Chr.7: 4983943-4983943 on Build GRCh38
rs373582047 Chr.7: 4983541-4983541 on Build GRCh38
rs796915112 Chr.7: 4983805-4983805 on Build GRCh38
rs761819821 Chr.7: 4983467-4983468 on Build GRCh38
rs574781409 Chr.7: 4983755-4983755 on Build GRCh38
rs529610556 Chr.7: 4983902-4983902 on Build GRCh38
rs186756592 Chr.7: 4983455-4983455 on Build GRCh38
rs566230944 Chr.7: 4983597-4983597 on Build GRCh38
rs907897191 Chr.7: 4983587-4983587 on Build GRCh38
rs999434491 Chr.7: 4983725-4983725 on Build GRCh38
rs6965043 Chr.7: 4983789-4983789 on Build GRCh38
rs928802532 Chr.7: 4983615-4983615 on Build GRCh38
rs180952341 Chr.7: 4983705-4983705 on Build GRCh38
rs937552610 Chr.7: 4983641-4983641 on Build GRCh38
rs564693499 Chr.7: 4983838-4983838 on Build GRCh38
rs535942473 Chr.7: 4983769-4983769 on Build GRCh38
rs962055483 Chr.7: 4983828-4983829 on Build GRCh38
rs547790419 Chr.7: 4983914-4983914 on Build GRCh38
rs559780587 Chr.7: 4983922-4983922 on Build GRCh38
rs189539289 Chr.7: 4983671-4983671 on Build GRCh38
rs974851289 Chr.7: 4983553-4983553 on Build GRCh38
rs576656189 Chr.7: 4983856-4983856 on Build GRCh38
rs901763606 Chr.7: 4983728-4983728 on Build GRCh38
rs1028837315 Chr.7: 4983760-4983760 on Build GRCh38
rs142915144 Chr.7: 4983958-4983958 on Build GRCh38
rs1018775322 Chr.7: 4983544-4983544 on Build GRCh38
rs1010670173 Chr.7: 4983681-4983681 on Build GRCh38
rs867074654 Chr.7: 4983608-4983608 on Build GRCh38
rs199562894 Chr.7: 4983504-4983507 on Build GRCh38
rs755811148 Chr.7: 4983464-4983464 on Build GRCh38
rs531930064 Chr.7: 4983509-4983509 on Build GRCh38
rs572584016 Chr.7: 4983790-4983790 on Build GRCh38
rs376162289 Chr.7: 4983534-4983534 on Build GRCh38
rs1021970105 Chr.7: 4983485-4983488 on Build GRCh38
rs71535160 Chr.7: 4983823-4983828 on Build GRCh38
rs1018912668 Chr.7: 4983726-4983726 on Build GRCh38
rs1043763550 Chr.7: 4983704-4983704 on Build GRCh38
rs1014866979 Chr.7: 4983822-4983822 on Build GRCh38
rs777915612 Chr.7: 4983449-4983449 on Build GRCh38
rs937617294 Chr.7: 4983874-4983874 on Build GRCh38
rs375558986 Chr.7: 4983822-4983823 on Build GRCh38
rs866207378 Chr.7: 4983738-4983738 on Build GRCh38
rs141733729 Chr.7: 4983665-4983665 on Build GRCh38
rs60609016 Chr.7: 4983473-4983474 on Build GRCh38
rs370468261 Chr.7: 4983468-4983469 on Build GRCh38
rs570194601 Chr.7: 4983823-4983825 on Build GRCh38
rs185135529 Chr.7: 4983813-4983813 on Build GRCh38
rs768008865 Chr.7: 4983825-4983826 on Build GRCh38
rs1043528630 Chr.7: 4983933-4983933 on Build GRCh38
rs551967908 Chr.7: 4983643-4983643 on Build GRCh38
rs952202743 Chr.7: 4983771-4983771 on Build GRCh38
rs148124723 Chr.7: 4983585-4983585 on Build GRCh38
rs564731901 Chr.7: 4983496-4983496 on Build GRCh38
rs562375396 Chr.7: 4983548-4983548 on Build GRCh38

View Full Product Details