Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1487647-1487946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD3B
Entrez Gene ID
83858
Gene Name
ATPase family, AAA domain containing 3B
Gene Aliases
AAA-TOB3, TOB3
UniGene
Hs.729021
Gene Chromosome Location
Chr.1: 1471732-1509466 on Build GRCh38
Transcripts
NM_001317238.1 NM_031921.5 XM_005244806.3 XM_011542241.2 XM_011542243.2 XM_011542244.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6871076 c.1262C>T p.A421V Chr.1: 1487910-1487910 on Build GRCh38
6606411 c.1243G>A p.A415T Chr.1: 1487891-1487891 on Build GRCh38
4984865 c.1258C>G p.R420G Chr.1: 1487906-1487906 on Build GRCh38
4021571 c.1242C>T p.D414D Chr.1: 1487890-1487890 on Build GRCh38
6019225 c.1239A>G p.A413A Chr.1: 1487887-1487887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749656044 Chr.1: 1487902-1487902 on Build GRCh38
rs147191465 Chr.1: 1487867-1487867 on Build GRCh38
rs755374112 Chr.1: 1487904-1487904 on Build GRCh38
rs939324224 Chr.1: 1487836-1487836 on Build GRCh38
rs956822321 Chr.1: 1487747-1487747 on Build GRCh38
rs186532722 Chr.1: 1487707-1487707 on Build GRCh38
rs1022454226 Chr.1: 1487819-1487819 on Build GRCh38
rs542039116 Chr.1: 1487793-1487793 on Build GRCh38
rs369424999 Chr.1: 1487796-1487796 on Build GRCh38
rs763773084 Chr.1: 1487893-1487893 on Build GRCh38
rs375527319 Chr.1: 1487824-1487824 on Build GRCh38
rs550703705 Chr.1: 1487676-1487676 on Build GRCh38
rs773103691 Chr.1: 1487907-1487907 on Build GRCh38
rs989583318 Chr.1: 1487760-1487760 on Build GRCh38
rs1031235704 Chr.1: 1487806-1487806 on Build GRCh38
rs187809296 Chr.1: 1487857-1487857 on Build GRCh38
rs774024590 Chr.1: 1487920-1487920 on Build GRCh38
rs573569267 Chr.1: 1487802-1487802 on Build GRCh38
rs763719670 Chr.1: 1487852-1487852 on Build GRCh38
rs1056996934 Chr.1: 1487687-1487687 on Build GRCh38
rs145173454 Chr.1: 1487849-1487849 on Build GRCh38
rs772485433 Chr.1: 1487904-1487905 on Build GRCh38
rs373064964 Chr.1: 1487809-1487809 on Build GRCh38
rs777949818 Chr.1: 1487844-1487844 on Build GRCh38
rs1056938667 Chr.1: 1487777-1487777 on Build GRCh38
rs766973280 Chr.1: 1487899-1487899 on Build GRCh38
rs759328778 Chr.1: 1487890-1487890 on Build GRCh38
rs756780839 Chr.1: 1487897-1487897 on Build GRCh38
rs779302285 Chr.1: 1487905-1487905 on Build GRCh38
rs769690327 Chr.1: 1487910-1487910 on Build GRCh38
rs577666979 Chr.1: 1487773-1487773 on Build GRCh38
rs773514564 Chr.1: 1487915-1487918 on Build GRCh38
rs767143072 Chr.1: 1487925-1487925 on Build GRCh38
rs142126053 Chr.1: 1487914-1487914 on Build GRCh38
rs762708254 Chr.1: 1487912-1487912 on Build GRCh38
rs748289945 Chr.1: 1487885-1487885 on Build GRCh38
rs1031185579 Chr.1: 1487737-1487737 on Build GRCh38
rs79849353 Chr.1: 1487901-1487901 on Build GRCh38
rs938619685 Chr.1: 1487771-1487771 on Build GRCh38
rs1029055659 Chr.1: 1487717-1487717 on Build GRCh38
rs199705339 Chr.1: 1487900-1487900 on Build GRCh38
rs530396032 Chr.1: 1487875-1487875 on Build GRCh38
rs191811766 Chr.1: 1487795-1487795 on Build GRCh38
rs781660761 Chr.1: 1487818-1487818 on Build GRCh38
rs571285142 Chr.1: 1487906-1487906 on Build GRCh38
rs756532841 Chr.1: 1487862-1487862 on Build GRCh38
rs567098844 Chr.1: 1487723-1487723 on Build GRCh38
rs374004343 Chr.1: 1487902-1487906 on Build GRCh38
rs915300702 Chr.1: 1487761-1487761 on Build GRCh38
rs940496964 Chr.1: 1487778-1487778 on Build GRCh38
rs375185569 Chr.1: 1487794-1487794 on Build GRCh38
rs772233246 Chr.1: 1487886-1487886 on Build GRCh38
rs765089718 Chr.1: 1487891-1487891 on Build GRCh38
rs547187655 Chr.1: 1487708-1487708 on Build GRCh38
rs1004590840 Chr.1: 1487696-1487696 on Build GRCh38
rs780224737 Chr.1: 1487871-1487871 on Build GRCh38
rs749717160 Chr.1: 1487829-1487830 on Build GRCh38
rs886239083 Chr.1: 1487694-1487694 on Build GRCh38
rs779191408 Chr.1: 1487846-1487849 on Build GRCh38
rs530983508 Chr.1: 1487674-1487674 on Build GRCh38
rs759401597 Chr.1: 1487840-1487840 on Build GRCh38
rs763829941 Chr.1: 1487915-1487915 on Build GRCh38
rs988122122 Chr.1: 1487735-1487735 on Build GRCh38
rs183587099 Chr.1: 1487798-1487798 on Build GRCh38
rs757926457 Chr.1: 1487850-1487850 on Build GRCh38
rs746269526 Chr.1: 1487820-1487820 on Build GRCh38
rs148709813 Chr.1: 1487876-1487876 on Build GRCh38
rs819977 Chr.1: 1487887-1487887 on Build GRCh38
rs181389167 Chr.1: 1487677-1487677 on Build GRCh38
rs141072509 Chr.1: 1487837-1487837 on Build GRCh38
rs773985897 Chr.1: 1487846-1487847 on Build GRCh38
rs369873036 Chr.1: 1487825-1487825 on Build GRCh38
rs534698017 Chr.1: 1487724-1487724 on Build GRCh38
rs751189348 Chr.1: 1487895-1487895 on Build GRCh38
rs557880639 Chr.1: 1487727-1487727 on Build GRCh38
rs865852435 Chr.1: 1487896-1487896 on Build GRCh38
rs565127375 Chr.1: 1487670-1487670 on Build GRCh38
rs373211432 Chr.1: 1487834-1487834 on Build GRCh38
rs763044901 Chr.1: 1487833-1487833 on Build GRCh38
rs751134174 Chr.1: 1487854-1487854 on Build GRCh38
rs948056706 Chr.1: 1487673-1487673 on Build GRCh38
rs1037482174 Chr.1: 1487792-1487792 on Build GRCh38
rs372066277 Chr.1: 1487827-1487827 on Build GRCh38
rs1009156781 Chr.1: 1487805-1487805 on Build GRCh38
rs766462525 Chr.1: 1487838-1487838 on Build GRCh38
rs373575774 Chr.1: 1487855-1487855 on Build GRCh38
rs753808119 Chr.1: 1487839-1487839 on Build GRCh38
rs980821130 Chr.1: 1487767-1487767 on Build GRCh38
rs751526787 Chr.1: 1487799-1487799 on Build GRCh38
rs556588325 Chr.1: 1487669-1487669 on Build GRCh38
rs559103079 Chr.1: 1487810-1487810 on Build GRCh38
rs955025220 Chr.1: 1487722-1487722 on Build GRCh38

View Full Product Details