Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112455835-112456009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPN11
Entrez Gene ID
5781
Gene Name
protein tyrosine phosphatase, non-receptor type 11
Gene Aliases
BPTP3, CFC, JMML, METCDS, NS1, PTP-1D, PTP2C, SH-PTP2, SH-PTP3, SHP2
UniGene
Hs.506852
Gene Chromosome Location
Chr.12: 112418732-112509913 on Build GRCh38
Transcripts
NM_002834.3 NM_080601.1 XM_006719526.2 XM_011538613.2 XM_017019722.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6399855 c.659G>A p.R220H Chr.12: 112455966-112455966 on Build GRCh38
6325411 c.643-6G>A p.? Chr.12: 112455944-112455944 on Build GRCh38
692039 c.673G>T p.E225* Chr.12: 112455980-112455980 on Build GRCh38
1222515 c.675A>C p.E225D Chr.12: 112455982-112455982 on Build GRCh38
3359483 c.687A>G p.R229R Chr.12: 112455994-112455994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568340391 Chr.12: 112455961-112455961 on Build GRCh38
rs943458246 Chr.12: 112455887-112455887 on Build GRCh38
rs780911423 Chr.12: 112455966-112455966 on Build GRCh38
rs183857035 Chr.12: 112455889-112455889 on Build GRCh38
rs140552217 Chr.12: 112455872-112455872 on Build GRCh38
rs755400416 Chr.12: 112455913-112455913 on Build GRCh38
rs566654281 Chr.12: 112455872-112455874 on Build GRCh38
rs750520685 Chr.12: 112455941-112455941 on Build GRCh38
rs928283406 Chr.12: 112455871-112455871 on Build GRCh38
rs397516807 Chr.12: 112455968-112455968 on Build GRCh38
rs398122861 Chr.12: 112455948-112455948 on Build GRCh38
rs757773210 Chr.12: 112455921-112455921 on Build GRCh38
rs758889732 Chr.12: 112455943-112455944 on Build GRCh38
rs397516806 Chr.12: 112455968-112455968 on Build GRCh38
rs768555552 Chr.12: 112455959-112455959 on Build GRCh38
rs758575627 Chr.12: 112455942-112455942 on Build GRCh38
rs895890372 Chr.12: 112455872-112455873 on Build GRCh38
rs779021487 Chr.12: 112455933-112455933 on Build GRCh38
rs569070852 Chr.12: 112455874-112455874 on Build GRCh38
rs1022983191 Chr.12: 112455922-112455922 on Build GRCh38
rs754356317 Chr.12: 112455917-112455917 on Build GRCh38
rs886925676 Chr.12: 112455860-112455860 on Build GRCh38
rs374084684 Chr.12: 112455899-112455899 on Build GRCh38
rs780387568 Chr.12: 112455944-112455944 on Build GRCh38
rs1014313069 Chr.12: 112455900-112455900 on Build GRCh38
rs747243342 Chr.12: 112455958-112455958 on Build GRCh38
rs905837482 Chr.12: 112455926-112455926 on Build GRCh38
rs193226579 Chr.12: 112455877-112455877 on Build GRCh38

View Full Product Details