Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 8055742-8055883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA3
Entrez Gene ID
2625
Gene Name
GATA binding protein 3
Gene Aliases
HDR, HDRS
UniGene
Hs.524134
Gene Chromosome Location
Chr.10: 8045421-8075201 on Build GRCh38
Transcripts
NM_001002295.1 NM_002051.2 XM_005252442.2 XM_005252443.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4771385 c.200C>G p.S67W Chr.10: 8055855-8055855 on Build GRCh38
1207942 c.202G>T p.V68F Chr.10: 8055857-8055857 on Build GRCh38
4427532 c.121T>C p.Y41H Chr.10: 8055776-8055776 on Build GRCh38
6571666 c.137_138insG p.V47fs*6 Chr.10: 8055792-8055793 on Build GRCh38
7345128 c.183_184insC p.Y63fs*241 Chr.10: 8055838-8055839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745997183 Chr.10: 8055782-8055782 on Build GRCh38
rs769859105 Chr.10: 8055787-8055787 on Build GRCh38
rs939494825 Chr.10: 8055761-8055761 on Build GRCh38
rs976703495 Chr.10: 8055766-8055766 on Build GRCh38
rs774975933 Chr.10: 8055779-8055779 on Build GRCh38
rs763798635 Chr.10: 8055847-8055847 on Build GRCh38
rs545189339 Chr.10: 8055769-8055769 on Build GRCh38
rs774154154 Chr.10: 8055860-8055860 on Build GRCh38
rs371853270 Chr.10: 8055843-8055843 on Build GRCh38
rs932686919 Chr.10: 8055861-8055861 on Build GRCh38
rs763176056 Chr.10: 8055818-8055818 on Build GRCh38
rs778144734 Chr.10: 8055764-8055764 on Build GRCh38
rs775745771 Chr.10: 8055803-8055803 on Build GRCh38
rs747477962 Chr.10: 8055767-8055767 on Build GRCh38
rs761676499 Chr.10: 8055864-8055864 on Build GRCh38

View Full Product Details