Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 112766343-112766497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APC
Entrez Gene ID
324
Gene Name
APC, WNT signaling pathway regulator
Gene Aliases
BTPS2, DP2, DP2.5, DP3, GS, PPP1R46
UniGene
Hs.158932
Gene Chromosome Location
Chr.5: 112707505-112846239 on Build GRCh38
Transcripts
NM_000038.5 NM_001127510.2 NM_001127511.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6396851 c.182C>A p.A61D Chr.5: 112766372-112766372 on Build GRCh38
6019368 c.211C>T p.R71C Chr.5: 112766401-112766401 on Build GRCh38
6475382 c.190G>A p.G64R Chr.5: 112766380-112766380 on Build GRCh38
6601962 c.174A>C p.E58D Chr.5: 112766364-112766364 on Build GRCh38
5356439 c.208G>A p.E70K Chr.5: 112766398-112766398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973491846 Chr.5: 112766414-112766414 on Build GRCh38
rs777631286 Chr.5: 112766460-112766460 on Build GRCh38
rs374729480 Chr.5: 112766454-112766454 on Build GRCh38
rs761957941 Chr.5: 112766400-112766400 on Build GRCh38
rs750503329 Chr.5: 112766402-112766402 on Build GRCh38
rs754354718 Chr.5: 112766429-112766429 on Build GRCh38
rs764962781 Chr.5: 112766386-112766386 on Build GRCh38
rs200746873 Chr.5: 112766438-112766438 on Build GRCh38
rs786201989 Chr.5: 112766371-112766371 on Build GRCh38
rs876660575 Chr.5: 112766377-112766379 on Build GRCh38
rs927125994 Chr.5: 112766428-112766428 on Build GRCh38
rs863225325 Chr.5: 112766397-112766397 on Build GRCh38
rs863224539 Chr.5: 112766399-112766399 on Build GRCh38
rs756214797 Chr.5: 112766426-112766426 on Build GRCh38
rs864622493 Chr.5: 112766420-112766422 on Build GRCh38
rs776469589 Chr.5: 112766377-112766377 on Build GRCh38
rs587781809 Chr.5: 112766412-112766412 on Build GRCh38
rs771834686 Chr.5: 112766453-112766453 on Build GRCh38
rs1054485399 Chr.5: 112766441-112766441 on Build GRCh38
rs752519066 Chr.5: 112766396-112766396 on Build GRCh38
rs79323615 Chr.5: 112766380-112766380 on Build GRCh38
rs767741687 Chr.5: 112766401-112766401 on Build GRCh38
rs74727182 Chr.5: 112766375-112766375 on Build GRCh38
rs937184733 Chr.5: 112766467-112766467 on Build GRCh38
rs863225320 Chr.5: 112766383-112766383 on Build GRCh38
rs74340509 Chr.5: 112766448-112766448 on Build GRCh38
rs748533525 Chr.5: 112766449-112766449 on Build GRCh38
rs779383707 Chr.5: 112766440-112766440 on Build GRCh38
rs876658941 Chr.5: 112766410-112766410 on Build GRCh38
rs759352811 Chr.5: 112766379-112766379 on Build GRCh38
rs878853421 Chr.5: 112766381-112766381 on Build GRCh38

View Full Product Details