Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 31722048-31722186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABALON
Entrez Gene ID
103021294
Gene Name
apoptotic BCL2L1-antisense long non-coding RNA
Gene Aliases
INXS
UniGene
Gene Chromosome Location
Chr.20: 31721507-31723409 on Build GRCh38
Transcripts
NR_131907.1 NM_001191.3 NM_001317919.1 NM_001317920.1 NM_001317921.1 NM_001322239.1 NM_001322240.1 NM_001322242.1 NM_138578.2 NR_134257.1 XM_011528964.2 XM_017027993.1

Gene Symbol
BCL2L1
Entrez Gene ID
598
Gene Name
BCL2 like 1
Gene Aliases
BCL-XL/S, BCL2L, BCLX, Bcl-X, PPP1R52
UniGene
Hs.516966
Gene Chromosome Location
Chr.20: 31664452-31723999 on Build GRCh38
Transcripts
NR_131907.1 NM_001191.3 NM_001317919.1 NM_001317920.1 NM_001317921.1 NM_001322239.1 NM_001322240.1 NM_001322242.1 NM_138578.2 NR_134257.1 XM_011528964.2 XM_017027993.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2888609 c.96G>T p.E32D Chr.20: 31722123-31722123 on Build GRCh38
1307252 c.90G>A p.V30V Chr.20: 31722129-31722129 on Build GRCh38
5544522 c.116A>T p.E39V Chr.20: 31722103-31722103 on Build GRCh38
159351 c.76C>A p.Q26K Chr.20: 31722143-31722143 on Build GRCh38
4290947 c.99C>T p.N33N Chr.20: 31722120-31722120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753681012 Chr.20: 31722137-31722137 on Build GRCh38
rs774465563 Chr.20: 31722101-31722101 on Build GRCh38
rs766199813 Chr.20: 31722124-31722124 on Build GRCh38
rs769141052 Chr.20: 31722100-31722100 on Build GRCh38
rs752845959 Chr.20: 31722119-31722121 on Build GRCh38
rs376431497 Chr.20: 31722077-31722077 on Build GRCh38
rs761851595 Chr.20: 31722102-31722102 on Build GRCh38
rs760750448 Chr.20: 31722123-31722123 on Build GRCh38
rs777448557 Chr.20: 31722068-31722068 on Build GRCh38
rs145910874 Chr.20: 31722091-31722091 on Build GRCh38
rs772005799 Chr.20: 31722111-31722111 on Build GRCh38
rs773505559 Chr.20: 31722118-31722118 on Build GRCh38
rs770213331 Chr.20: 31722087-31722087 on Build GRCh38
rs11550476 Chr.20: 31722090-31722090 on Build GRCh38

View Full Product Details