Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 122892977-122893220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF2
Entrez Gene ID
2247
Gene Name
fibroblast growth factor 2
Gene Aliases
BFGF, FGF-2, FGFB, HBGF-2
UniGene
Hs.284244
Gene Chromosome Location
Chr.4: 122826708-122898235 on Build GRCh38
Transcripts
NM_002006.4 NM_007083.4 NM_198041.2

Gene Symbol
NUDT6
Entrez Gene ID
11162
Gene Name
nudix hydrolase 6
Gene Aliases
ASFGF2, FGF-AS, FGF2AS, GFG-1, GFG1
UniGene
Hs.558459
Gene Chromosome Location
Chr.4: 122892644-122923004 on Build GRCh38
Transcripts
NM_002006.4 NM_007083.4 NM_198041.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4980678 c.802G>T p.A268S Chr.4: 122892977-122892977 on Build GRCh38
5400417 c.251A>G p.D84G Chr.4: 122893021-122893021 on Build GRCh38
5400416 c.758A>G p.D253G Chr.4: 122893021-122893021 on Build GRCh38
5595010 c.191C>T p.S64L Chr.4: 122893081-122893081 on Build GRCh38
5737082 c.82G>T p.G28C Chr.4: 122893190-122893190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891833866 Chr.4: 122893164-122893164 on Build GRCh38
rs201458145 Chr.4: 122893166-122893166 on Build GRCh38
rs769818641 Chr.4: 122893129-122893129 on Build GRCh38
rs751612205 Chr.4: 122893056-122893056 on Build GRCh38
rs776567689 Chr.4: 122893017-122893017 on Build GRCh38
rs761529651 Chr.4: 122893024-122893024 on Build GRCh38
rs201373471 Chr.4: 122893080-122893080 on Build GRCh38
rs765178237 Chr.4: 122893033-122893033 on Build GRCh38
rs775987914 Chr.4: 122893111-122893111 on Build GRCh38
rs931437932 Chr.4: 122893085-122893085 on Build GRCh38
rs755045449 Chr.4: 122893070-122893070 on Build GRCh38
rs770075653 Chr.4: 122893005-122893005 on Build GRCh38
rs113836715 Chr.4: 122893167-122893167 on Build GRCh38
rs372728120 Chr.4: 122893150-122893150 on Build GRCh38
rs369473521 Chr.4: 122893122-122893122 on Build GRCh38
rs746508996 Chr.4: 122893097-122893097 on Build GRCh38
rs771682042 Chr.4: 122893012-122893012 on Build GRCh38
rs142756927 Chr.4: 122893096-122893096 on Build GRCh38
rs749632195 Chr.4: 122893087-122893087 on Build GRCh38
rs45606640 Chr.4: 122893009-122893009 on Build GRCh38
rs756611528 Chr.4: 122893077-122893077 on Build GRCh38
rs201412128 Chr.4: 122893190-122893190 on Build GRCh38
rs763303117 Chr.4: 122893044-122893044 on Build GRCh38
rs746637657 Chr.4: 122893194-122893194 on Build GRCh38
rs938922060 Chr.4: 122893146-122893146 on Build GRCh38
rs781553867 Chr.4: 122893072-122893072 on Build GRCh38
rs374780215 Chr.4: 122893194-122893194 on Build GRCh38
rs778205008 Chr.4: 122893192-122893192 on Build GRCh38
rs752689945 Chr.4: 122893176-122893176 on Build GRCh38
rs768158192 Chr.4: 122893104-122893104 on Build GRCh38
rs1048201 Chr.4: 122893153-122893153 on Build GRCh38
rs760186313 Chr.4: 122893014-122893014 on Build GRCh38
rs1005973997 Chr.4: 122893100-122893100 on Build GRCh38
rs778098774 Chr.4: 122893008-122893008 on Build GRCh38
rs766682539 Chr.4: 122893050-122893050 on Build GRCh38
rs750236034 Chr.4: 122893043-122893043 on Build GRCh38
rs751736046 Chr.4: 122893154-122893154 on Build GRCh38
rs199533328 Chr.4: 122893130-122893130 on Build GRCh38
rs753188971 Chr.4: 122893076-122893076 on Build GRCh38
rs138619142 Chr.4: 122893015-122893015 on Build GRCh38
rs761906165 Chr.4: 122893114-122893114 on Build GRCh38
rs771737486 Chr.4: 122893092-122893092 on Build GRCh38

View Full Product Details