Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127942047-127942300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAP130
Entrez Gene ID
79595
Gene Name
Sin3A associated protein 130
Gene Aliases
-
UniGene
Hs.32995
Gene Chromosome Location
Chr.2: 127941217-128028093 on Build GRCh38
Transcripts
NM_001145928.1 NM_024545.3 XM_005263767.3 XM_005263768.3 XM_005263769.3 XM_005263770.3 XM_006712748.3 XM_006712749.3 XM_017004903.1 XM_017004904.1 XM_017004905.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3797947 c.3143G>C p.R1048T Chr.2: 127942115-127942115 on Build GRCh38
5550824 c.3109T>G p.C1037G Chr.2: 127942149-127942149 on Build GRCh38
5776041 c.3100G>A p.G1034R Chr.2: 127942053-127942053 on Build GRCh38
1006349 c.2999A>G p.Q1000R Chr.2: 127942154-127942154 on Build GRCh38
1006348 c.3099C>T p.N1033N Chr.2: 127942054-127942054 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764146024 Chr.2: 127942180-127942180 on Build GRCh38
rs369335777 Chr.2: 127942208-127942208 on Build GRCh38
rs915655841 Chr.2: 127942102-127942102 on Build GRCh38
rs765351007 Chr.2: 127942196-127942196 on Build GRCh38
rs367556794 Chr.2: 127942080-127942080 on Build GRCh38
rs773513955 Chr.2: 127942224-127942224 on Build GRCh38
rs755430289 Chr.2: 127942203-127942203 on Build GRCh38
rs117858259 Chr.2: 127942195-127942195 on Build GRCh38
rs191853862 Chr.2: 127942182-127942182 on Build GRCh38
rs2304520 Chr.2: 127942202-127942202 on Build GRCh38
rs770450229 Chr.2: 127942075-127942075 on Build GRCh38
rs1041360718 Chr.2: 127942251-127942251 on Build GRCh38
rs2304519 Chr.2: 127942176-127942176 on Build GRCh38
rs748430537 Chr.2: 127942211-127942211 on Build GRCh38
rs1370229 Chr.2: 127942259-127942259 on Build GRCh38
rs758950225 Chr.2: 127942092-127942092 on Build GRCh38
rs747199615 Chr.2: 127942182-127942183 on Build GRCh38
rs750715157 Chr.2: 127942197-127942197 on Build GRCh38
rs758403332 Chr.2: 127942199-127942199 on Build GRCh38
rs140784554 Chr.2: 127942129-127942129 on Build GRCh38
rs774094805 Chr.2: 127942081-127942081 on Build GRCh38
rs767117312 Chr.2: 127942108-127942108 on Build GRCh38
rs768624695 Chr.2: 127942187-127942190 on Build GRCh38

View Full Product Details