Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6957427-6957684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPN6
Entrez Gene ID
5777
Gene Name
protein tyrosine phosphatase, non-receptor type 6
Gene Aliases
HCP, HCPH, HPTP1C, PTP-1C, SH-PTP1, SHP-1, SHP-1L, SHP1
UniGene
Hs.63489
Gene Chromosome Location
Chr.12: 6946576-6961316 on Build GRCh38
Transcripts
NM_002831.5 NM_080548.4 NM_080549.3 XM_006718994.1 XM_011520988.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6137756 c.1105G>A p.E369K Chr.12: 6957678-6957678 on Build GRCh38
4718980 c.1096C>T p.P366S Chr.12: 6957675-6957675 on Build GRCh38
6137757 c.1099G>A p.E367K Chr.12: 6957678-6957678 on Build GRCh38
6505247 c.1087C>T p.P363S Chr.12: 6957666-6957666 on Build GRCh38
4718979 c.1102C>T p.P368S Chr.12: 6957675-6957675 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377113136 Chr.12: 6957463-6957464 on Build GRCh38
rs782271177 Chr.12: 6957634-6957634 on Build GRCh38
rs374616743 Chr.12: 6957586-6957586 on Build GRCh38
rs782455038 Chr.12: 6957563-6957563 on Build GRCh38
rs781941676 Chr.12: 6957658-6957658 on Build GRCh38
rs782361392 Chr.12: 6957631-6957631 on Build GRCh38
rs782198051 Chr.12: 6957593-6957593 on Build GRCh38
rs1050329125 Chr.12: 6957598-6957598 on Build GRCh38
rs782219452 Chr.12: 6957629-6957629 on Build GRCh38
rs771890941 Chr.12: 6957591-6957591 on Build GRCh38
rs1037287633 Chr.12: 6957572-6957572 on Build GRCh38
rs1021369049 Chr.12: 6957641-6957641 on Build GRCh38
rs186735874 Chr.12: 6957637-6957637 on Build GRCh38
rs992586793 Chr.12: 6957501-6957501 on Build GRCh38
rs977344478 Chr.12: 6957452-6957452 on Build GRCh38
rs782761798 Chr.12: 6957663-6957663 on Build GRCh38
rs377374678 Chr.12: 6957483-6957483 on Build GRCh38
rs781867603 Chr.12: 6957537-6957537 on Build GRCh38
rs889945266 Chr.12: 6957605-6957605 on Build GRCh38
rs782406485 Chr.12: 6957635-6957635 on Build GRCh38
rs782540889 Chr.12: 6957486-6957486 on Build GRCh38
rs370294562 Chr.12: 6957504-6957504 on Build GRCh38
rs781783816 Chr.12: 6957562-6957562 on Build GRCh38
rs782636166 Chr.12: 6957633-6957633 on Build GRCh38
rs782587727 Chr.12: 6957553-6957553 on Build GRCh38
rs748182101 Chr.12: 6957538-6957538 on Build GRCh38
rs782578622 Chr.12: 6957615-6957615 on Build GRCh38
rs959869075 Chr.12: 6957493-6957493 on Build GRCh38
rs145915773 Chr.12: 6957638-6957638 on Build GRCh38
rs901389063 Chr.12: 6957574-6957574 on Build GRCh38
rs142240608 Chr.12: 6957539-6957539 on Build GRCh38
rs190247406 Chr.12: 6957662-6957662 on Build GRCh38
rs782322399 Chr.12: 6957640-6957640 on Build GRCh38

View Full Product Details