Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31393258-31393501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGAD
Entrez Gene ID
3681
Gene Name
integrin subunit alpha D
Gene Aliases
ADB2, CD11D
UniGene
Hs.679163
Gene Chromosome Location
Chr.16: 31393224-31426513 on Build GRCh38
Transcripts
NM_001318185.1 NM_005353.2 XM_011545835.2 XM_011545836.2 XM_011545837.2 XM_011545838.2 XM_011545839.2 XM_011545841.2 XM_011545842.2 XM_011545843.2 XM_011545844.2 XM_011545845.2 XM_011545846.2 XM_011545847.2 XM_011545848.2 XM_017023215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7198252 c.31+1_31+4delGTAA p.? Chr.16: 31393392-31393395 on Build GRCh38
3888457 c.9C>T p.F3F Chr.16: 31393369-31393369 on Build GRCh38
5500794 c.25C>T p.L9L Chr.16: 31393385-31393385 on Build GRCh38
6464351 c.31+7G>T p.? Chr.16: 31393398-31393398 on Build GRCh38
5461722 c.5C>T p.T2I Chr.16: 31393365-31393365 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773732427 Chr.16: 31393440-31393440 on Build GRCh38
rs773502631 Chr.16: 31393369-31393369 on Build GRCh38
rs557325094 Chr.16: 31393300-31393300 on Build GRCh38
rs766741737 Chr.16: 31393370-31393370 on Build GRCh38
rs779471589 Chr.16: 31393412-31393412 on Build GRCh38
rs750795204 Chr.16: 31393292-31393292 on Build GRCh38
rs751767956 Chr.16: 31393372-31393372 on Build GRCh38
rs538703511 Chr.16: 31393290-31393290 on Build GRCh38
rs201306686 Chr.16: 31393310-31393310 on Build GRCh38
rs767770744 Chr.16: 31393386-31393386 on Build GRCh38
rs746506279 Chr.16: 31393413-31393413 on Build GRCh38
rs748229323 Chr.16: 31393288-31393289 on Build GRCh38
rs769139065 Chr.16: 31393442-31393442 on Build GRCh38
rs376827502 Chr.16: 31393348-31393348 on Build GRCh38
rs752632275 Chr.16: 31393321-31393321 on Build GRCh38
rs763520017 Chr.16: 31393347-31393347 on Build GRCh38
rs756052454 Chr.16: 31393395-31393395 on Build GRCh38
rs752786427 Chr.16: 31393394-31393394 on Build GRCh38
rs777464701 Chr.16: 31393329-31393329 on Build GRCh38
rs145889371 Chr.16: 31393350-31393350 on Build GRCh38
rs759815381 Chr.16: 31393380-31393380 on Build GRCh38
rs899213465 Chr.16: 31393476-31393476 on Build GRCh38
rs568697272 Chr.16: 31393464-31393464 on Build GRCh38
rs749082859 Chr.16: 31393339-31393339 on Build GRCh38
rs771710652 Chr.16: 31393407-31393407 on Build GRCh38
rs575606728 Chr.16: 31393301-31393301 on Build GRCh38
rs374854824 Chr.16: 31393397-31393397 on Build GRCh38
rs977574966 Chr.16: 31393340-31393340 on Build GRCh38
rs1036198393 Chr.16: 31393454-31393454 on Build GRCh38
rs774968517 Chr.16: 31393351-31393351 on Build GRCh38
rs949637520 Chr.16: 31393459-31393459 on Build GRCh38
rs866824566 Chr.16: 31393374-31393374 on Build GRCh38
rs756072412 Chr.16: 31393323-31393323 on Build GRCh38
rs577566056 Chr.16: 31393466-31393466 on Build GRCh38
rs770229292 Chr.16: 31393429-31393429 on Build GRCh38
rs1009562702 Chr.16: 31393299-31393299 on Build GRCh38
rs781283115 Chr.16: 31393312-31393312 on Build GRCh38
rs1030256892 Chr.16: 31393307-31393307 on Build GRCh38
rs762253941 Chr.16: 31393284-31393284 on Build GRCh38
rs369567409 Chr.16: 31393349-31393349 on Build GRCh38
rs758588863 Chr.16: 31393295-31393295 on Build GRCh38
rs753606163 Chr.16: 31393400-31393400 on Build GRCh38
rs745579406 Chr.16: 31393406-31393406 on Build GRCh38
rs762296013 Chr.16: 31393352-31393352 on Build GRCh38
rs770305378 Chr.16: 31393353-31393353 on Build GRCh38
rs756977248 Chr.16: 31393401-31393401 on Build GRCh38
rs778548568 Chr.16: 31393402-31393402 on Build GRCh38

View Full Product Details