Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 41345761-41346034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LPIN3
Entrez Gene ID
64900
Gene Name
lipin 3
Gene Aliases
LIPN3L, SMP2
UniGene
Hs.25897
Gene Chromosome Location
Chr.20: 41340779-41360585 on Build GRCh38
Transcripts
NM_001301860.1 NR_126051.1 XM_005260516.2 XM_006723863.2 XM_011528996.1 XM_011528997.2 XM_011528998.2 XM_011528999.2 XM_011529000.1 XM_011529001.1 XM_011529003.1 XM_011529004.1 XM_011529005.2 XM_011529006.2 XM_017028020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6159729 c.131_132GG>TT p.R44L Chr.20: 41345934-41345935 on Build GRCh38
3785398 c.84C>T p.S28S Chr.20: 41345887-41345887 on Build GRCh38
6702595 c.58C>T p.R20W Chr.20: 41345861-41345861 on Build GRCh38
6702611 c.175C>T p.R59W Chr.20: 41345978-41345978 on Build GRCh38
5949927 c.130C>T p.R44W Chr.20: 41345933-41345933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777842165 Chr.20: 41345796-41345796 on Build GRCh38
rs75620173 Chr.20: 41345862-41345862 on Build GRCh38
rs778876096 Chr.20: 41345939-41345939 on Build GRCh38
rs948523587 Chr.20: 41346007-41346007 on Build GRCh38
rs377188716 Chr.20: 41345833-41345833 on Build GRCh38
rs747446012 Chr.20: 41345899-41345899 on Build GRCh38
rs772847019 Chr.20: 41345826-41345826 on Build GRCh38
rs771462288 Chr.20: 41345799-41345799 on Build GRCh38
rs143723483 Chr.20: 41345997-41345997 on Build GRCh38
rs138855546 Chr.20: 41345933-41345933 on Build GRCh38
rs755941094 Chr.20: 41345929-41345929 on Build GRCh38
rs760478064 Chr.20: 41345829-41345830 on Build GRCh38
rs770415979 Chr.20: 41345955-41345955 on Build GRCh38
rs772358991 Chr.20: 41345811-41345811 on Build GRCh38
rs970215211 Chr.20: 41346001-41346001 on Build GRCh38
rs781342933 Chr.20: 41345781-41345781 on Build GRCh38
rs41277014 Chr.20: 41345944-41345944 on Build GRCh38
rs766319923 Chr.20: 41345831-41345831 on Build GRCh38
rs763885142 Chr.20: 41345978-41345978 on Build GRCh38
rs760410304 Chr.20: 41345976-41345976 on Build GRCh38
rs368891655 Chr.20: 41345845-41345845 on Build GRCh38
rs781423258 Chr.20: 41345995-41345995 on Build GRCh38
rs61730992 Chr.20: 41345827-41345827 on Build GRCh38
rs775986460 Chr.20: 41345890-41345890 on Build GRCh38
rs779438805 Chr.20: 41345872-41345872 on Build GRCh38
rs765836868 Chr.20: 41345909-41345909 on Build GRCh38
rs199630486 Chr.20: 41345903-41345903 on Build GRCh38
rs774756851 Chr.20: 41345802-41345802 on Build GRCh38
rs765565818 Chr.20: 41345982-41345982 on Build GRCh38
rs761930233 Chr.20: 41345980-41345980 on Build GRCh38
rs767500365 Chr.20: 41345923-41345923 on Build GRCh38
rs372455946 Chr.20: 41345855-41345855 on Build GRCh38
rs201955942 Chr.20: 41345972-41345972 on Build GRCh38
rs41277016 Chr.20: 41345984-41345984 on Build GRCh38
rs140565889 Chr.20: 41345844-41345844 on Build GRCh38
rs370587902 Chr.20: 41345996-41345996 on Build GRCh38
rs139051268 Chr.20: 41345979-41345979 on Build GRCh38
rs377187175 Chr.20: 41345922-41345922 on Build GRCh38
rs765043169 Chr.20: 41345822-41345822 on Build GRCh38
rs770772409 Chr.20: 41345908-41345908 on Build GRCh38
rs370435400 Chr.20: 41345950-41345950 on Build GRCh38
rs755326841 Chr.20: 41345993-41345993 on Build GRCh38
rs146667115 Chr.20: 41345813-41345813 on Build GRCh38
rs774490083 Chr.20: 41345804-41345810 on Build GRCh38
rs780659323 Chr.20: 41345985-41345985 on Build GRCh38
rs16985673 Chr.20: 41345842-41345842 on Build GRCh38
rs772509983 Chr.20: 41345888-41345888 on Build GRCh38
rs144051050 Chr.20: 41345832-41345832 on Build GRCh38
rs759561120 Chr.20: 41345975-41345976 on Build GRCh38
rs754631598 Chr.20: 41345971-41345971 on Build GRCh38
rs748896618 Chr.20: 41345954-41345954 on Build GRCh38
rs1053057566 Chr.20: 41345801-41345801 on Build GRCh38
rs767734329 Chr.20: 41345900-41345900 on Build GRCh38
rs142640782 Chr.20: 41345940-41345940 on Build GRCh38
rs748399491 Chr.20: 41345785-41345785 on Build GRCh38
rs749383919 Chr.20: 41345798-41345798 on Build GRCh38
rs188502586 Chr.20: 41345924-41345924 on Build GRCh38
rs545382844 Chr.20: 41345918-41345918 on Build GRCh38
rs376560998 Chr.20: 41345983-41345983 on Build GRCh38
rs759112424 Chr.20: 41345966-41345966 on Build GRCh38
rs752143630 Chr.20: 41345992-41345992 on Build GRCh38
rs548778766 Chr.20: 41346002-41346002 on Build GRCh38
rs769797474 Chr.20: 41345786-41345786 on Build GRCh38
rs773719321 Chr.20: 41345957-41345957 on Build GRCh38
rs373805814 Chr.20: 41345951-41345951 on Build GRCh38
rs201841300 Chr.20: 41345934-41345934 on Build GRCh38
rs41277012 Chr.20: 41345812-41345812 on Build GRCh38
rs201526389 Chr.20: 41345874-41345874 on Build GRCh38
rs758972856 Chr.20: 41345941-41345941 on Build GRCh38
rs776288770 Chr.20: 41345968-41345968 on Build GRCh38
rs761934621 Chr.20: 41345887-41345887 on Build GRCh38
rs150453522 Chr.20: 41345861-41345861 on Build GRCh38

View Full Product Details