Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 9039599-9039848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC2A5
Entrez Gene ID
6518
Gene Name
solute carrier family 2 member 5
Gene Aliases
GLUT-5, GLUT5
UniGene
Hs.530003
Gene Chromosome Location
Chr.1: 9036941-9093407 on Build GRCh38
Transcripts
NM_003039.2 XM_005263491.3 XM_017002133.1 XM_017002134.1 XM_017002135.1 XM_017002136.1 XM_017002137.1 XM_017002138.1 XM_017002139.1 XM_017002140.1 XM_017002141.1 XM_017002142.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6506891 c.859G>T p.G287C Chr.1: 9039826-9039826 on Build GRCh38
1344803 c.877G>A p.V293I Chr.1: 9039808-9039808 on Build GRCh38
3493268 c.840C>T p.S280S Chr.1: 9039845-9039845 on Build GRCh38
7301372 c.939C>T p.H313H Chr.1: 9039609-9039609 on Build GRCh38
6781371 c.949G>A p.V317M Chr.1: 9039599-9039599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745684759 Chr.1: 9039801-9039801 on Build GRCh38
rs371249455 Chr.1: 9039635-9039635 on Build GRCh38
rs752225785 Chr.1: 9039769-9039769 on Build GRCh38
rs576402186 Chr.1: 9039671-9039671 on Build GRCh38
rs576666332 Chr.1: 9039648-9039648 on Build GRCh38
rs754230868 Chr.1: 9039714-9039714 on Build GRCh38
rs200136395 Chr.1: 9039790-9039790 on Build GRCh38
rs941009948 Chr.1: 9039621-9039621 on Build GRCh38
rs779890796 Chr.1: 9039826-9039826 on Build GRCh38
rs970683909 Chr.1: 9039634-9039634 on Build GRCh38
rs924210151 Chr.1: 9039646-9039646 on Build GRCh38
rs759769831 Chr.1: 9039756-9039756 on Build GRCh38
rs778387531 Chr.1: 9039618-9039618 on Build GRCh38
rs764956075 Chr.1: 9039760-9039760 on Build GRCh38
rs758065939 Chr.1: 9039770-9039770 on Build GRCh38
rs910856917 Chr.1: 9039619-9039619 on Build GRCh38
rs769897 Chr.1: 9039668-9039668 on Build GRCh38
rs368017715 Chr.1: 9039687-9039687 on Build GRCh38
rs574449436 Chr.1: 9039779-9039779 on Build GRCh38
rs897095346 Chr.1: 9039713-9039713 on Build GRCh38
rs13306769 Chr.1: 9039696-9039696 on Build GRCh38
rs537378085 Chr.1: 9039666-9039666 on Build GRCh38
rs918367474 Chr.1: 9039631-9039631 on Build GRCh38
rs766840134 Chr.1: 9039705-9039705 on Build GRCh38
rs772105932 Chr.1: 9039698-9039698 on Build GRCh38
rs773317655 Chr.1: 9039827-9039827 on Build GRCh38
rs751093090 Chr.1: 9039788-9039788 on Build GRCh38
rs757787611 Chr.1: 9039623-9039623 on Build GRCh38
rs932393603 Chr.1: 9039678-9039678 on Build GRCh38
rs959196431 Chr.1: 9039761-9039761 on Build GRCh38
rs936925789 Chr.1: 9039650-9039650 on Build GRCh38
rs761217627 Chr.1: 9039704-9039704 on Build GRCh38
rs768959238 Chr.1: 9039675-9039675 on Build GRCh38
rs367787656 Chr.1: 9039791-9039791 on Build GRCh38
rs995408895 Chr.1: 9039759-9039759 on Build GRCh38
rs747715661 Chr.1: 9039622-9039622 on Build GRCh38
rs747010390 Chr.1: 9039629-9039629 on Build GRCh38
rs769421798 Chr.1: 9039813-9039813 on Build GRCh38

View Full Product Details