Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108141462-108141719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAT1
Entrez Gene ID
38
Gene Name
acetyl-CoA acetyltransferase 1
Gene Aliases
ACAT, MAT, T2, THIL
UniGene
Hs.232375
Gene Chromosome Location
Chr.11: 108121531-108148168 on Build GRCh38
Transcripts
NM_000019.3 XM_006718835.3 XM_017017681.1 XM_017017682.1 XM_017017683.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3699970 c.767A>C p.Y256S Chr.11: 108141641-108141641 on Build GRCh38
7353903 c.816G>T p.Q272H Chr.11: 108141690-108141690 on Build GRCh38
5516587 c.752_754delAAG p.E252delE Chr.11: 108141626-108141628 on Build GRCh38
1181554 c.772C>T p.R258C Chr.11: 108141646-108141646 on Build GRCh38
6213028 c.741T>A p.D247E Chr.11: 108141615-108141615 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765961551 Chr.11: 108141626-108141628 on Build GRCh38
rs753816946 Chr.11: 108141646-108141646 on Build GRCh38
rs753583314 Chr.11: 108141551-108141551 on Build GRCh38
rs370652435 Chr.11: 108141644-108141644 on Build GRCh38
rs954526075 Chr.11: 108141616-108141616 on Build GRCh38
rs762416339 Chr.11: 108141563-108141566 on Build GRCh38
rs768923639 Chr.11: 108141684-108141684 on Build GRCh38
rs747309345 Chr.11: 108141686-108141686 on Build GRCh38
rs190607067 Chr.11: 108141526-108141526 on Build GRCh38
rs373300665 Chr.11: 108141556-108141556 on Build GRCh38
rs11212523 Chr.11: 108141522-108141522 on Build GRCh38
rs761192857 Chr.11: 108141565-108141565 on Build GRCh38
rs794727893 Chr.11: 108141639-108141639 on Build GRCh38
rs752216720 Chr.11: 108141603-108141603 on Build GRCh38
rs747071980 Chr.11: 108141664-108141664 on Build GRCh38
rs542087102 Chr.11: 108141647-108141647 on Build GRCh38
rs751053463 Chr.11: 108141634-108141636 on Build GRCh38
rs755683200 Chr.11: 108141627-108141627 on Build GRCh38
rs869312946 Chr.11: 108141631-108141631 on Build GRCh38
rs762262012 Chr.11: 108141576-108141576 on Build GRCh38
rs983216159 Chr.11: 108141633-108141633 on Build GRCh38
rs1031095816 Chr.11: 108141512-108141512 on Build GRCh38
rs905197066 Chr.11: 108141489-108141489 on Build GRCh38
rs746059964 Chr.11: 108141650-108141650 on Build GRCh38
rs1056468321 Chr.11: 108141544-108141544 on Build GRCh38
rs780221377 Chr.11: 108141662-108141662 on Build GRCh38
rs563370385 Chr.11: 108141658-108141658 on Build GRCh38
rs112489357 Chr.11: 108141577-108141577 on Build GRCh38
rs778592070 Chr.11: 108141648-108141648 on Build GRCh38
rs777334733 Chr.11: 108141571-108141571 on Build GRCh38
rs999582929 Chr.11: 108141509-108141509 on Build GRCh38
rs1011451895 Chr.11: 108141529-108141529 on Build GRCh38
rs574669264 Chr.11: 108141599-108141599 on Build GRCh38
rs759144164 Chr.11: 108141590-108141590 on Build GRCh38
rs794727894 Chr.11: 108141640-108141640 on Build GRCh38

View Full Product Details