Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4208647-4208918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD24
Entrez Gene ID
170961
Gene Name
ankyrin repeat domain 24
Gene Aliases
-
UniGene
Hs.197872
Gene Chromosome Location
Chr.19: 4182618-4224814 on Build GRCh38
Transcripts
NM_133475.1 XM_011527756.2 XM_011527757.2 XM_011527758.2 XM_011527761.1 XM_011527764.2 XM_011527765.2 XM_017026438.1 XM_017026439.1 XM_017026440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
997053 c.537G>A p.A179A Chr.19: 4208792-4208792 on Build GRCh38
997051 c.861G>A p.A287A Chr.19: 4208792-4208792 on Build GRCh38
1318316 c.860C>T p.A287V Chr.19: 4208791-4208791 on Build GRCh38
997052 c.1131G>A p.A377A Chr.19: 4208792-4208792 on Build GRCh38
1318318 c.536C>T p.A179V Chr.19: 4208791-4208791 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760745612 Chr.19: 4208778-4208778 on Build GRCh38
rs755701005 Chr.19: 4208818-4208818 on Build GRCh38
rs368869089 Chr.19: 4208675-4208675 on Build GRCh38
rs775537168 Chr.19: 4208764-4208764 on Build GRCh38
rs367858704 Chr.19: 4208721-4208721 on Build GRCh38
rs990243221 Chr.19: 4208772-4208772 on Build GRCh38
rs372451476 Chr.19: 4208792-4208792 on Build GRCh38
rs768805818 Chr.19: 4208779-4208779 on Build GRCh38
rs772621169 Chr.19: 4208724-4208724 on Build GRCh38
rs970646032 Chr.19: 4208669-4208669 on Build GRCh38
rs117198027 Chr.19: 4208791-4208791 on Build GRCh38
rs748935041 Chr.19: 4208673-4208673 on Build GRCh38
rs747332164 Chr.19: 4208813-4208813 on Build GRCh38
rs568712628 Chr.19: 4208849-4208849 on Build GRCh38
rs756977635 Chr.19: 4208848-4208848 on Build GRCh38
rs889280629 Chr.19: 4208855-4208855 on Build GRCh38
rs956290652 Chr.19: 4208676-4208676 on Build GRCh38
rs369594610 Chr.19: 4208782-4208782 on Build GRCh38
rs980712621 Chr.19: 4208681-4208681 on Build GRCh38
rs868290834 Chr.19: 4208786-4208786 on Build GRCh38
rs990388168 Chr.19: 4208688-4208688 on Build GRCh38
rs762765184 Chr.19: 4208787-4208787 on Build GRCh38
rs767627880 Chr.19: 4208800-4208800 on Build GRCh38
rs914563353 Chr.19: 4208694-4208694 on Build GRCh38
rs753464599 Chr.19: 4208830-4208830 on Build GRCh38
rs770512940 Chr.19: 4208720-4208720 on Build GRCh38
rs552044280 Chr.19: 4208805-4208805 on Build GRCh38
rs777235354 Chr.19: 4208846-4208846 on Build GRCh38
rs371819536 Chr.19: 4208730-4208730 on Build GRCh38
rs372618819 Chr.19: 4208828-4208828 on Build GRCh38

View Full Product Details