Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 74965570-74965821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPCS2
Entrez Gene ID
9789
Gene Name
signal peptidase complex subunit 2
Gene Aliases
-
UniGene
Hs.282700
Gene Chromosome Location
Chr.11: 74949247-74979031 on Build GRCh38
Transcripts
NM_014752.2

SNP Detail

SNP ID Chromosome Location
rs776223048 Chr.11: 74965731-74965731 on Build GRCh38
rs1022778734 Chr.11: 74965600-74965600 on Build GRCh38
rs928323586 Chr.11: 74965662-74965662 on Build GRCh38
rs765057722 Chr.11: 74965789-74965789 on Build GRCh38
rs988311767 Chr.11: 74965643-74965643 on Build GRCh38
rs968125933 Chr.11: 74965677-74965677 on Build GRCh38
rs562115770 Chr.11: 74965736-74965736 on Build GRCh38
rs745538503 Chr.11: 74965733-74965733 on Build GRCh38
rs187578102 Chr.11: 74965790-74965790 on Build GRCh38
rs514534 Chr.11: 74965749-74965749 on Build GRCh38
rs754066700 Chr.11: 74965783-74965783 on Build GRCh38
rs760638756 Chr.11: 74965766-74965766 on Build GRCh38
rs1018775183 Chr.11: 74965611-74965611 on Build GRCh38
rs926245344 Chr.11: 74965718-74965718 on Build GRCh38
rs770025908 Chr.11: 74965758-74965758 on Build GRCh38
rs370914590 Chr.11: 74965685-74965685 on Build GRCh38
rs542264236 Chr.11: 74965678-74965678 on Build GRCh38
rs185019099 Chr.11: 74965626-74965626 on Build GRCh38
rs548226933 Chr.11: 74965727-74965727 on Build GRCh38
rs777837005 Chr.11: 74965728-74965728 on Build GRCh38
rs770526117 Chr.11: 74965730-74965730 on Build GRCh38
rs541577774 Chr.11: 74965785-74965785 on Build GRCh38
rs746490806 Chr.11: 74965729-74965729 on Build GRCh38
rs937585616 Chr.11: 74965725-74965725 on Build GRCh38
rs774698482 Chr.11: 74965737-74965737 on Build GRCh38
rs1020672035 Chr.11: 74965675-74965675 on Build GRCh38
rs768763248 Chr.11: 74965705-74965705 on Build GRCh38
rs979081695 Chr.11: 74965708-74965708 on Build GRCh38
rs920159264 Chr.11: 74965688-74965688 on Build GRCh38
rs981152947 Chr.11: 74965621-74965621 on Build GRCh38
rs748040776 Chr.11: 74965720-74965720 on Build GRCh38
rs754782108 Chr.11: 74965715-74965715 on Build GRCh38
rs762305174 Chr.11: 74965745-74965745 on Build GRCh38
rs543306138 Chr.11: 74965773-74965773 on Build GRCh38
rs969808081 Chr.11: 74965604-74965604 on Build GRCh38
rs752318651 Chr.11: 74965675-74965675 on Build GRCh38

View Full Product Details