Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 112503955-112504226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUBGCP3
Entrez Gene ID
10426
Gene Name
tubulin gamma complex associated protein 3
Gene Aliases
104p, GCP3, Grip104, SPBC98, Spc98, Spc98p
UniGene
Hs.224152
Gene Chromosome Location
Chr.13: 112485005-112606469 on Build GRCh38
Transcripts
NM_001286277.1 NM_001286278.1 NM_006322.5 XM_005268293.1 XM_011537462.2 XM_011537463.2 XM_011537464.2 XM_017020323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4760243 c.2276T>C p.I759T Chr.13: 112504063-112504063 on Build GRCh38
695974 c.2282G>T p.R761L Chr.13: 112504057-112504057 on Build GRCh38
1365524 c.2253C>T p.H751H Chr.13: 112504086-112504086 on Build GRCh38
355032 c.2217G>C p.Q739H Chr.13: 112504122-112504122 on Build GRCh38
4842092 c.2205G>C p.W735C Chr.13: 112504134-112504134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754889683 Chr.13: 112504106-112504106 on Build GRCh38
rs781219529 Chr.13: 112504010-112504010 on Build GRCh38
rs779744188 Chr.13: 112503994-112503994 on Build GRCh38
rs769584279 Chr.13: 112504197-112504197 on Build GRCh38
rs779974511 Chr.13: 112504001-112504001 on Build GRCh38
rs756337745 Chr.13: 112504158-112504158 on Build GRCh38
rs1050351973 Chr.13: 112504117-112504117 on Build GRCh38
rs746768027 Chr.13: 112504170-112504172 on Build GRCh38
rs748419852 Chr.13: 112504013-112504013 on Build GRCh38
rs371232252 Chr.13: 112504186-112504186 on Build GRCh38
rs940888988 Chr.13: 112504058-112504058 on Build GRCh38
rs377470399 Chr.13: 112504022-112504022 on Build GRCh38
rs202245851 Chr.13: 112504021-112504021 on Build GRCh38
rs964905515 Chr.13: 112503980-112503980 on Build GRCh38
rs774983294 Chr.13: 112504041-112504041 on Build GRCh38
rs761575204 Chr.13: 112504072-112504072 on Build GRCh38
rs778141776 Chr.13: 112504019-112504019 on Build GRCh38
rs751579089 Chr.13: 112504093-112504093 on Build GRCh38
rs768319104 Chr.13: 112504057-112504057 on Build GRCh38
rs758298576 Chr.13: 112503992-112503992 on Build GRCh38
rs374347551 Chr.13: 112504015-112504015 on Build GRCh38
rs752735566 Chr.13: 112504128-112504128 on Build GRCh38
rs201106975 Chr.13: 112504180-112504180 on Build GRCh38
rs751603637 Chr.13: 112504005-112504005 on Build GRCh38
rs764939077 Chr.13: 112504085-112504085 on Build GRCh38
rs199549355 Chr.13: 112504187-112504187 on Build GRCh38
rs777997746 Chr.13: 112504166-112504166 on Build GRCh38
rs762750913 Chr.13: 112504086-112504086 on Build GRCh38
rs781272584 Chr.13: 112504107-112504107 on Build GRCh38
rs779113123 Chr.13: 112504183-112504183 on Build GRCh38
rs776256523 Chr.13: 112504060-112504060 on Build GRCh38
rs747594842 Chr.13: 112504194-112504194 on Build GRCh38
rs145266974 Chr.13: 112504007-112504007 on Build GRCh38
rs759960025 Chr.13: 112504043-112504043 on Build GRCh38

View Full Product Details