Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 66448734-66448967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADHFE1
Entrez Gene ID
137872
Gene Name
alcohol dehydrogenase, iron containing 1
Gene Aliases
ADH8, HMFT2263, HOT
UniGene
Hs.720023
Gene Chromosome Location
Chr.8: 66432483-66468809 on Build GRCh38
Transcripts
NM_144650.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5904309 c.674C>T p.P225L Chr.8: 66448910-66448910 on Build GRCh38
1244512 c.709G>A p.A237T Chr.8: 66448945-66448945 on Build GRCh38
7126363 c.639G>A p.S213S Chr.8: 66448875-66448875 on Build GRCh38
6113579 c.689A>T p.H230L Chr.8: 66448925-66448925 on Build GRCh38
32509 c.581A>T p.D194V Chr.8: 66448961-66448961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924317883 Chr.8: 66448796-66448796 on Build GRCh38
rs776461759 Chr.8: 66448762-66448762 on Build GRCh38
rs12544088 Chr.8: 66448783-66448783 on Build GRCh38
rs756683160 Chr.8: 66448865-66448865 on Build GRCh38
rs1041644132 Chr.8: 66448835-66448835 on Build GRCh38
rs775542691 Chr.8: 66448819-66448819 on Build GRCh38
rs776500463 Chr.8: 66448910-66448910 on Build GRCh38
rs187158237 Chr.8: 66448894-66448894 on Build GRCh38
rs766239380 Chr.8: 66448937-66448937 on Build GRCh38
rs752008861 Chr.8: 66448947-66448948 on Build GRCh38
rs577285802 Chr.8: 66448904-66448904 on Build GRCh38
rs372547396 Chr.8: 66448877-66448877 on Build GRCh38
rs755106872 Chr.8: 66448855-66448855 on Build GRCh38
rs62511186 Chr.8: 66448837-66448837 on Build GRCh38
rs763974255 Chr.8: 66448829-66448829 on Build GRCh38
rs913088633 Chr.8: 66448924-66448924 on Build GRCh38
rs781299907 Chr.8: 66448857-66448857 on Build GRCh38
rs536915554 Chr.8: 66448853-66448853 on Build GRCh38
rs866457584 Chr.8: 66448946-66448947 on Build GRCh38
rs375449644 Chr.8: 66448827-66448827 on Build GRCh38
rs766462372 Chr.8: 66448927-66448927 on Build GRCh38
rs748624079 Chr.8: 66448861-66448861 on Build GRCh38
rs369269266 Chr.8: 66448831-66448831 on Build GRCh38
rs762882078 Chr.8: 66448914-66448914 on Build GRCh38
rs199559170 Chr.8: 66448945-66448945 on Build GRCh38
rs771673334 Chr.8: 66448886-66448886 on Build GRCh38
rs1004191376 Chr.8: 66448902-66448902 on Build GRCh38
rs61743396 Chr.8: 66448871-66448871 on Build GRCh38
rs767582148 Chr.8: 66448939-66448939 on Build GRCh38
rs775204738 Chr.8: 66448893-66448893 on Build GRCh38
rs62511187 Chr.8: 66448841-66448841 on Build GRCh38
rs778098866 Chr.8: 66448867-66448867 on Build GRCh38
rs368995778 Chr.8: 66448874-66448874 on Build GRCh38
rs751783445 Chr.8: 66448936-66448936 on Build GRCh38
rs745324789 Chr.8: 66448878-66448878 on Build GRCh38

View Full Product Details