Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58338670-58338899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN22
Entrez Gene ID
342945
Gene Name
zinc finger and SCAN domain containing 22
Gene Aliases
HKR2, ZNF50
UniGene
Hs.388162
Gene Chromosome Location
Chr.19: 58327000-58342346 on Build GRCh38
Transcripts
NM_001321116.1 NM_001321117.1 NM_181846.2 XM_006723192.3 XM_011526917.2 XM_011526918.2 XM_017026746.1 XM_017026747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
386996 c.1004G>A p.R335Q Chr.19: 58338854-58338854 on Build GRCh38
6834330 c.821G>T p.R274M Chr.19: 58338671-58338671 on Build GRCh38
5458797 c.849C>T p.L283L Chr.19: 58338699-58338699 on Build GRCh38
6086217 c.963G>A p.A321A Chr.19: 58338813-58338813 on Build GRCh38
7399886 c.988G>C p.G330R Chr.19: 58338838-58338838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751497329 Chr.19: 58338819-58338819 on Build GRCh38
rs768089290 Chr.19: 58338845-58338845 on Build GRCh38
rs753931190 Chr.19: 58338871-58338871 on Build GRCh38
rs777374992 Chr.19: 58338747-58338747 on Build GRCh38
rs368284797 Chr.19: 58338699-58338699 on Build GRCh38
rs149655686 Chr.19: 58338869-58338869 on Build GRCh38
rs568354675 Chr.19: 58338784-58338784 on Build GRCh38
rs1056736041 Chr.19: 58338843-58338843 on Build GRCh38
rs139283341 Chr.19: 58338813-58338813 on Build GRCh38
rs375935212 Chr.19: 58338695-58338695 on Build GRCh38
rs745575034 Chr.19: 58338769-58338769 on Build GRCh38
rs750661237 Chr.19: 58338797-58338797 on Build GRCh38
rs761045526 Chr.19: 58338801-58338801 on Build GRCh38
rs146020373 Chr.19: 58338821-58338821 on Build GRCh38
rs554590877 Chr.19: 58338707-58338707 on Build GRCh38
rs556783304 Chr.19: 58338849-58338849 on Build GRCh38
rs758415041 Chr.19: 58338733-58338733 on Build GRCh38
rs144167717 Chr.19: 58338817-58338817 on Build GRCh38
rs1025815561 Chr.19: 58338738-58338738 on Build GRCh38
rs762209131 Chr.19: 58338783-58338783 on Build GRCh38
rs756244183 Chr.19: 58338724-58338724 on Build GRCh38
rs763391851 Chr.19: 58338777-58338777 on Build GRCh38
rs770669020 Chr.19: 58338761-58338761 on Build GRCh38
rs773720957 Chr.19: 58338858-58338858 on Build GRCh38
rs766504588 Chr.19: 58338810-58338810 on Build GRCh38
rs769449537 Chr.19: 58338823-58338823 on Build GRCh38
rs769285873 Chr.19: 58338770-58338770 on Build GRCh38
rs780874744 Chr.19: 58338762-58338762 on Build GRCh38
rs373749935 Chr.19: 58338748-58338748 on Build GRCh38
rs372290763 Chr.19: 58338776-58338776 on Build GRCh38
rs537926318 Chr.19: 58338696-58338696 on Build GRCh38
rs751963267 Chr.19: 58338700-58338700 on Build GRCh38
rs60710360 Chr.19: 58338812-58338812 on Build GRCh38
rs780342708 Chr.19: 58338725-58338725 on Build GRCh38
rs748093868 Chr.19: 58338730-58338730 on Build GRCh38
rs762285917 Chr.19: 58338853-58338853 on Build GRCh38
rs766771221 Chr.19: 58338854-58338854 on Build GRCh38
rs763518054 Chr.19: 58338698-58338698 on Build GRCh38
rs767810106 Chr.19: 58338708-58338708 on Build GRCh38
rs953033778 Chr.19: 58338746-58338746 on Build GRCh38
rs750654412 Chr.19: 58338723-58338723 on Build GRCh38
rs577439792 Chr.19: 58338827-58338827 on Build GRCh38
rs200660868 Chr.19: 58338693-58338693 on Build GRCh38
rs774537954 Chr.19: 58338780-58338780 on Build GRCh38
rs760926419 Chr.19: 58338861-58338861 on Build GRCh38
rs964107090 Chr.19: 58338752-58338752 on Build GRCh38
rs62116373 Chr.19: 58338840-58338840 on Build GRCh38
rs148314941 Chr.19: 58338692-58338692 on Build GRCh38

View Full Product Details