Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 69414942-69415197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TACR2
Entrez Gene ID
6865
Gene Name
tachykinin receptor 2
Gene Aliases
NK2R, NKNAR, SKR, TAC2R
UniGene
Hs.88372
Gene Chromosome Location
Chr.10: 69404202-69416918 on Build GRCh38
Transcripts
NM_001057.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6307347 c.491C>T p.S164F Chr.10: 69415041-69415041 on Build GRCh38
4731898 c.530G>A p.G177D Chr.10: 69415002-69415002 on Build GRCh38
7392907 c.578C>T p.T193M Chr.10: 69414954-69414954 on Build GRCh38
5803947 c.521T>A p.M174K Chr.10: 69415011-69415011 on Build GRCh38
2160094 c.579G>A p.T193T Chr.10: 69414953-69414953 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752971766 Chr.10: 69415168-69415168 on Build GRCh38
rs201152189 Chr.10: 69415081-69415081 on Build GRCh38
rs913231363 Chr.10: 69415127-69415127 on Build GRCh38
rs148031991 Chr.10: 69415051-69415051 on Build GRCh38
rs773150827 Chr.10: 69414986-69414986 on Build GRCh38
rs199608050 Chr.10: 69415107-69415107 on Build GRCh38
rs769929557 Chr.10: 69414975-69414975 on Build GRCh38
rs752073460 Chr.10: 69415161-69415161 on Build GRCh38
rs200692704 Chr.10: 69415108-69415108 on Build GRCh38
rs781477753 Chr.10: 69415058-69415058 on Build GRCh38
rs747354247 Chr.10: 69414963-69414963 on Build GRCh38
rs371826214 Chr.10: 69415099-69415099 on Build GRCh38
rs535568032 Chr.10: 69415144-69415144 on Build GRCh38
rs777877576 Chr.10: 69415171-69415171 on Build GRCh38
rs528407251 Chr.10: 69414990-69414990 on Build GRCh38
rs200987720 Chr.10: 69414973-69414973 on Build GRCh38
rs141827389 Chr.10: 69414967-69414967 on Build GRCh38
rs759381167 Chr.10: 69415140-69415141 on Build GRCh38
rs561960711 Chr.10: 69415004-69415004 on Build GRCh38
rs752022242 Chr.10: 69415020-69415020 on Build GRCh38
rs781424764 Chr.10: 69414970-69414970 on Build GRCh38
rs753977676 Chr.10: 69415038-69415038 on Build GRCh38
rs200975784 Chr.10: 69415019-69415019 on Build GRCh38
rs372622967 Chr.10: 69415117-69415117 on Build GRCh38
rs970785006 Chr.10: 69415052-69415052 on Build GRCh38
rs201054804 Chr.10: 69415111-69415111 on Build GRCh38
rs753570963 Chr.10: 69415132-69415132 on Build GRCh38
rs201476389 Chr.10: 69415135-69415135 on Build GRCh38
rs923828091 Chr.10: 69415074-69415074 on Build GRCh38
rs200556691 Chr.10: 69415164-69415164 on Build GRCh38
rs141759934 Chr.10: 69415065-69415065 on Build GRCh38
rs765539277 Chr.10: 69415080-69415080 on Build GRCh38
rs761569938 Chr.10: 69415120-69415120 on Build GRCh38
rs752739560 Chr.10: 69415151-69415152 on Build GRCh38
rs968709176 Chr.10: 69415151-69415151 on Build GRCh38
rs200230426 Chr.10: 69415066-69415066 on Build GRCh38
rs764775979 Chr.10: 69415142-69415142 on Build GRCh38
rs199825032 Chr.10: 69415154-69415154 on Build GRCh38
rs201322308 Chr.10: 69415096-69415096 on Build GRCh38
rs202196664 Chr.10: 69414989-69414989 on Build GRCh38
rs1015973683 Chr.10: 69414964-69414964 on Build GRCh38
rs760845670 Chr.10: 69414966-69414967 on Build GRCh38
rs762402134 Chr.10: 69415148-69415148 on Build GRCh38
rs376335183 Chr.10: 69415169-69415169 on Build GRCh38
rs755464752 Chr.10: 69415163-69415163 on Build GRCh38
rs964555719 Chr.10: 69414974-69414974 on Build GRCh38
rs201947329 Chr.10: 69415165-69415165 on Build GRCh38
rs988719658 Chr.10: 69415083-69415083 on Build GRCh38
rs760564971 Chr.10: 69414993-69414993 on Build GRCh38
rs146283790 Chr.10: 69415018-69415018 on Build GRCh38
rs761361786 Chr.10: 69415002-69415002 on Build GRCh38
rs200013426 Chr.10: 69414996-69414996 on Build GRCh38
rs757826293 Chr.10: 69415163-69415163 on Build GRCh38
rs780509295 Chr.10: 69414960-69414960 on Build GRCh38
rs768961049 Chr.10: 69414965-69414965 on Build GRCh38
rs574436938 Chr.10: 69415126-69415126 on Build GRCh38
rs758569718 Chr.10: 69414988-69414988 on Build GRCh38
rs202091754 Chr.10: 69415082-69415082 on Build GRCh38
rs200605647 Chr.10: 69415085-69415085 on Build GRCh38
rs765082792 Chr.10: 69415149-69415150 on Build GRCh38
rs766611204 Chr.10: 69415100-69415101 on Build GRCh38
rs765810504 Chr.10: 69415158-69415158 on Build GRCh38
rs749481857 Chr.10: 69415173-69415173 on Build GRCh38

View Full Product Details