Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 75816232-75816491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSH4
Entrez Gene ID
4438
Gene Name
mutS homolog 4
Gene Aliases
-
UniGene
Hs.216639
Gene Chromosome Location
Chr.1: 75796871-75913238 on Build GRCh38
Transcripts
NM_002440.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5381526 c.875C>T p.S292L Chr.1: 75816432-75816432 on Build GRCh38
4009908 c.919G>T p.E307* Chr.1: 75816476-75816476 on Build GRCh38
5934333 c.919G>A p.E307K Chr.1: 75816476-75816476 on Build GRCh38
4511446 c.868C>T p.Q290* Chr.1: 75816425-75816425 on Build GRCh38
5661380 c.933G>C p.M311I Chr.1: 75816490-75816490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773595755 Chr.1: 75816382-75816382 on Build GRCh38
rs1045275689 Chr.1: 75816314-75816314 on Build GRCh38
rs144767182 Chr.1: 75816435-75816435 on Build GRCh38
rs951730527 Chr.1: 75816274-75816274 on Build GRCh38
rs754120637 Chr.1: 75816407-75816407 on Build GRCh38
rs997571586 Chr.1: 75816434-75816434 on Build GRCh38
rs748733234 Chr.1: 75816340-75816340 on Build GRCh38
rs372643929 Chr.1: 75816336-75816336 on Build GRCh38
rs774652462 Chr.1: 75816349-75816349 on Build GRCh38
rs766562265 Chr.1: 75816399-75816399 on Build GRCh38
rs140131641 Chr.1: 75816392-75816392 on Build GRCh38
rs772370871 Chr.1: 75816364-75816364 on Build GRCh38
rs1015939576 Chr.1: 75816308-75816308 on Build GRCh38
rs750213260 Chr.1: 75816462-75816462 on Build GRCh38
rs768278883 Chr.1: 75816346-75816346 on Build GRCh38
rs756022702 Chr.1: 75816466-75816466 on Build GRCh38
rs751398721 Chr.1: 75816442-75816442 on Build GRCh38
rs759534906 Chr.1: 75816412-75816412 on Build GRCh38
rs370426879 Chr.1: 75816359-75816359 on Build GRCh38
rs143902879 Chr.1: 75816437-75816437 on Build GRCh38
rs1036503432 Chr.1: 75816345-75816345 on Build GRCh38
rs142299451 Chr.1: 75816460-75816460 on Build GRCh38
rs757241895 Chr.1: 75816454-75816454 on Build GRCh38
rs779894202 Chr.1: 75816338-75816338 on Build GRCh38
rs556902744 Chr.1: 75816312-75816312 on Build GRCh38
rs1004516833 Chr.1: 75816285-75816285 on Build GRCh38
rs909408234 Chr.1: 75816330-75816330 on Build GRCh38

View Full Product Details