Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 121758813-121759076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAB2IP
Entrez Gene ID
153090
Gene Name
DAB2 interacting protein
Gene Aliases
AF9Q34, AIP-1, AIP1, DIP1/2
UniGene
Hs.522378
Gene Chromosome Location
Chr.9: 121566883-121785530 on Build GRCh38
Transcripts
NM_032552.3 NM_138709.2 XM_005251719.4 XM_005251721.1 XM_005251724.4 XM_011518264.2 XM_011518265.2 XM_011518266.2 XM_011518267.2 XM_011518270.2 XM_011518271.2 XM_017014298.1 XM_017014299.1 XM_017014300.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7211900 c.433-3C>G p.? Chr.9: 121758895-121758895 on Build GRCh38
3323017 c.437C>T p.T146M Chr.9: 121758902-121758902 on Build GRCh38
7211901 c.145-3C>G p.? Chr.9: 121758895-121758895 on Build GRCh38
3323018 c.149C>T p.T50M Chr.9: 121758902-121758902 on Build GRCh38
7363625 c.608C>T p.P203L Chr.9: 121758989-121758989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780140648 Chr.9: 121758908-121758908 on Build GRCh38
rs770718886 Chr.9: 121759023-121759023 on Build GRCh38
rs559896738 Chr.9: 121758906-121758906 on Build GRCh38
rs369054346 Chr.9: 121758888-121758888 on Build GRCh38
rs746972369 Chr.9: 121758860-121758860 on Build GRCh38
rs768400658 Chr.9: 121758862-121758862 on Build GRCh38
rs773984806 Chr.9: 121759029-121759029 on Build GRCh38
rs758625862 Chr.9: 121758989-121758989 on Build GRCh38
rs754243563 Chr.9: 121758902-121758902 on Build GRCh38
rs780053460 Chr.9: 121759002-121759002 on Build GRCh38
rs778501675 Chr.9: 121759038-121759038 on Build GRCh38
rs758120340 Chr.9: 121759040-121759041 on Build GRCh38
rs976026810 Chr.9: 121758887-121758887 on Build GRCh38
rs775457416 Chr.9: 121758853-121758853 on Build GRCh38
rs768112950 Chr.9: 121758876-121758876 on Build GRCh38
rs1032410166 Chr.9: 121758898-121758898 on Build GRCh38
rs774000651 Chr.9: 121758973-121758973 on Build GRCh38
rs753153245 Chr.9: 121759012-121759012 on Build GRCh38
rs879577732 Chr.9: 121758892-121758892 on Build GRCh38
rs751948717 Chr.9: 121758865-121758865 on Build GRCh38
rs1008484111 Chr.9: 121758874-121758874 on Build GRCh38
rs200515341 Chr.9: 121758893-121758893 on Build GRCh38
rs746884928 Chr.9: 121758928-121758928 on Build GRCh38
rs748045477 Chr.9: 121758947-121758947 on Build GRCh38
rs190940502 Chr.9: 121758903-121758903 on Build GRCh38
rs769580713 Chr.9: 121759009-121759009 on Build GRCh38
rs765001856 Chr.9: 121758867-121758867 on Build GRCh38
rs764485096 Chr.9: 121758897-121758897 on Build GRCh38
rs187297125 Chr.9: 121758868-121758868 on Build GRCh38
rs748880997 Chr.9: 121759020-121759020 on Build GRCh38
rs769297007 Chr.9: 121758952-121758952 on Build GRCh38
rs913905979 Chr.9: 121758909-121758909 on Build GRCh38
rs377190430 Chr.9: 121759007-121759007 on Build GRCh38
rs3747851 Chr.9: 121758981-121758981 on Build GRCh38
rs780694138 Chr.9: 121758918-121758918 on Build GRCh38
rs368193656 Chr.9: 121758974-121758974 on Build GRCh38
rs892349601 Chr.9: 121758924-121758924 on Build GRCh38
rs777542550 Chr.9: 121759045-121759046 on Build GRCh38
rs1005197237 Chr.9: 121759041-121759041 on Build GRCh38
rs777456627 Chr.9: 121759018-121759018 on Build GRCh38
rs200088258 Chr.9: 121759006-121759006 on Build GRCh38
rs761730279 Chr.9: 121758980-121758980 on Build GRCh38
rs1047347979 Chr.9: 121759019-121759019 on Build GRCh38
rs750619917 Chr.9: 121758984-121758984 on Build GRCh38
rs988112588 Chr.9: 121758907-121758907 on Build GRCh38
rs771874753 Chr.9: 121758850-121758850 on Build GRCh38
rs200002701 Chr.9: 121758953-121758953 on Build GRCh38
rs776418092 Chr.9: 121758864-121758864 on Build GRCh38
rs777147346 Chr.9: 121759048-121759048 on Build GRCh38
rs747152150 Chr.9: 121759004-121759004 on Build GRCh38
rs147323804 Chr.9: 121758934-121758934 on Build GRCh38
rs753138176 Chr.9: 121758884-121758884 on Build GRCh38
rs769265042 Chr.9: 121759040-121759040 on Build GRCh38
rs767935698 Chr.9: 121758880-121758880 on Build GRCh38
rs765674461 Chr.9: 121758882-121758883 on Build GRCh38
rs776280750 Chr.9: 121758936-121758936 on Build GRCh38
rs374038020 Chr.9: 121758954-121758954 on Build GRCh38
rs114266755 Chr.9: 121758976-121758976 on Build GRCh38
rs371328283 Chr.9: 121759031-121759031 on Build GRCh38
rs765943680 Chr.9: 121758968-121758968 on Build GRCh38
rs567507053 Chr.9: 121758977-121758977 on Build GRCh38
rs762560743 Chr.9: 121758870-121758870 on Build GRCh38
rs762533050 Chr.9: 121758956-121758956 on Build GRCh38
rs780771429 Chr.9: 121758876-121758877 on Build GRCh38
rs754909939 Chr.9: 121759005-121759005 on Build GRCh38
rs370347740 Chr.9: 121758930-121758930 on Build GRCh38

View Full Product Details