Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151507289-151507534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT2
Entrez Gene ID
2196
Gene Name
FAT atypical cadherin 2
Gene Aliases
CDHF8, CDHR9, HFAT2, MEGF1
UniGene
Hs.591255
Gene Chromosome Location
Chr.5: 151504092-151594822 on Build GRCh38
Transcripts
NM_001447.2 XM_006714761.3 XM_011537600.2 XM_011537603.2 XM_017009224.1 XM_017009225.1 XM_011537595.2 XM_017009217.1 XM_017009219.1

Gene Symbol
SLC36A1
Entrez Gene ID
206358
Gene Name
solute carrier family 36 member 1
Gene Aliases
Dct1, LYAAT1, PAT1, TRAMD3
UniGene
Hs.269004
Gene Chromosome Location
Chr.5: 151411344-151556318 on Build GRCh38
Transcripts
NM_001447.2 XM_006714761.3 XM_011537600.2 XM_011537603.2 XM_017009224.1 XM_017009225.1 XM_011537595.2 XM_017009217.1 XM_017009219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4683535 c.12211C>T p.R4071C Chr.5: 151507460-151507460 on Build GRCh38
4683534 c.12218G>A p.C4073Y Chr.5: 151507453-151507453 on Build GRCh38
7098600 c.12342C>T p.L4114L Chr.5: 151507329-151507329 on Build GRCh38
3761045 c.12350C>T p.P4117L Chr.5: 151507321-151507321 on Build GRCh38
1064561 c.12215G>A p.R4072H Chr.5: 151507456-151507456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772772158 Chr.5: 151507480-151507480 on Build GRCh38
rs1105168 Chr.5: 151507321-151507321 on Build GRCh38
rs749389986 Chr.5: 151507453-151507453 on Build GRCh38
rs745784430 Chr.5: 151507343-151507343 on Build GRCh38
rs762302252 Chr.5: 151507488-151507488 on Build GRCh38
rs138705803 Chr.5: 151507351-151507351 on Build GRCh38
rs777974723 Chr.5: 151507417-151507417 on Build GRCh38
rs760967696 Chr.5: 151507506-151507506 on Build GRCh38
rs904759464 Chr.5: 151507320-151507320 on Build GRCh38
rs201825044 Chr.5: 151507404-151507404 on Build GRCh38
rs746980768 Chr.5: 151507458-151507458 on Build GRCh38
rs755238824 Chr.5: 151507400-151507400 on Build GRCh38
rs927235826 Chr.5: 151507340-151507340 on Build GRCh38
rs143801705 Chr.5: 151507479-151507479 on Build GRCh38
rs763502422 Chr.5: 151507372-151507372 on Build GRCh38
rs143949083 Chr.5: 151507505-151507505 on Build GRCh38
rs755223311 Chr.5: 151507315-151507315 on Build GRCh38
rs779535153 Chr.5: 151507407-151507407 on Build GRCh38
rs770681196 Chr.5: 151507337-151507338 on Build GRCh38
rs533240264 Chr.5: 151507337-151507337 on Build GRCh38
rs201517523 Chr.5: 151507508-151507508 on Build GRCh38
rs756209489 Chr.5: 151507333-151507333 on Build GRCh38
rs766860387 Chr.5: 151507386-151507386 on Build GRCh38
rs768938668 Chr.5: 151507447-151507447 on Build GRCh38
rs776442798 Chr.5: 151507462-151507462 on Build GRCh38
rs374289322 Chr.5: 151507456-151507456 on Build GRCh38
rs767959528 Chr.5: 151507501-151507501 on Build GRCh38
rs776208786 Chr.5: 151507355-151507355 on Build GRCh38
rs918406526 Chr.5: 151507414-151507414 on Build GRCh38
rs752111755 Chr.5: 151507391-151507391 on Build GRCh38
rs770489194 Chr.5: 151507490-151507490 on Build GRCh38
rs755763462 Chr.5: 151507392-151507392 on Build GRCh38
rs749663238 Chr.5: 151507409-151507409 on Build GRCh38
rs761476586 Chr.5: 151507359-151507359 on Build GRCh38
rs774668674 Chr.5: 151507495-151507495 on Build GRCh38
rs768447645 Chr.5: 151507353-151507353 on Build GRCh38
rs756442595 Chr.5: 151507416-151507416 on Build GRCh38
rs768392091 Chr.5: 151507460-151507460 on Build GRCh38
rs867210819 Chr.5: 151507502-151507502 on Build GRCh38
rs138553780 Chr.5: 151507500-151507500 on Build GRCh38
rs748391997 Chr.5: 151507322-151507322 on Build GRCh38
rs531281319 Chr.5: 151507403-151507403 on Build GRCh38
rs545597751 Chr.5: 151507348-151507348 on Build GRCh38
rs199556909 Chr.5: 151507457-151507457 on Build GRCh38
rs367689507 Chr.5: 151507367-151507367 on Build GRCh38
rs200773290 Chr.5: 151507360-151507360 on Build GRCh38
rs747769250 Chr.5: 151507478-151507478 on Build GRCh38

View Full Product Details