Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 124319232-124319481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR8D2
Entrez Gene ID
283160
Gene Name
olfactory receptor family 8 subfamily D member 2 (gene/pseudogene)
Gene Aliases
JCG2
UniGene
Hs.504212
Gene Chromosome Location
Chr.11: 124319262-124320197 on Build GRCh38
Transcripts
NM_001002918.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6000153 c.860C>A p.P287H Chr.11: 124319338-124319338 on Build GRCh38
3359066 c.859C>A p.P287T Chr.11: 124319339-124319339 on Build GRCh38
4561971 c.906G>A p.K302K Chr.11: 124319292-124319292 on Build GRCh38
686649 c.864A>T p.L288L Chr.11: 124319334-124319334 on Build GRCh38
686648 c.747C>A p.G249G Chr.11: 124319451-124319451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770491562 Chr.11: 124319339-124319339 on Build GRCh38
rs767892209 Chr.11: 124319398-124319398 on Build GRCh38
rs201649690 Chr.11: 124319402-124319402 on Build GRCh38
rs906280675 Chr.11: 124319330-124319330 on Build GRCh38
rs768506974 Chr.11: 124319453-124319453 on Build GRCh38
rs370641158 Chr.11: 124319424-124319424 on Build GRCh38
rs762882611 Chr.11: 124319378-124319378 on Build GRCh38
rs745554869 Chr.11: 124319345-124319345 on Build GRCh38
rs755327113 Chr.11: 124319308-124319308 on Build GRCh38
rs200457402 Chr.11: 124319393-124319393 on Build GRCh38
rs756684360 Chr.11: 124319431-124319431 on Build GRCh38
rs1019079301 Chr.11: 124319412-124319412 on Build GRCh38
rs139150329 Chr.11: 124319347-124319347 on Build GRCh38
rs763157502 Chr.11: 124319282-124319282 on Build GRCh38
rs781713212 Chr.11: 124319312-124319312 on Build GRCh38
rs776685001 Chr.11: 124319377-124319377 on Build GRCh38
rs947907120 Chr.11: 124319263-124319263 on Build GRCh38
rs779744609 Chr.11: 124319443-124319443 on Build GRCh38
rs201080079 Chr.11: 124319346-124319346 on Build GRCh38
rs200162903 Chr.11: 124319317-124319317 on Build GRCh38
rs74550564 Chr.11: 124319396-124319396 on Build GRCh38
rs750435044 Chr.11: 124319261-124319261 on Build GRCh38
rs898456544 Chr.11: 124319428-124319428 on Build GRCh38
rs758011345 Chr.11: 124319439-124319439 on Build GRCh38
rs368245211 Chr.11: 124319444-124319444 on Build GRCh38
rs769058169 Chr.11: 124319288-124319288 on Build GRCh38
rs966121755 Chr.11: 124319430-124319430 on Build GRCh38
rs145955830 Chr.11: 124319363-124319363 on Build GRCh38
rs778472925 Chr.11: 124319343-124319343 on Build GRCh38
rs2466620 Chr.11: 124319410-124319410 on Build GRCh38
rs773832849 Chr.11: 124319381-124319392 on Build GRCh38
rs150761485 Chr.11: 124319323-124319323 on Build GRCh38
rs897466353 Chr.11: 124319349-124319349 on Build GRCh38
rs762224579 Chr.11: 124319333-124319333 on Build GRCh38
rs748735376 Chr.11: 124319328-124319328 on Build GRCh38
rs533604016 Chr.11: 124319294-124319294 on Build GRCh38
rs745434790 Chr.11: 124319437-124319437 on Build GRCh38
rs777132806 Chr.11: 124319288-124319290 on Build GRCh38
rs778412692 Chr.11: 124319324-124319324 on Build GRCh38
rs751843408 Chr.11: 124319307-124319307 on Build GRCh38
rs754376662 Chr.11: 124319429-124319429 on Build GRCh38
rs775018205 Chr.11: 124319285-124319287 on Build GRCh38
rs374312619 Chr.11: 124319435-124319435 on Build GRCh38
rs201126939 Chr.11: 124319306-124319306 on Build GRCh38
rs765975859 Chr.11: 124319365-124319366 on Build GRCh38
rs778421563 Chr.11: 124319433-124319433 on Build GRCh38

View Full Product Details