Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154460128-154460353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLXNA3
Entrez Gene ID
55558
Gene Name
plexin A3
Gene Aliases
6.3, HSSEXGENE, PLXN3, PLXN4, XAP-6
UniGene
Hs.632839
Gene Chromosome Location
Chr.X: 154458281-154473646 on Build GRCh38
Transcripts
NM_017514.4 XM_005274705.4 XM_005274706.4 XM_006724829.3

SNP Detail

SNP ID Chromosome Location
rs781950701 Chr.X: 154460220-154460220 on Build GRCh38
rs782671802 Chr.X: 154460178-154460178 on Build GRCh38
rs144427846 Chr.X: 154460278-154460278 on Build GRCh38
rs782142719 Chr.X: 154460153-154460153 on Build GRCh38
rs149878050 Chr.X: 154460254-154460254 on Build GRCh38
rs782478833 Chr.X: 154460228-154460228 on Build GRCh38
rs782247826 Chr.X: 154460224-154460225 on Build GRCh38
rs782437535 Chr.X: 154460180-154460180 on Build GRCh38
rs782551566 Chr.X: 154460322-154460322 on Build GRCh38
rs149048914 Chr.X: 154460262-154460262 on Build GRCh38
rs375310385 Chr.X: 154460225-154460225 on Build GRCh38
rs782218651 Chr.X: 154460188-154460188 on Build GRCh38
rs782516007 Chr.X: 154460189-154460189 on Build GRCh38
rs782359318 Chr.X: 154460213-154460213 on Build GRCh38
rs782442410 Chr.X: 154460306-154460306 on Build GRCh38
rs782177013 Chr.X: 154460208-154460208 on Build GRCh38
rs782125563 Chr.X: 154460221-154460221 on Build GRCh38
rs782185732 Chr.X: 154460242-154460242 on Build GRCh38
rs781844352 Chr.X: 154460316-154460316 on Build GRCh38
rs781920321 Chr.X: 154460285-154460285 on Build GRCh38
rs782784290 Chr.X: 154460230-154460230 on Build GRCh38
rs1043167831 Chr.X: 154460226-154460226 on Build GRCh38
rs201924446 Chr.X: 154460222-154460222 on Build GRCh38
rs782264572 Chr.X: 154460179-154460179 on Build GRCh38
rs781882951 Chr.X: 154460303-154460303 on Build GRCh38
rs782698823 Chr.X: 154460307-154460307 on Build GRCh38
rs782215325 Chr.X: 154460253-154460253 on Build GRCh38
rs782472371 Chr.X: 154460311-154460311 on Build GRCh38
rs781929516 Chr.X: 154460204-154460204 on Build GRCh38
rs912596332 Chr.X: 154460187-154460187 on Build GRCh38
rs141311907 Chr.X: 154460327-154460327 on Build GRCh38
rs782808493 Chr.X: 154460302-154460302 on Build GRCh38
rs376190456 Chr.X: 154460170-154460170 on Build GRCh38
rs782122729 Chr.X: 154460301-154460301 on Build GRCh38
rs1054864178 Chr.X: 154460240-154460240 on Build GRCh38
rs995344595 Chr.X: 154460268-154460268 on Build GRCh38
rs782432583 Chr.X: 154460247-154460247 on Build GRCh38
rs144950198 Chr.X: 154460261-154460261 on Build GRCh38
rs1026852450 Chr.X: 154460273-154460273 on Build GRCh38
rs200397807 Chr.X: 154460182-154460182 on Build GRCh38
rs782737285 Chr.X: 154460295-154460295 on Build GRCh38
rs782709405 Chr.X: 154460159-154460159 on Build GRCh38
rs781859171 Chr.X: 154460231-154460231 on Build GRCh38
rs782614980 Chr.X: 154460249-154460249 on Build GRCh38
rs202070666 Chr.X: 154460239-154460239 on Build GRCh38
rs200617248 Chr.X: 154460238-154460238 on Build GRCh38
rs781830544 Chr.X: 154460309-154460309 on Build GRCh38
rs782361229 Chr.X: 154460225-154460225 on Build GRCh38
rs782095183 Chr.X: 154460171-154460171 on Build GRCh38
rs782009812 Chr.X: 154460297-154460297 on Build GRCh38
rs782271984 Chr.X: 154460223-154460223 on Build GRCh38
rs782733882 Chr.X: 154460269-154460269 on Build GRCh38
rs147816954 Chr.X: 154460321-154460321 on Build GRCh38
rs368407407 Chr.X: 154460172-154460172 on Build GRCh38
rs782031983 Chr.X: 154460147-154460147 on Build GRCh38
rs35880504 Chr.X: 154460195-154460195 on Build GRCh38
rs781867355 Chr.X: 154460173-154460173 on Build GRCh38

View Full Product Details