Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95823146-95823419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD1
Entrez Gene ID
953
Gene Name
ectonucleoside triphosphate diphosphohydrolase 1
Gene Aliases
ATPDase, CD39, NTPDase-1, SPG64
UniGene
Hs.576612
Gene Chromosome Location
Chr.10: 95710901-95877266 on Build GRCh38
Transcripts
NM_001098175.1 NM_001164178.1 NM_001164179.1 NM_001164181.1 NM_001164182.1 NM_001164183.1 NM_001312654.1 NM_001320916.1 NM_001776.5 XM_011540370.2 XM_011540371.2 XM_011540372.2 XM_011540373.2 XM_011540374.2 XM_011540376.2 XM_011540377.2 XM_017016958.1 XM_017016959.1 XM_017016960.1 XM_017016961.1 XM_017016962.1 XM_017016963.1 XM_017016964.1 NR_038444.1

Gene Symbol
ENTPD1-AS1
Entrez Gene ID
728558
Gene Name
ENTPD1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.538374
Gene Chromosome Location
Chr.10: 95753206-96090235 on Build GRCh38
Transcripts
NM_001098175.1 NM_001164178.1 NM_001164179.1 NM_001164181.1 NM_001164182.1 NM_001164183.1 NM_001312654.1 NM_001320916.1 NM_001776.5 XM_011540370.2 XM_011540371.2 XM_011540372.2 XM_011540373.2 XM_011540374.2 XM_011540376.2 XM_011540377.2 XM_017016958.1 XM_017016959.1 XM_017016960.1 XM_017016961.1 XM_017016962.1 XM_017016963.1 XM_017016964.1 NR_038444.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6272671 c.38-7G>T p.? Chr.10: 95823230-95823230 on Build GRCh38
6067143 c.164A>T p.K55M Chr.10: 95823363-95823363 on Build GRCh38
5050432 c.114C>T p.T38T Chr.10: 95823334-95823334 on Build GRCh38
4172061 c.113C>T p.T38I Chr.10: 95823333-95823333 on Build GRCh38
4172062 c.134C>T p.T45I Chr.10: 95823333-95823333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763581010 Chr.10: 95823258-95823258 on Build GRCh38
rs749013354 Chr.10: 95823218-95823218 on Build GRCh38
rs751980010 Chr.10: 95823260-95823260 on Build GRCh38
rs1033246962 Chr.10: 95823333-95823333 on Build GRCh38
rs370422237 Chr.10: 95823347-95823347 on Build GRCh38
rs778623881 Chr.10: 95823353-95823353 on Build GRCh38
rs746482729 Chr.10: 95823380-95823380 on Build GRCh38
rs768166538 Chr.10: 95823391-95823391 on Build GRCh38
rs777736628 Chr.10: 95823286-95823286 on Build GRCh38
rs755284270 Chr.10: 95823199-95823199 on Build GRCh38
rs558050218 Chr.10: 95823178-95823178 on Build GRCh38
rs777162789 Chr.10: 95823384-95823385 on Build GRCh38
rs757189483 Chr.10: 95823322-95823322 on Build GRCh38
rs920578320 Chr.10: 95823175-95823175 on Build GRCh38
rs745455500 Chr.10: 95823237-95823237 on Build GRCh38
rs1000841129 Chr.10: 95823331-95823331 on Build GRCh38
rs766814208 Chr.10: 95823259-95823259 on Build GRCh38
rs745722207 Chr.10: 95823356-95823356 on Build GRCh38
rs375705285 Chr.10: 95823225-95823225 on Build GRCh38
rs150772804 Chr.10: 95823245-95823245 on Build GRCh38
rs756263091 Chr.10: 95823284-95823284 on Build GRCh38
rs3176889 Chr.10: 95823211-95823211 on Build GRCh38
rs1055858489 Chr.10: 95823205-95823205 on Build GRCh38
rs776038920 Chr.10: 95823397-95823397 on Build GRCh38
rs752685478 Chr.10: 95823280-95823280 on Build GRCh38
rs773822293 Chr.10: 95823249-95823249 on Build GRCh38
rs748314965 Chr.10: 95823208-95823208 on Build GRCh38
rs937342069 Chr.10: 95823177-95823177 on Build GRCh38
rs779796722 Chr.10: 95823376-95823376 on Build GRCh38
rs576943385 Chr.10: 95823396-95823396 on Build GRCh38
rs755235175 Chr.10: 95823271-95823271 on Build GRCh38
rs770662343 Chr.10: 95823224-95823224 on Build GRCh38
rs755997413 Chr.10: 95823209-95823209 on Build GRCh38
rs753767695 Chr.10: 95823296-95823296 on Build GRCh38
rs201306382 Chr.10: 95823239-95823239 on Build GRCh38
rs771708732 Chr.10: 95823359-95823359 on Build GRCh38
rs758290973 Chr.10: 95823191-95823191 on Build GRCh38
rs781360486 Chr.10: 95823200-95823200 on Build GRCh38
rs767914674 Chr.10: 95823274-95823274 on Build GRCh38
rs866596463 Chr.10: 95823236-95823236 on Build GRCh38
rs560634896 Chr.10: 95823388-95823388 on Build GRCh38
rs368856010 Chr.10: 95823244-95823244 on Build GRCh38

View Full Product Details