Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5529026-5529293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTB
Entrez Gene ID
60
Gene Name
actin beta
Gene Aliases
BRWS1, PS1TP5BP1
UniGene
Hs.520640
Gene Chromosome Location
Chr.7: 5527148-5530601 on Build GRCh38
Transcripts
NM_001101.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6197157 c.358A>G p.T120A Chr.7: 5529166-5529166 on Build GRCh38
3639803 c.298G>A p.E100K Chr.7: 5529226-5529226 on Build GRCh38
6592849 c.252C>T p.D84D Chr.7: 5529281-5529281 on Build GRCh38
3639802 c.340G>A p.A114T Chr.7: 5529184-5529184 on Build GRCh38
5655559 c.286G>A p.E96K Chr.7: 5529247-5529247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs928322034 Chr.7: 5529052-5529052 on Build GRCh38
rs753268077 Chr.7: 5529148-5529148 on Build GRCh38
rs962058369 Chr.7: 5529114-5529114 on Build GRCh38
rs1041149457 Chr.7: 5529170-5529170 on Build GRCh38
rs771252379 Chr.7: 5529129-5529129 on Build GRCh38
rs373647923 Chr.7: 5529214-5529214 on Build GRCh38
rs752229167 Chr.7: 5529057-5529057 on Build GRCh38
rs751558283 Chr.7: 5529179-5529179 on Build GRCh38
rs762414570 Chr.7: 5529058-5529058 on Build GRCh38
rs754623772 Chr.7: 5529151-5529151 on Build GRCh38
rs372760503 Chr.7: 5529135-5529135 on Build GRCh38
rs766794678 Chr.7: 5529053-5529053 on Build GRCh38
rs753696029 Chr.7: 5529152-5529152 on Build GRCh38
rs765858205 Chr.7: 5529056-5529056 on Build GRCh38
rs770397951 Chr.7: 5529092-5529092 on Build GRCh38
rs551699029 Chr.7: 5529185-5529185 on Build GRCh38
rs145281194 Chr.7: 5529244-5529244 on Build GRCh38
rs397843863 Chr.7: 5529177-5529177 on Build GRCh38
rs951845713 Chr.7: 5529093-5529093 on Build GRCh38
rs746032511 Chr.7: 5529200-5529200 on Build GRCh38
rs17851374 Chr.7: 5529235-5529235 on Build GRCh38
rs771307382 Chr.7: 5529085-5529085 on Build GRCh38
rs574370199 Chr.7: 5529068-5529068 on Build GRCh38
rs749687095 Chr.7: 5529212-5529212 on Build GRCh38
rs765265404 Chr.7: 5529173-5529173 on Build GRCh38
rs760332854 Chr.7: 5529149-5529149 on Build GRCh38
rs778460789 Chr.7: 5529272-5529272 on Build GRCh38
rs747131945 Chr.7: 5529126-5529126 on Build GRCh38
rs139795454 Chr.7: 5529209-5529209 on Build GRCh38
rs761089427 Chr.7: 5529048-5529048 on Build GRCh38
rs758699008 Chr.7: 5529227-5529227 on Build GRCh38
rs991680762 Chr.7: 5529125-5529125 on Build GRCh38
rs763457988 Chr.7: 5529102-5529102 on Build GRCh38
rs747643184 Chr.7: 5529239-5529239 on Build GRCh38
rs587779773 Chr.7: 5529168-5529168 on Build GRCh38
rs574137728 Chr.7: 5529260-5529260 on Build GRCh38
rs11546933 Chr.7: 5529233-5529233 on Build GRCh38
rs537580307 Chr.7: 5529236-5529236 on Build GRCh38
rs541771250 Chr.7: 5529078-5529078 on Build GRCh38
rs760023216 Chr.7: 5529088-5529088 on Build GRCh38
rs371726724 Chr.7: 5529269-5529269 on Build GRCh38
rs748819558 Chr.7: 5529078-5529079 on Build GRCh38
rs11546910 Chr.7: 5529232-5529232 on Build GRCh38
rs750196776 Chr.7: 5529107-5529107 on Build GRCh38
rs772711453 Chr.7: 5529248-5529248 on Build GRCh38
rs201751687 Chr.7: 5529206-5529206 on Build GRCh38
rs13447399 Chr.7: 5529131-5529131 on Build GRCh38
rs587779774 Chr.7: 5529165-5529165 on Build GRCh38
rs780900079 Chr.7: 5529072-5529072 on Build GRCh38
rs763801135 Chr.7: 5529063-5529063 on Build GRCh38
rs767179841 Chr.7: 5529104-5529104 on Build GRCh38
rs760078095 Chr.7: 5529055-5529055 on Build GRCh38
rs781554752 Chr.7: 5529197-5529197 on Build GRCh38
rs761362868 Chr.7: 5529141-5529141 on Build GRCh38
rs756148153 Chr.7: 5529075-5529075 on Build GRCh38
rs981220862 Chr.7: 5529115-5529116 on Build GRCh38
rs760279394 Chr.7: 5529257-5529257 on Build GRCh38
rs772907450 Chr.7: 5529146-5529146 on Build GRCh38
rs184552929 Chr.7: 5529118-5529118 on Build GRCh38
rs769021550 Chr.7: 5529218-5529218 on Build GRCh38
rs779636227 Chr.7: 5529077-5529078 on Build GRCh38
rs200359258 Chr.7: 5529124-5529124 on Build GRCh38
rs531876233 Chr.7: 5529103-5529103 on Build GRCh38
rs143729583 Chr.7: 5529096-5529096 on Build GRCh38
rs374419443 Chr.7: 5529139-5529139 on Build GRCh38
rs397515470 Chr.7: 5529175-5529175 on Build GRCh38
rs780243010 Chr.7: 5529211-5529211 on Build GRCh38
rs564566763 Chr.7: 5529099-5529099 on Build GRCh38
rs754820594 Chr.7: 5529156-5529156 on Build GRCh38
rs550177210 Chr.7: 5529108-5529108 on Build GRCh38
rs531570981 Chr.7: 5529230-5529230 on Build GRCh38
rs189833026 Chr.7: 5529130-5529130 on Build GRCh38
rs560194132 Chr.7: 5529080-5529080 on Build GRCh38
rs779953111 Chr.7: 5529113-5529113 on Build GRCh38
rs11546931 Chr.7: 5529237-5529237 on Build GRCh38
rs587779772 Chr.7: 5529217-5529217 on Build GRCh38
rs73332405 Chr.7: 5529145-5529145 on Build GRCh38
rs769239344 Chr.7: 5529136-5529136 on Build GRCh38
rs527433029 Chr.7: 5529082-5529082 on Build GRCh38
rs750334287 Chr.7: 5529052-5529053 on Build GRCh38
rs755998236 Chr.7: 5529075-5529075 on Build GRCh38

View Full Product Details