Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 76154806-76155063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPATCH2L
Entrez Gene ID
55668
Gene Name
G-patch domain containing 2 like
Gene Aliases
C14orf118
UniGene
Hs.410231
Gene Chromosome Location
Chr.14: 76151885-76214473 on Build GRCh38
Transcripts
NM_001322026.1 NM_001322027.1 NM_001322028.1 NM_001322029.1 NM_001322030.1 NM_001322031.1 NM_001322032.1 NM_017926.3 NM_017972.3 NR_110314.1 XM_006720191.2 XM_017021427.1 XM_017021428.1 XM_017021429.1 XM_017021430.1 XM_017021431.1 XM_017021432.1 XM_017021433.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5890798 c.572G>A p.R191K Chr.14: 76154935-76154935 on Build GRCh38
4850159 c.449C>T p.S150L Chr.14: 76154812-76154812 on Build GRCh38
1748994 c.519G>C p.E173D Chr.14: 76154882-76154882 on Build GRCh38
1660835 c.513G>T p.W171C Chr.14: 76154876-76154876 on Build GRCh38
4850160 c.449C>T p.S150L Chr.14: 76154812-76154812 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113776212 Chr.14: 76154880-76154880 on Build GRCh38
rs778831501 Chr.14: 76154871-76154871 on Build GRCh38
rs747861571 Chr.14: 76154872-76154872 on Build GRCh38
rs1025455323 Chr.14: 76154955-76154955 on Build GRCh38
rs924419126 Chr.14: 76154957-76154957 on Build GRCh38
rs754855329 Chr.14: 76154870-76154870 on Build GRCh38
rs141077103 Chr.14: 76154862-76154862 on Build GRCh38
rs758226453 Chr.14: 76155015-76155015 on Build GRCh38
rs755584927 Chr.14: 76154861-76154861 on Build GRCh38
rs946279786 Chr.14: 76154849-76154849 on Build GRCh38
rs747114916 Chr.14: 76154903-76154903 on Build GRCh38
rs774855958 Chr.14: 76154949-76154949 on Build GRCh38
rs776838335 Chr.14: 76154909-76154909 on Build GRCh38
rs772385336 Chr.14: 76154879-76154879 on Build GRCh38
rs767138836 Chr.14: 76154847-76154847 on Build GRCh38
rs778689856 Chr.14: 76154993-76154993 on Build GRCh38
rs933138459 Chr.14: 76155009-76155009 on Build GRCh38
rs144915442 Chr.14: 76154912-76154912 on Build GRCh38
rs765632272 Chr.14: 76154913-76154913 on Build GRCh38
rs755638035 Chr.14: 76154958-76154958 on Build GRCh38
rs763183489 Chr.14: 76154926-76154926 on Build GRCh38
rs773518569 Chr.14: 76154884-76154884 on Build GRCh38
rs550282443 Chr.14: 76154852-76154852 on Build GRCh38
rs200600695 Chr.14: 76155032-76155032 on Build GRCh38
rs753280501 Chr.14: 76154982-76154982 on Build GRCh38
rs765929654 Chr.14: 76154960-76154960 on Build GRCh38
rs781462839 Chr.14: 76155033-76155033 on Build GRCh38
rs756975330 Chr.14: 76154840-76154840 on Build GRCh38
rs775972956 Chr.14: 76154915-76154915 on Build GRCh38
rs752559402 Chr.14: 76154995-76154995 on Build GRCh38
rs184669989 Chr.14: 76154943-76154943 on Build GRCh38
rs771066166 Chr.14: 76154904-76154904 on Build GRCh38
rs747168138 Chr.14: 76155030-76155030 on Build GRCh38
rs750014209 Chr.14: 76154952-76154952 on Build GRCh38
rs779973670 Chr.14: 76154869-76154869 on Build GRCh38

View Full Product Details