Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 55073735-55074000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP24
Entrez Gene ID
23358
Gene Name
ubiquitin specific peptidase 24
Gene Aliases
-
UniGene
Hs.477009
Gene Chromosome Location
Chr.1: 55066359-55215374 on Build GRCh38
Transcripts
NM_015306.2 XM_005270690.3 XM_017000831.1 XM_017000832.1 XM_017000833.1 XM_017000834.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6126861 c.7490A>G p.Y2497C Chr.1: 55073864-55073864 on Build GRCh38
4846421 c.7522C>T p.Q2508* Chr.1: 55073832-55073832 on Build GRCh38
3769891 c.7451T>A p.L2484* Chr.1: 55073903-55073903 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs866351596 Chr.1: 55073870-55073870 on Build GRCh38
rs748111001 Chr.1: 55073771-55073771 on Build GRCh38
rs746335287 Chr.1: 55073886-55073886 on Build GRCh38
rs761579082 Chr.1: 55073912-55073912 on Build GRCh38
rs775674913 Chr.1: 55073899-55073899 on Build GRCh38
rs770231986 Chr.1: 55073891-55073891 on Build GRCh38
rs575163919 Chr.1: 55073916-55073916 on Build GRCh38
rs371558492 Chr.1: 55073909-55073909 on Build GRCh38
rs777856263 Chr.1: 55073796-55073796 on Build GRCh38
rs867610842 Chr.1: 55073907-55073907 on Build GRCh38
rs550799268 Chr.1: 55073949-55073949 on Build GRCh38
rs983308738 Chr.1: 55073938-55073938 on Build GRCh38
rs545142111 Chr.1: 55073815-55073815 on Build GRCh38
rs747037617 Chr.1: 55073804-55073804 on Build GRCh38
rs956751323 Chr.1: 55073857-55073857 on Build GRCh38
rs970342367 Chr.1: 55073779-55073779 on Build GRCh38
rs1001007307 Chr.1: 55073816-55073816 on Build GRCh38
rs765795640 Chr.1: 55073928-55073928 on Build GRCh38
rs139906161 Chr.1: 55073972-55073972 on Build GRCh38
rs199933857 Chr.1: 55073884-55073884 on Build GRCh38
rs369426260 Chr.1: 55073847-55073847 on Build GRCh38
rs753444698 Chr.1: 55073936-55073936 on Build GRCh38
rs543828329 Chr.1: 55073921-55073921 on Build GRCh38
rs772853280 Chr.1: 55073918-55073918 on Build GRCh38
rs771436698 Chr.1: 55073860-55073860 on Build GRCh38
rs955298246 Chr.1: 55073959-55073959 on Build GRCh38

View Full Product Details