Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108830492-108830751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKNAD1
Entrez Gene ID
254268
Gene Name
AKNA domain containing 1
Gene Aliases
C1orf62
UniGene
Hs.729441
Gene Chromosome Location
Chr.1: 108815898-108858242 on Build GRCh38
Transcripts
NM_152763.4 NR_049760.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5040454 c.956A>T p.E319V Chr.1: 108830562-108830562 on Build GRCh38
5040452 c.1835A>T p.E612V Chr.1: 108830562-108830562 on Build GRCh38
7417967 c.1750G>T p.D584Y Chr.1: 108830647-108830647 on Build GRCh38
4020246 c.1822C>T p.R608C Chr.1: 108830575-108830575 on Build GRCh38
4868619 c.944G>A p.R315H Chr.1: 108830574-108830574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140157073 Chr.1: 108830586-108830586 on Build GRCh38
rs1034909465 Chr.1: 108830731-108830731 on Build GRCh38
rs778045627 Chr.1: 108830527-108830527 on Build GRCh38
rs779732862 Chr.1: 108830700-108830700 on Build GRCh38
rs765244096 Chr.1: 108830605-108830605 on Build GRCh38
rs543233868 Chr.1: 108830671-108830671 on Build GRCh38
rs375465135 Chr.1: 108830703-108830703 on Build GRCh38
rs199923486 Chr.1: 108830604-108830604 on Build GRCh38
rs753811728 Chr.1: 108830515-108830515 on Build GRCh38
rs376307747 Chr.1: 108830628-108830628 on Build GRCh38
rs540620131 Chr.1: 108830711-108830711 on Build GRCh38
rs771249763 Chr.1: 108830655-108830655 on Build GRCh38
rs200960053 Chr.1: 108830698-108830698 on Build GRCh38
rs980698475 Chr.1: 108830726-108830726 on Build GRCh38
rs776811283 Chr.1: 108830603-108830603 on Build GRCh38
rs755555676 Chr.1: 108830518-108830518 on Build GRCh38
rs143433407 Chr.1: 108830574-108830574 on Build GRCh38
rs755783040 Chr.1: 108830699-108830699 on Build GRCh38
rs745341212 Chr.1: 108830575-108830575 on Build GRCh38
rs202008360 Chr.1: 108830663-108830663 on Build GRCh38
rs370337411 Chr.1: 108830638-108830638 on Build GRCh38
rs969742182 Chr.1: 108830719-108830719 on Build GRCh38
rs769893580 Chr.1: 108830666-108830666 on Build GRCh38
rs368452927 Chr.1: 108830651-108830651 on Build GRCh38
rs982545504 Chr.1: 108830609-108830609 on Build GRCh38
rs752030271 Chr.1: 108830679-108830679 on Build GRCh38
rs192411720 Chr.1: 108830670-108830670 on Build GRCh38
rs892513954 Chr.1: 108830704-108830704 on Build GRCh38
rs769916637 Chr.1: 108830579-108830579 on Build GRCh38
rs768665987 Chr.1: 108830648-108830648 on Build GRCh38
rs200586359 Chr.1: 108830592-108830592 on Build GRCh38
rs1031021927 Chr.1: 108830532-108830532 on Build GRCh38
rs765731018 Chr.1: 108830523-108830523 on Build GRCh38
rs374736119 Chr.1: 108830589-108830589 on Build GRCh38
rs764480180 Chr.1: 108830672-108830672 on Build GRCh38
rs750005326 Chr.1: 108830634-108830634 on Build GRCh38
rs749175799 Chr.1: 108830588-108830588 on Build GRCh38
rs758777168 Chr.1: 108830525-108830525 on Build GRCh38
rs779071649 Chr.1: 108830682-108830683 on Build GRCh38
rs764468459 Chr.1: 108830620-108830620 on Build GRCh38
rs771034859 Chr.1: 108830602-108830602 on Build GRCh38
rs369842462 Chr.1: 108830597-108830597 on Build GRCh38
rs993735377 Chr.1: 108830599-108830599 on Build GRCh38
rs767861873 Chr.1: 108830685-108830685 on Build GRCh38
rs749265436 Chr.1: 108830644-108830644 on Build GRCh38
rs955435803 Chr.1: 108830715-108830716 on Build GRCh38
rs201555708 Chr.1: 108830654-108830654 on Build GRCh38
rs1022288751 Chr.1: 108830709-108830709 on Build GRCh38
rs753207825 Chr.1: 108830616-108830616 on Build GRCh38
rs773977078 Chr.1: 108830591-108830591 on Build GRCh38
rs544090925 Chr.1: 108830631-108830631 on Build GRCh38
rs139022413 Chr.1: 108830585-108830585 on Build GRCh38
rs762068972 Chr.1: 108830683-108830683 on Build GRCh38
rs1011365440 Chr.1: 108830705-108830705 on Build GRCh38
rs140857271 Chr.1: 108830639-108830639 on Build GRCh38

View Full Product Details