Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 22747250-22747511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBA3
Entrez Gene ID
57733
Gene Name
glucosylceramidase beta 3 (gene/pseudogene)
Gene Aliases
CBG, CBGL1, GLUC, KLRP
UniGene
Hs.653107
Gene Chromosome Location
Chr.4: 22692914-22819575 on Build GRCh38
Transcripts
NM_001128432.2 NM_001277225.1 NM_020973.4 NR_102355.1 NR_102356.1 NR_102357.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1485915 c.453T>C p.S151S Chr.4: 22747462-22747462 on Build GRCh38
4406205 c.327A>C p.K109N Chr.4: 22747336-22747336 on Build GRCh38
733694 c.287-2A>G p.? Chr.4: 22747294-22747294 on Build GRCh38
6100174 c.292G>T p.D98Y Chr.4: 22747301-22747301 on Build GRCh38
733692 c.341C>A p.P114H Chr.4: 22747350-22747350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754269435 Chr.4: 22747327-22747327 on Build GRCh38
rs761978614 Chr.4: 22747306-22747306 on Build GRCh38
rs372194282 Chr.4: 22747303-22747303 on Build GRCh38
rs868685564 Chr.4: 22747372-22747372 on Build GRCh38
rs755329225 Chr.4: 22747333-22747333 on Build GRCh38
rs751288202 Chr.4: 22747445-22747445 on Build GRCh38
rs750984356 Chr.4: 22747322-22747322 on Build GRCh38
rs760753418 Chr.4: 22747399-22747399 on Build GRCh38
rs778567555 Chr.4: 22747476-22747476 on Build GRCh38
rs749114036 Chr.4: 22747380-22747380 on Build GRCh38
rs781239183 Chr.4: 22747292-22747292 on Build GRCh38
rs755579073 Chr.4: 22747464-22747464 on Build GRCh38
rs765662114 Chr.4: 22747425-22747425 on Build GRCh38
rs887048047 Chr.4: 22747427-22747427 on Build GRCh38
rs189910504 Chr.4: 22747388-22747388 on Build GRCh38
rs575224803 Chr.4: 22747321-22747321 on Build GRCh38
rs773156605 Chr.4: 22747398-22747398 on Build GRCh38
rs777588471 Chr.4: 22747342-22747342 on Build GRCh38
rs111989254 Chr.4: 22747385-22747385 on Build GRCh38
rs202139282 Chr.4: 22747343-22747343 on Build GRCh38
rs760024988 Chr.4: 22747411-22747411 on Build GRCh38
rs746559077 Chr.4: 22747281-22747281 on Build GRCh38
rs201348337 Chr.4: 22747415-22747415 on Build GRCh38
rs767139688 Chr.4: 22747453-22747453 on Build GRCh38
rs749904393 Chr.4: 22747462-22747462 on Build GRCh38
rs779897804 Chr.4: 22747377-22747377 on Build GRCh38
rs769821749 Chr.4: 22747345-22747345 on Build GRCh38
rs573260891 Chr.4: 22747396-22747396 on Build GRCh38
rs745872404 Chr.4: 22747368-22747368 on Build GRCh38
rs370054152 Chr.4: 22747475-22747475 on Build GRCh38
rs769693291 Chr.4: 22747296-22747296 on Build GRCh38
rs779932053 Chr.4: 22747325-22747325 on Build GRCh38
rs756832208 Chr.4: 22747351-22747351 on Build GRCh38
rs756845869 Chr.4: 22747447-22747447 on Build GRCh38
rs749201905 Chr.4: 22747474-22747474 on Build GRCh38
rs780559452 Chr.4: 22747363-22747363 on Build GRCh38
rs376004241 Chr.4: 22747383-22747383 on Build GRCh38
rs770296771 Chr.4: 22747288-22747288 on Build GRCh38
rs775248473 Chr.4: 22747299-22747299 on Build GRCh38
rs751675029 Chr.4: 22747358-22747359 on Build GRCh38
rs745761646 Chr.4: 22747294-22747294 on Build GRCh38
rs754345059 Chr.4: 22747404-22747404 on Build GRCh38
rs187070546 Chr.4: 22747325-22747325 on Build GRCh38
rs755204237 Chr.4: 22747416-22747416 on Build GRCh38
rs769733975 Chr.4: 22747370-22747370 on Build GRCh38
rs1053845933 Chr.4: 22747473-22747473 on Build GRCh38
rs199890228 Chr.4: 22747305-22747305 on Build GRCh38
rs372609182 Chr.4: 22747402-22747402 on Build GRCh38

View Full Product Details