Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16537562-16537823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC72
Entrez Gene ID
100506049
Gene Name
leucine rich repeat containing 72
Gene Aliases
-
UniGene
Hs.278036
Gene Chromosome Location
Chr.7: 16526827-16581554 on Build GRCh38
Transcripts
NM_001195280.1 XM_011515057.1

SNP Detail

SNP ID Chromosome Location
rs796278752 Chr.7: 16537680-16537680 on Build GRCh38
rs983542075 Chr.7: 16537763-16537763 on Build GRCh38
rs948125312 Chr.7: 16537622-16537622 on Build GRCh38
rs917939078 Chr.7: 16537620-16537620 on Build GRCh38
rs764596576 Chr.7: 16537738-16537738 on Build GRCh38
rs557239496 Chr.7: 16537725-16537725 on Build GRCh38
rs1051417045 Chr.7: 16537739-16537739 on Build GRCh38
rs997459874 Chr.7: 16537703-16537707 on Build GRCh38
rs890613723 Chr.7: 16537607-16537607 on Build GRCh38
rs1001453483 Chr.7: 16537791-16537791 on Build GRCh38
rs6961390 Chr.7: 16537751-16537751 on Build GRCh38
rs377378983 Chr.7: 16537715-16537715 on Build GRCh38
rs1017843479 Chr.7: 16537690-16537690 on Build GRCh38
rs766756519 Chr.7: 16537587-16537587 on Build GRCh38
rs532780210 Chr.7: 16537608-16537609 on Build GRCh38
rs769819148 Chr.7: 16537688-16537688 on Build GRCh38
rs746079302 Chr.7: 16537761-16537761 on Build GRCh38
rs968661307 Chr.7: 16537591-16537591 on Build GRCh38
rs754666345 Chr.7: 16537701-16537701 on Build GRCh38
rs979668803 Chr.7: 16537601-16537601 on Build GRCh38
rs941869197 Chr.7: 16537671-16537671 on Build GRCh38
rs992137812 Chr.7: 16537611-16537611 on Build GRCh38
rs369407971 Chr.7: 16537679-16537679 on Build GRCh38
rs567048171 Chr.7: 16537765-16537765 on Build GRCh38
rs1045158194 Chr.7: 16537645-16537645 on Build GRCh38
rs909042119 Chr.7: 16537655-16537655 on Build GRCh38
rs557130317 Chr.7: 16537609-16537609 on Build GRCh38

View Full Product Details