Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 124038966-124039227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR10G7
Entrez Gene ID
390265
Gene Name
olfactory receptor family 10 subfamily G member 7
Gene Aliases
OR11-283
UniGene
Hs.553761
Gene Chromosome Location
Chr.11: 124038066-124039001 on Build GRCh38
Transcripts
NM_001004463.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
75709 c.35T>A p.L12H Chr.11: 124038967-124038967 on Build GRCh38
4709597 c.22A>T p.T8S Chr.11: 124038980-124038980 on Build GRCh38
4978734 c.9C>T p.N3N Chr.11: 124038993-124038993 on Build GRCh38
5907570 c.25G>A p.A9T Chr.11: 124038977-124038977 on Build GRCh38
6461606 c.36C>T p.L12L Chr.11: 124038966-124038966 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914742153 Chr.11: 124039115-124039115 on Build GRCh38
rs147004963 Chr.11: 124039179-124039179 on Build GRCh38
rs776429062 Chr.11: 124038987-124038987 on Build GRCh38
rs549859141 Chr.11: 124038995-124038995 on Build GRCh38
rs766385492 Chr.11: 124038999-124038999 on Build GRCh38
rs780737223 Chr.11: 124039045-124039045 on Build GRCh38
rs531350307 Chr.11: 124038993-124038993 on Build GRCh38
rs746456749 Chr.11: 124039047-124039047 on Build GRCh38
rs988964449 Chr.11: 124039073-124039073 on Build GRCh38
rs754656342 Chr.11: 124039055-124039055 on Build GRCh38
rs756217204 Chr.11: 124039011-124039011 on Build GRCh38
rs3894197 Chr.11: 124038988-124038988 on Build GRCh38
rs764233293 Chr.11: 124039015-124039015 on Build GRCh38
rs912273471 Chr.11: 124039060-124039060 on Build GRCh38
rs1004364588 Chr.11: 124039131-124039131 on Build GRCh38
rs561584169 Chr.11: 124039037-124039037 on Build GRCh38
rs906941151 Chr.11: 124039084-124039084 on Build GRCh38
rs780910674 Chr.11: 124039049-124039049 on Build GRCh38
rs764931043 Chr.11: 124038992-124038992 on Build GRCh38
rs996165459 Chr.11: 124039189-124039189 on Build GRCh38
rs779117370 Chr.11: 124039031-124039031 on Build GRCh38
rs757555096 Chr.11: 124039019-124039019 on Build GRCh38
rs1044657028 Chr.11: 124039191-124039191 on Build GRCh38
rs187123603 Chr.11: 124039064-124039064 on Build GRCh38
rs1016167568 Chr.11: 124039143-124039143 on Build GRCh38
rs754071524 Chr.11: 124039017-124039017 on Build GRCh38
rs758783003 Chr.11: 124039043-124039043 on Build GRCh38

View Full Product Details