Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2975407-2975650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKMYT1
Entrez Gene ID
9088
Gene Name
protein kinase, membrane associated tyrosine/threonine 1
Gene Aliases
MYT1, PPP1R126
UniGene
Hs.732385
Gene Chromosome Location
Chr.16: 2972791-2980539 on Build GRCh38
Transcripts
NM_001258450.1 NM_001258451.1 NM_004203.4 NM_182687.2 XM_011522734.2 XM_011522735.2 XM_011522736.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6016014 c.638A>G p.Y213C Chr.16: 2975553-2975553 on Build GRCh38
5504160 c.721C>T p.L241L Chr.16: 2975470-2975470 on Build GRCh38
1377242 c.724G>A p.G242R Chr.16: 2975467-2975467 on Build GRCh38
6747459 c.560C>T p.T187M Chr.16: 2975631-2975631 on Build GRCh38
1220808 c.736C>T p.R246C Chr.16: 2975455-2975455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762073643 Chr.16: 2975547-2975547 on Build GRCh38
rs373318831 Chr.16: 2975433-2975433 on Build GRCh38
rs760711402 Chr.16: 2975533-2975533 on Build GRCh38
rs113508225 Chr.16: 2975511-2975511 on Build GRCh38
rs777250172 Chr.16: 2975561-2975561 on Build GRCh38
rs775478177 Chr.16: 2975527-2975527 on Build GRCh38
rs35192104 Chr.16: 2975454-2975454 on Build GRCh38
rs768874579 Chr.16: 2975606-2975606 on Build GRCh38
rs773458421 Chr.16: 2975625-2975625 on Build GRCh38
rs147158922 Chr.16: 2975464-2975464 on Build GRCh38
rs377489890 Chr.16: 2975470-2975470 on Build GRCh38
rs371206404 Chr.16: 2975627-2975627 on Build GRCh38
rs771871236 Chr.16: 2975598-2975598 on Build GRCh38
rs780471941 Chr.16: 2975553-2975553 on Build GRCh38
rs372008941 Chr.16: 2975631-2975631 on Build GRCh38
rs752679639 Chr.16: 2975456-2975456 on Build GRCh38
rs762153541 Chr.16: 2975621-2975621 on Build GRCh38
rs886279902 Chr.16: 2975565-2975565 on Build GRCh38
rs141696813 Chr.16: 2975461-2975461 on Build GRCh38
rs564993872 Chr.16: 2975574-2975574 on Build GRCh38
rs140937042 Chr.16: 2975491-2975491 on Build GRCh38
rs778792546 Chr.16: 2975579-2975579 on Build GRCh38
rs765324345 Chr.16: 2975624-2975624 on Build GRCh38
rs748953207 Chr.16: 2975563-2975563 on Build GRCh38
rs752072941 Chr.16: 2975556-2975556 on Build GRCh38
rs775360025 Chr.16: 2975601-2975601 on Build GRCh38
rs760679296 Chr.16: 2975605-2975605 on Build GRCh38
rs143167027 Chr.16: 2975541-2975541 on Build GRCh38
rs764268414 Chr.16: 2975540-2975540 on Build GRCh38
rs777441360 Chr.16: 2975501-2975501 on Build GRCh38
rs56012022 Chr.16: 2975492-2975492 on Build GRCh38
rs773950604 Chr.16: 2975516-2975516 on Build GRCh38
rs763942626 Chr.16: 2975604-2975606 on Build GRCh38
rs562372351 Chr.16: 2975508-2975508 on Build GRCh38
rs531556877 Chr.16: 2975521-2975521 on Build GRCh38
rs762783250 Chr.16: 2975434-2975434 on Build GRCh38
rs771856495 Chr.16: 2975524-2975524 on Build GRCh38
rs1005835026 Chr.16: 2975447-2975447 on Build GRCh38
rs747559818 Chr.16: 2975497-2975497 on Build GRCh38
rs752062422 Chr.16: 2975630-2975630 on Build GRCh38
rs755687070 Chr.16: 2975559-2975559 on Build GRCh38
rs369663008 Chr.16: 2975548-2975548 on Build GRCh38
rs370544187 Chr.16: 2975626-2975626 on Build GRCh38
rs776790359 Chr.16: 2975613-2975613 on Build GRCh38
rs4149797 Chr.16: 2975551-2975551 on Build GRCh38
rs373163800 Chr.16: 2975590-2975590 on Build GRCh38
rs748933063 Chr.16: 2975507-2975507 on Build GRCh38
rs542397918 Chr.16: 2975498-2975498 on Build GRCh38
rs760981093 Chr.16: 2975460-2975460 on Build GRCh38
rs370010108 Chr.16: 2975485-2975485 on Build GRCh38
rs745700411 Chr.16: 2975586-2975586 on Build GRCh38
rs542714330 Chr.16: 2975435-2975435 on Build GRCh38
rs767640370 Chr.16: 2975455-2975455 on Build GRCh38

View Full Product Details