Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 23807203-23807474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2E1
Entrez Gene ID
7324
Gene Name
ubiquitin conjugating enzyme E2 E1
Gene Aliases
UBCH6
UniGene
Hs.164853
Gene Chromosome Location
Chr.3: 23805893-23891640 on Build GRCh38
Transcripts
NM_003341.4 NM_182666.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2948562 c.67G>T p.E23* Chr.3: 23807336-23807336 on Build GRCh38
1043317 c.124T>A p.S42T Chr.3: 23807393-23807393 on Build GRCh38
1043316 c.28A>G p.T10A Chr.3: 23807297-23807297 on Build GRCh38
7396487 c.103A>G p.K35E Chr.3: 23807372-23807372 on Build GRCh38
6443237 c.97G>A p.E33K Chr.3: 23807366-23807366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766193190 Chr.3: 23807368-23807368 on Build GRCh38
rs747890878 Chr.3: 23807362-23807365 on Build GRCh38
rs761780211 Chr.3: 23807443-23807443 on Build GRCh38
rs758156787 Chr.3: 23807258-23807258 on Build GRCh38
rs373825917 Chr.3: 23807399-23807399 on Build GRCh38
rs200438173 Chr.3: 23807401-23807401 on Build GRCh38
rs563516000 Chr.3: 23807433-23807433 on Build GRCh38
rs776563677 Chr.3: 23807355-23807355 on Build GRCh38
rs918391207 Chr.3: 23807390-23807390 on Build GRCh38
rs1009974721 Chr.3: 23807231-23807231 on Build GRCh38
rs918041203 Chr.3: 23807444-23807444 on Build GRCh38
rs771126027 Chr.3: 23807420-23807420 on Build GRCh38
rs955063000 Chr.3: 23807251-23807251 on Build GRCh38
rs780534779 Chr.3: 23807262-23807262 on Build GRCh38
rs554807476 Chr.3: 23807450-23807450 on Build GRCh38
rs758101921 Chr.3: 23807365-23807365 on Build GRCh38
rs36060625 Chr.3: 23807344-23807344 on Build GRCh38
rs771179273 Chr.3: 23807330-23807330 on Build GRCh38
rs749415679 Chr.3: 23807418-23807418 on Build GRCh38
rs368849761 Chr.3: 23807252-23807252 on Build GRCh38
rs918710649 Chr.3: 23807264-23807264 on Build GRCh38
rs777873920 Chr.3: 23807408-23807408 on Build GRCh38
rs140553923 Chr.3: 23807287-23807287 on Build GRCh38
rs781155886 Chr.3: 23807357-23807359 on Build GRCh38
rs749941203 Chr.3: 23807363-23807363 on Build GRCh38
rs3214837 Chr.3: 23807442-23807442 on Build GRCh38
rs764012134 Chr.3: 23807353-23807353 on Build GRCh38
rs774620737 Chr.3: 23807427-23807427 on Build GRCh38
rs760518877 Chr.3: 23807352-23807352 on Build GRCh38
rs756856856 Chr.3: 23807241-23807241 on Build GRCh38
rs755329307 Chr.3: 23807380-23807380 on Build GRCh38
rs951168153 Chr.3: 23807275-23807275 on Build GRCh38
rs745648610 Chr.3: 23807358-23807358 on Build GRCh38
rs748643952 Chr.3: 23807392-23807392 on Build GRCh38
rs768440125 Chr.3: 23807442-23807442 on Build GRCh38
rs746093181 Chr.3: 23807432-23807432 on Build GRCh38
rs781427154 Chr.3: 23807391-23807391 on Build GRCh38
rs200711549 Chr.3: 23807248-23807248 on Build GRCh38
rs769620688 Chr.3: 23807431-23807432 on Build GRCh38
rs755530456 Chr.3: 23807271-23807271 on Build GRCh38
rs976944058 Chr.3: 23807249-23807249 on Build GRCh38
rs879182045 Chr.3: 23807259-23807259 on Build GRCh38
rs759588393 Chr.3: 23807432-23807433 on Build GRCh38
rs772767761 Chr.3: 23807449-23807449 on Build GRCh38
rs764065201 Chr.3: 23807240-23807240 on Build GRCh38
rs749481934 Chr.3: 23807311-23807311 on Build GRCh38
rs973979390 Chr.3: 23807384-23807384 on Build GRCh38
rs373602200 Chr.3: 23807442-23807443 on Build GRCh38
rs751969832 Chr.3: 23807376-23807376 on Build GRCh38
rs536582921 Chr.3: 23807356-23807356 on Build GRCh38
rs201307853 Chr.3: 23807448-23807448 on Build GRCh38
rs773191666 Chr.3: 23807304-23807304 on Build GRCh38
rs747429325 Chr.3: 23807269-23807269 on Build GRCh38
rs985562945 Chr.3: 23807343-23807343 on Build GRCh38
rs748610486 Chr.3: 23807298-23807298 on Build GRCh38

View Full Product Details