Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46184665-46184896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN1
Entrez Gene ID
10103
Gene Name
tetraspanin 1
Gene Aliases
NET1, TM4C, TM4SF
UniGene
Hs.38972
Gene Chromosome Location
Chr.1: 46175077-46196489 on Build GRCh38
Transcripts
NM_005727.3 XM_011540460.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4008550 c.388T>C p.Y130H Chr.1: 46184833-46184833 on Build GRCh38
4631590 c.350T>C p.F117S Chr.1: 46184795-46184795 on Build GRCh38
7163102 c.355A>G p.T119A Chr.1: 46184800-46184800 on Build GRCh38
3689737 c.356C>T p.T119M Chr.1: 46184801-46184801 on Build GRCh38
6810581 c.360G>A p.L120L Chr.1: 46184805-46184805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776058739 Chr.1: 46184802-46184802 on Build GRCh38
rs765375799 Chr.1: 46184743-46184743 on Build GRCh38
rs200825766 Chr.1: 46184751-46184751 on Build GRCh38
rs747354292 Chr.1: 46184792-46184792 on Build GRCh38
rs1042794130 Chr.1: 46184812-46184812 on Build GRCh38
rs777916985 Chr.1: 46184782-46184782 on Build GRCh38
rs781635698 Chr.1: 46184805-46184805 on Build GRCh38
rs767148620 Chr.1: 46184692-46184692 on Build GRCh38
rs776656323 Chr.1: 46184848-46184848 on Build GRCh38
rs767827146 Chr.1: 46184745-46184747 on Build GRCh38
rs765158447 Chr.1: 46184867-46184867 on Build GRCh38
rs766393208 Chr.1: 46184701-46184701 on Build GRCh38
rs760601622 Chr.1: 46184700-46184700 on Build GRCh38
rs928027063 Chr.1: 46184823-46184823 on Build GRCh38
rs767993057 Chr.1: 46184817-46184817 on Build GRCh38
rs768126575 Chr.1: 46184835-46184835 on Build GRCh38
rs540727697 Chr.1: 46184843-46184843 on Build GRCh38
rs140948297 Chr.1: 46184802-46184802 on Build GRCh38
rs41286815 Chr.1: 46184870-46184870 on Build GRCh38
rs200522147 Chr.1: 46184754-46184754 on Build GRCh38
rs773045756 Chr.1: 46184839-46184839 on Build GRCh38
rs746396355 Chr.1: 46184815-46184815 on Build GRCh38
rs924254322 Chr.1: 46184801-46184801 on Build GRCh38
rs759526347 Chr.1: 46184862-46184862 on Build GRCh38
rs773916045 Chr.1: 46184832-46184832 on Build GRCh38
rs537019116 Chr.1: 46184726-46184726 on Build GRCh38
rs747677012 Chr.1: 46184834-46184834 on Build GRCh38
rs752798173 Chr.1: 46184752-46184752 on Build GRCh38
rs369161050 Chr.1: 46184702-46184702 on Build GRCh38
rs748867539 Chr.1: 46184762-46184762 on Build GRCh38
rs569552283 Chr.1: 46184711-46184711 on Build GRCh38
rs773134043 Chr.1: 46184699-46184699 on Build GRCh38

View Full Product Details