Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29561623-29561894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIPSNAP1
Entrez Gene ID
8508
Gene Name
nipsnap homolog 1 (C. elegans)
Gene Aliases
-
UniGene
Hs.173878
Gene Chromosome Location
Chr.22: 29554808-29581337 on Build GRCh38
Transcripts
NM_001202502.1 NM_003634.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
725984 c.458G>T p.R153M Chr.22: 29561627-29561627 on Build GRCh38
5799852 c.382T>A p.F128I Chr.22: 29561848-29561848 on Build GRCh38
1259134 c.432C>T p.N144N Chr.22: 29561798-29561798 on Build GRCh38
1714254 c.424C>T p.L142F Chr.22: 29561806-29561806 on Build GRCh38
5744087 c.406A>G p.M136V Chr.22: 29561824-29561824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760016355 Chr.22: 29561691-29561691 on Build GRCh38
rs951713221 Chr.22: 29561688-29561688 on Build GRCh38
rs756699489 Chr.22: 29561777-29561777 on Build GRCh38
rs772325535 Chr.22: 29561669-29561669 on Build GRCh38
rs763697334 Chr.22: 29561767-29561767 on Build GRCh38
rs755847660 Chr.22: 29561660-29561660 on Build GRCh38
rs779261784 Chr.22: 29561661-29561661 on Build GRCh38
rs138586681 Chr.22: 29561810-29561810 on Build GRCh38
rs1028733942 Chr.22: 29561670-29561670 on Build GRCh38
rs771133886 Chr.22: 29561872-29561872 on Build GRCh38
rs779689147 Chr.22: 29561808-29561808 on Build GRCh38
rs181718552 Chr.22: 29561870-29561870 on Build GRCh38
rs771565074 Chr.22: 29561686-29561686 on Build GRCh38
rs968752872 Chr.22: 29561720-29561720 on Build GRCh38
rs966615573 Chr.22: 29561649-29561649 on Build GRCh38
rs144660913 Chr.22: 29561846-29561846 on Build GRCh38
rs753617295 Chr.22: 29561817-29561817 on Build GRCh38
rs35475756 Chr.22: 29561645-29561646 on Build GRCh38
rs561154100 Chr.22: 29561730-29561730 on Build GRCh38
rs777970851 Chr.22: 29561668-29561669 on Build GRCh38
rs147998965 Chr.22: 29561837-29561837 on Build GRCh38
rs529824220 Chr.22: 29561759-29561759 on Build GRCh38
rs778071409 Chr.22: 29561850-29561850 on Build GRCh38
rs767502401 Chr.22: 29561779-29561779 on Build GRCh38
rs201733738 Chr.22: 29561687-29561687 on Build GRCh38
rs753863845 Chr.22: 29561726-29561726 on Build GRCh38
rs370460779 Chr.22: 29561678-29561678 on Build GRCh38
rs375850456 Chr.22: 29561683-29561683 on Build GRCh38
rs368283450 Chr.22: 29561744-29561744 on Build GRCh38
rs78944745 Chr.22: 29561715-29561715 on Build GRCh38
rs773445013 Chr.22: 29561681-29561681 on Build GRCh38
rs748285469 Chr.22: 29561666-29561666 on Build GRCh38
rs372172570 Chr.22: 29561752-29561752 on Build GRCh38
rs750322712 Chr.22: 29561795-29561795 on Build GRCh38
rs747096139 Chr.22: 29561869-29561869 on Build GRCh38
rs560791182 Chr.22: 29561713-29561713 on Build GRCh38
rs540137406 Chr.22: 29561844-29561844 on Build GRCh38
rs376765893 Chr.22: 29561760-29561760 on Build GRCh38
rs995906144 Chr.22: 29561665-29561665 on Build GRCh38
rs116836672 Chr.22: 29561807-29561807 on Build GRCh38

View Full Product Details