Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77266443-77266708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NGB
Entrez Gene ID
58157
Gene Name
neuroglobin
Gene Aliases
-
UniGene
Hs.274363
Gene Chromosome Location
Chr.14: 77265491-77271312 on Build GRCh38
Transcripts
NM_021257.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5342501 c.450C>T p.G150G Chr.14: 77266542-77266542 on Build GRCh38
3690213 c.417C>T p.A139A Chr.14: 77266575-77266575 on Build GRCh38
958119 c.335C>T p.S112F Chr.14: 77266657-77266657 on Build GRCh38
5090629 c.412G>A p.G138R Chr.14: 77266580-77266580 on Build GRCh38
699310 c.437G>C p.R146P Chr.14: 77266555-77266555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758209751 Chr.14: 77266499-77266500 on Build GRCh38
rs776271598 Chr.14: 77266573-77266574 on Build GRCh38
rs749793272 Chr.14: 77266608-77266608 on Build GRCh38
rs751777411 Chr.14: 77266581-77266581 on Build GRCh38
rs775347758 Chr.14: 77266648-77266648 on Build GRCh38
rs1008372237 Chr.14: 77266488-77266488 on Build GRCh38
rs892750677 Chr.14: 77266475-77266475 on Build GRCh38
rs758198892 Chr.14: 77266521-77266521 on Build GRCh38
rs760352166 Chr.14: 77266544-77266544 on Build GRCh38
rs915752077 Chr.14: 77266644-77266644 on Build GRCh38
rs749676611 Chr.14: 77266541-77266541 on Build GRCh38
rs149698049 Chr.14: 77266629-77266629 on Build GRCh38
rs765233408 Chr.14: 77266506-77266506 on Build GRCh38
rs956724350 Chr.14: 77266484-77266484 on Build GRCh38
rs372062527 Chr.14: 77266613-77266613 on Build GRCh38
rs576905261 Chr.14: 77266525-77266525 on Build GRCh38
rs368017108 Chr.14: 77266503-77266503 on Build GRCh38
rs143417735 Chr.14: 77266580-77266580 on Build GRCh38
rs968725170 Chr.14: 77266524-77266524 on Build GRCh38
rs756125208 Chr.14: 77266603-77266603 on Build GRCh38
rs563520691 Chr.14: 77266661-77266661 on Build GRCh38
rs542940662 Chr.14: 77266685-77266685 on Build GRCh38
rs770731188 Chr.14: 77266624-77266624 on Build GRCh38
rs780592741 Chr.14: 77266538-77266538 on Build GRCh38
rs192468767 Chr.14: 77266622-77266622 on Build GRCh38
rs150881015 Chr.14: 77266555-77266555 on Build GRCh38
rs762661328 Chr.14: 77266578-77266578 on Build GRCh38
rs555053943 Chr.14: 77266553-77266553 on Build GRCh38
rs767684404 Chr.14: 77266588-77266588 on Build GRCh38
rs750729239 Chr.14: 77266604-77266604 on Build GRCh38
rs978532025 Chr.14: 77266663-77266663 on Build GRCh38
rs529143000 Chr.14: 77266662-77266662 on Build GRCh38
rs34654851 Chr.14: 77266533-77266533 on Build GRCh38
rs745376735 Chr.14: 77266627-77266627 on Build GRCh38
rs200664951 Chr.14: 77266463-77266463 on Build GRCh38
rs746726662 Chr.14: 77266511-77266511 on Build GRCh38
rs768040023 Chr.14: 77266525-77266525 on Build GRCh38
rs753847011 Chr.14: 77266497-77266497 on Build GRCh38
rs779543496 Chr.14: 77266503-77266506 on Build GRCh38
rs555682650 Chr.14: 77266562-77266562 on Build GRCh38
rs781201201 Chr.14: 77266526-77266526 on Build GRCh38
rs770328948 Chr.14: 77266532-77266532 on Build GRCh38
rs564212509 Chr.14: 77266592-77266592 on Build GRCh38
rs866506585 Chr.14: 77266551-77266551 on Build GRCh38
rs201654532 Chr.14: 77266527-77266527 on Build GRCh38
rs772750347 Chr.14: 77266542-77266542 on Build GRCh38
rs756862816 Chr.14: 77266520-77266520 on Build GRCh38
rs925716038 Chr.14: 77266684-77266684 on Build GRCh38
rs998700540 Chr.14: 77266465-77266465 on Build GRCh38
rs775492814 Chr.14: 77266575-77266575 on Build GRCh38
rs368186692 Chr.14: 77266582-77266582 on Build GRCh38
rs543596986 Chr.14: 77266650-77266650 on Build GRCh38
rs959964289 Chr.14: 77266615-77266615 on Build GRCh38
rs751738344 Chr.14: 77266522-77266522 on Build GRCh38
rs779308596 Chr.14: 77266614-77266614 on Build GRCh38
rs772243940 Chr.14: 77266482-77266483 on Build GRCh38
rs776365950 Chr.14: 77266626-77266626 on Build GRCh38
rs746408394 Chr.14: 77266531-77266531 on Build GRCh38
rs765144506 Chr.14: 77266574-77266574 on Build GRCh38

View Full Product Details