Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45193946-45194203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2F2
Entrez Gene ID
2963
Gene Name
general transcription factor IIF subunit 2
Gene Aliases
BTF4, RAP30, TF2F2, TFIIF
UniGene
Hs.654582
Gene Chromosome Location
Chr.13: 45119830-45284105 on Build GRCh38
Transcripts
NM_004128.2 XM_011535052.2 XM_017020551.1 NM_198404.2

Gene Symbol
KCTD4
Entrez Gene ID
386618
Gene Name
potassium channel tetramerization domain containing 4
Gene Aliases
bA321C24.3
UniGene
Hs.23406
Gene Chromosome Location
Chr.13: 45192853-45201040 on Build GRCh38
Transcripts
NM_004128.2 XM_011535052.2 XM_017020551.1 NM_198404.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3384851 c.542A>C p.D181A Chr.13: 45194026-45194026 on Build GRCh38
3417618 c.451C>T p.R151C Chr.13: 45194117-45194117 on Build GRCh38
6895641 c.492C>T p.F164F Chr.13: 45194076-45194076 on Build GRCh38
4549226 c.467G>A p.R156K Chr.13: 45194101-45194101 on Build GRCh38
321149 c.556G>A p.E186K Chr.13: 45194012-45194012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201038607 Chr.13: 45194177-45194177 on Build GRCh38
rs780718179 Chr.13: 45194081-45194081 on Build GRCh38
rs770159016 Chr.13: 45194150-45194151 on Build GRCh38
rs748258708 Chr.13: 45194006-45194006 on Build GRCh38
rs747047179 Chr.13: 45194124-45194124 on Build GRCh38
rs763759913 Chr.13: 45194148-45194148 on Build GRCh38
rs1028298701 Chr.13: 45194077-45194077 on Build GRCh38
rs750333759 Chr.13: 45194086-45194086 on Build GRCh38
rs34555048 Chr.13: 45194121-45194121 on Build GRCh38
rs771518022 Chr.13: 45193999-45193999 on Build GRCh38
rs762666968 Chr.13: 45194048-45194048 on Build GRCh38
rs17851609 Chr.13: 45193984-45193984 on Build GRCh38
rs779893068 Chr.13: 45194092-45194092 on Build GRCh38
rs751179380 Chr.13: 45194165-45194165 on Build GRCh38
rs150359535 Chr.13: 45193987-45193987 on Build GRCh38
rs996773137 Chr.13: 45194055-45194055 on Build GRCh38
rs772248786 Chr.13: 45193988-45193988 on Build GRCh38
rs765613388 Chr.13: 45194038-45194038 on Build GRCh38
rs753064064 Chr.13: 45194046-45194046 on Build GRCh38
rs1046431135 Chr.13: 45193973-45193973 on Build GRCh38
rs760161994 Chr.13: 45194017-45194017 on Build GRCh38
rs766997687 Chr.13: 45194173-45194173 on Build GRCh38
rs776899557 Chr.13: 45194139-45194139 on Build GRCh38
rs750001763 Chr.13: 45194175-45194175 on Build GRCh38
rs779758432 Chr.13: 45193983-45193983 on Build GRCh38
rs756788877 Chr.13: 45194072-45194072 on Build GRCh38
rs143597700 Chr.13: 45194116-45194116 on Build GRCh38
rs185309053 Chr.13: 45194147-45194147 on Build GRCh38
rs761498026 Chr.13: 45194168-45194168 on Build GRCh38
rs903340778 Chr.13: 45193993-45193993 on Build GRCh38
rs777310266 Chr.13: 45194003-45194003 on Build GRCh38
rs138094135 Chr.13: 45194049-45194049 on Build GRCh38
rs376571272 Chr.13: 45193998-45193998 on Build GRCh38
rs772277733 Chr.13: 45194119-45194119 on Build GRCh38
rs756060711 Chr.13: 45194088-45194088 on Build GRCh38
rs368124187 Chr.13: 45194057-45194057 on Build GRCh38
rs551823576 Chr.13: 45193990-45193990 on Build GRCh38
rs760107154 Chr.13: 45194146-45194146 on Build GRCh38
rs957743678 Chr.13: 45194053-45194053 on Build GRCh38
rs534142034 Chr.13: 45194127-45194127 on Build GRCh38

View Full Product Details