Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 91630091-91630316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R3C
Entrez Gene ID
5507
Gene Name
protein phosphatase 1 regulatory subunit 3C
Gene Aliases
PPP1R5
UniGene
Hs.303090
Gene Chromosome Location
Chr.10: 91628440-91633101 on Build GRCh38
Transcripts
NM_005398.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
125539 c.688C>A p.Q230K Chr.10: 91630193-91630193 on Build GRCh38
6518098 c.625G>T p.G209C Chr.10: 91630256-91630256 on Build GRCh38
4572479 c.760T>C p.Y254H Chr.10: 91630121-91630121 on Build GRCh38
6754105 c.667C>T p.P223S Chr.10: 91630214-91630214 on Build GRCh38
686099 c.690G>A p.Q230Q Chr.10: 91630191-91630191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374238734 Chr.10: 91630166-91630166 on Build GRCh38
rs775691105 Chr.10: 91630237-91630237 on Build GRCh38
rs553748981 Chr.10: 91630165-91630165 on Build GRCh38
rs760157762 Chr.10: 91630219-91630219 on Build GRCh38
rs746486788 Chr.10: 91630151-91630151 on Build GRCh38
rs937535855 Chr.10: 91630245-91630245 on Build GRCh38
rs142799521 Chr.10: 91630252-91630252 on Build GRCh38
rs760810195 Chr.10: 91630248-91630248 on Build GRCh38
rs929470089 Chr.10: 91630221-91630221 on Build GRCh38
rs774750791 Chr.10: 91630215-91630216 on Build GRCh38
rs779017862 Chr.10: 91630129-91630129 on Build GRCh38
rs144290379 Chr.10: 91630287-91630287 on Build GRCh38
rs983863368 Chr.10: 91630131-91630131 on Build GRCh38
rs138872156 Chr.10: 91630202-91630202 on Build GRCh38
rs373998228 Chr.10: 91630135-91630135 on Build GRCh38
rs368346528 Chr.10: 91630132-91630132 on Build GRCh38
rs949125386 Chr.10: 91630278-91630278 on Build GRCh38
rs749681328 Chr.10: 91630196-91630196 on Build GRCh38
rs774730445 Chr.10: 91630209-91630209 on Build GRCh38
rs199996471 Chr.10: 91630225-91630225 on Build GRCh38
rs752149540 Chr.10: 91630286-91630286 on Build GRCh38
rs778620061 Chr.10: 91630209-91630210 on Build GRCh38
rs372071073 Chr.10: 91630134-91630134 on Build GRCh38
rs898007088 Chr.10: 91630175-91630175 on Build GRCh38
rs754068775 Chr.10: 91630271-91630271 on Build GRCh38
rs41286878 Chr.10: 91630285-91630285 on Build GRCh38
rs771234227 Chr.10: 91630199-91630199 on Build GRCh38
rs764482329 Chr.10: 91630276-91630276 on Build GRCh38
rs954817068 Chr.10: 91630118-91630118 on Build GRCh38
rs761717915 Chr.10: 91630277-91630277 on Build GRCh38
rs942304191 Chr.10: 91630136-91630136 on Build GRCh38

View Full Product Details