Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41489649-41489892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT36
Entrez Gene ID
8689
Gene Name
keratin 36
Gene Aliases
HA6, KRTHA6, hHa6
UniGene
Hs.248189
Gene Chromosome Location
Chr.17: 41486136-41489864 on Build GRCh38
Transcripts
NM_003771.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2840846 c.175C>A p.L59I Chr.17: 41489690-41489690 on Build GRCh38
4883584 c.30C>A p.F10L Chr.17: 41489835-41489835 on Build GRCh38
5816695 c.95G>T p.R32L Chr.17: 41489770-41489770 on Build GRCh38
4914141 c.22C>A p.P8T Chr.17: 41489843-41489843 on Build GRCh38
979202 c.12G>T p.Q4H Chr.17: 41489853-41489853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769219679 Chr.17: 41489825-41489825 on Build GRCh38
rs954919645 Chr.17: 41489814-41489814 on Build GRCh38
rs578258715 Chr.17: 41489700-41489700 on Build GRCh38
rs751470845 Chr.17: 41489857-41489857 on Build GRCh38
rs868033616 Chr.17: 41489797-41489797 on Build GRCh38
rs202223747 Chr.17: 41489842-41489842 on Build GRCh38
rs780330505 Chr.17: 41489731-41489731 on Build GRCh38
rs776271282 Chr.17: 41489747-41489747 on Build GRCh38
rs1007651555 Chr.17: 41489815-41489815 on Build GRCh38
rs866013543 Chr.17: 41489791-41489791 on Build GRCh38
rs529507843 Chr.17: 41489771-41489771 on Build GRCh38
rs775017982 Chr.17: 41489827-41489827 on Build GRCh38
rs369080313 Chr.17: 41489703-41489703 on Build GRCh38
rs781275139 Chr.17: 41489788-41489788 on Build GRCh38
rs767573909 Chr.17: 41489771-41489771 on Build GRCh38
rs771485812 Chr.17: 41489818-41489818 on Build GRCh38
rs760020435 Chr.17: 41489824-41489824 on Build GRCh38
rs143124390 Chr.17: 41489795-41489795 on Build GRCh38
rs761215695 Chr.17: 41489745-41489745 on Build GRCh38
rs972255485 Chr.17: 41489723-41489748 on Build GRCh38
rs748338517 Chr.17: 41489801-41489801 on Build GRCh38
rs954968449 Chr.17: 41489828-41489828 on Build GRCh38
rs146627816 Chr.17: 41489765-41489765 on Build GRCh38
rs1017669475 Chr.17: 41489849-41489849 on Build GRCh38
rs753320791 Chr.17: 41489682-41489682 on Build GRCh38
rs762743688 Chr.17: 41489840-41489840 on Build GRCh38
rs371182923 Chr.17: 41489796-41489796 on Build GRCh38
rs200810410 Chr.17: 41489803-41489803 on Build GRCh38
rs746629464 Chr.17: 41489724-41489724 on Build GRCh38
rs772454130 Chr.17: 41489740-41489740 on Build GRCh38
rs376708541 Chr.17: 41489691-41489691 on Build GRCh38
rs777645641 Chr.17: 41489721-41489721 on Build GRCh38
rs751377257 Chr.17: 41489785-41489785 on Build GRCh38
rs917100172 Chr.17: 41489735-41489735 on Build GRCh38
rs748347652 Chr.17: 41489739-41489739 on Build GRCh38
rs764004912 Chr.17: 41489773-41489773 on Build GRCh38
rs778992405 Chr.17: 41489738-41489738 on Build GRCh38
rs746975955 Chr.17: 41489723-41489723 on Build GRCh38
rs763808931 Chr.17: 41489845-41489845 on Build GRCh38
rs772256330 Chr.17: 41489802-41489802 on Build GRCh38
rs529324683 Chr.17: 41489838-41489838 on Build GRCh38
rs868829636 Chr.17: 41489672-41489672 on Build GRCh38
rs759806098 Chr.17: 41489699-41489699 on Build GRCh38
rs200188804 Chr.17: 41489725-41489725 on Build GRCh38
rs760344754 Chr.17: 41489865-41489865 on Build GRCh38
rs777015511 Chr.17: 41489823-41489823 on Build GRCh38
rs758140884 Chr.17: 41489706-41489706 on Build GRCh38
rs752557571 Chr.17: 41489861-41489861 on Build GRCh38
rs1003842 Chr.17: 41489769-41489769 on Build GRCh38
rs765892890 Chr.17: 41489676-41489676 on Build GRCh38
rs747368762 Chr.17: 41489813-41489813 on Build GRCh38
rs551238900 Chr.17: 41489786-41489786 on Build GRCh38
rs142937964 Chr.17: 41489704-41489704 on Build GRCh38
rs749301115 Chr.17: 41489736-41489736 on Build GRCh38
rs150707929 Chr.17: 41489811-41489811 on Build GRCh38
rs758955999 Chr.17: 41489751-41489751 on Build GRCh38
rs375069940 Chr.17: 41489675-41489675 on Build GRCh38
rs199500732 Chr.17: 41489770-41489770 on Build GRCh38
rs773514074 Chr.17: 41489743-41489743 on Build GRCh38

View Full Product Details