Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 42566087-42566358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERHL2
Entrez Gene ID
253190
Gene Name
serine hydrolase-like 2
Gene Aliases
dJ222E13.1
UniGene
Hs.728878
Gene Chromosome Location
Chr.22: 42553857-42574382 on Build GRCh38
Transcripts
NM_001284334.1 NM_014509.4 NR_104300.1 NR_104301.1 XM_017028738.1 XM_017028739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6269137 c.614-1G>T p.? Chr.22: 42566303-42566303 on Build GRCh38
3964375 c.628A>G p.R210G Chr.22: 42566318-42566318 on Build GRCh38
1484277 c.634C>A p.Q212K Chr.22: 42566324-42566324 on Build GRCh38
7223583 c.614-4C>G p.? Chr.22: 42566300-42566300 on Build GRCh38
7106508 c.636G>C p.Q212H Chr.22: 42566326-42566326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs578027158 Chr.22: 42566269-42566269 on Build GRCh38
rs779038651 Chr.22: 42566326-42566326 on Build GRCh38
rs774695008 Chr.22: 42566247-42566247 on Build GRCh38
rs769217450 Chr.22: 42566258-42566258 on Build GRCh38
rs758711341 Chr.22: 42566313-42566313 on Build GRCh38
rs777269777 Chr.22: 42566262-42566262 on Build GRCh38
rs749248978 Chr.22: 42566264-42566264 on Build GRCh38
rs749914514 Chr.22: 42566180-42566180 on Build GRCh38
rs977119099 Chr.22: 42566325-42566325 on Build GRCh38
rs559702726 Chr.22: 42566242-42566242 on Build GRCh38
rs763405033 Chr.22: 42566304-42566304 on Build GRCh38
rs960012084 Chr.22: 42566241-42566241 on Build GRCh38
rs376999609 Chr.22: 42566232-42566232 on Build GRCh38
rs113712697 Chr.22: 42566119-42566119 on Build GRCh38
rs886094696 Chr.22: 42566177-42566177 on Build GRCh38
rs192958278 Chr.22: 42566202-42566202 on Build GRCh38
rs752295102 Chr.22: 42566321-42566321 on Build GRCh38
rs765998025 Chr.22: 42566322-42566323 on Build GRCh38
rs976883000 Chr.22: 42566222-42566222 on Build GRCh38
rs530514017 Chr.22: 42566332-42566332 on Build GRCh38
rs139690969 Chr.22: 42566153-42566154 on Build GRCh38
rs200238208 Chr.22: 42566324-42566324 on Build GRCh38
rs764097513 Chr.22: 42566293-42566293 on Build GRCh38
rs760405357 Chr.22: 42566303-42566303 on Build GRCh38
rs760886455 Chr.22: 42566287-42566287 on Build GRCh38
rs13056731 Chr.22: 42566223-42566223 on Build GRCh38
rs189019711 Chr.22: 42566116-42566116 on Build GRCh38
rs755732745 Chr.22: 42566322-42566322 on Build GRCh38
rs765773914 Chr.22: 42566296-42566296 on Build GRCh38
rs537596162 Chr.22: 42566123-42566123 on Build GRCh38
rs766725093 Chr.22: 42566316-42566316 on Build GRCh38
rs776750874 Chr.22: 42566294-42566294 on Build GRCh38
rs746770694 Chr.22: 42566339-42566339 on Build GRCh38
rs771443894 Chr.22: 42566252-42566253 on Build GRCh38
rs768990341 Chr.22: 42566340-42566340 on Build GRCh38
rs199947066 Chr.22: 42566333-42566333 on Build GRCh38
rs1027359358 Chr.22: 42566174-42566174 on Build GRCh38
rs1050308094 Chr.22: 42566137-42566137 on Build GRCh38
rs1003303537 Chr.22: 42566114-42566114 on Build GRCh38
rs9623626 Chr.22: 42566212-42566212 on Build GRCh38
rs370022127 Chr.22: 42566203-42566203 on Build GRCh38
rs771854904 Chr.22: 42566276-42566276 on Build GRCh38
rs113416470 Chr.22: 42566323-42566323 on Build GRCh38
rs776805648 Chr.22: 42566341-42566341 on Build GRCh38
rs759440293 Chr.22: 42566275-42566275 on Build GRCh38
rs142103668 Chr.22: 42566170-42566170 on Build GRCh38
rs762647416 Chr.22: 42566200-42566200 on Build GRCh38
rs888608128 Chr.22: 42566140-42566140 on Build GRCh38
rs993543029 Chr.22: 42566201-42566201 on Build GRCh38
rs539659116 Chr.22: 42566110-42566110 on Build GRCh38
rs897433396 Chr.22: 42566191-42566191 on Build GRCh38
rs761879869 Chr.22: 42566295-42566295 on Build GRCh38
rs1018684718 Chr.22: 42566211-42566211 on Build GRCh38
rs764736290 Chr.22: 42566131-42566131 on Build GRCh38
rs375194492 Chr.22: 42566271-42566271 on Build GRCh38
rs13057222 Chr.22: 42566224-42566224 on Build GRCh38
rs559229417 Chr.22: 42566178-42566178 on Build GRCh38
rs1041064485 Chr.22: 42566117-42566117 on Build GRCh38
rs775609800 Chr.22: 42566284-42566284 on Build GRCh38
rs906399161 Chr.22: 42566113-42566113 on Build GRCh38
rs745764539 Chr.22: 42566328-42566328 on Build GRCh38
rs932755426 Chr.22: 42566121-42566121 on Build GRCh38
rs750908786 Chr.22: 42566303-42566303 on Build GRCh38

View Full Product Details