Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 169424586-169424841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEK1
Entrez Gene ID
4750
Gene Name
NIMA related kinase 1
Gene Aliases
NY-REN-55, SRPS2, SRPS2A, SRTD6
UniGene
Hs.481181
Gene Chromosome Location
Chr.4: 169393270-169612629 on Build GRCh38
Transcripts
NM_001199397.1 NM_001199398.1 NM_001199399.1 NM_001199400.1 NM_012224.2 XM_006714228.1 XM_011532003.1 XM_011532004.1 XM_017008249.1 XM_017008250.1 XM_017008251.1 XM_017008252.1 XM_017008253.1 XM_017008254.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1180597 c.3091C>G p.Q1031E Chr.4: 169424684-169424684 on Build GRCh38
5535324 c.3058C>T p.H1020Y Chr.4: 169424717-169424717 on Build GRCh38
1053157 c.3125C>A p.A1042E Chr.4: 169424650-169424650 on Build GRCh38
6099924 c.3110C>T p.P1037L Chr.4: 169424665-169424665 on Build GRCh38
1053159 c.3049A>G p.K1017E Chr.4: 169424726-169424726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370713361 Chr.4: 169424640-169424640 on Build GRCh38
rs749326267 Chr.4: 169424627-169424627 on Build GRCh38
rs752555915 Chr.4: 169424784-169424784 on Build GRCh38
rs772227033 Chr.4: 169424801-169424801 on Build GRCh38
rs772130033 Chr.4: 169424706-169424706 on Build GRCh38
rs762454081 Chr.4: 169424645-169424645 on Build GRCh38
rs377607698 Chr.4: 169424635-169424635 on Build GRCh38
rs960473869 Chr.4: 169424781-169424781 on Build GRCh38
rs745888585 Chr.4: 169424798-169424798 on Build GRCh38
rs368669247 Chr.4: 169424692-169424692 on Build GRCh38
rs779767983 Chr.4: 169424660-169424660 on Build GRCh38
rs371727818 Chr.4: 169424664-169424664 on Build GRCh38
rs190807750 Chr.4: 169424634-169424634 on Build GRCh38
rs796302803 Chr.4: 169424687-169424687 on Build GRCh38
rs755980153 Chr.4: 169424786-169424786 on Build GRCh38
rs768418367 Chr.4: 169424612-169424612 on Build GRCh38
rs765841630 Chr.4: 169424739-169424739 on Build GRCh38
rs780040740 Chr.4: 169424802-169424802 on Build GRCh38
rs769037266 Chr.4: 169424725-169424725 on Build GRCh38
rs375212680 Chr.4: 169424740-169424740 on Build GRCh38
rs954917764 Chr.4: 169424658-169424658 on Build GRCh38
rs375686118 Chr.4: 169424632-169424632 on Build GRCh38
rs765906291 Chr.4: 169424743-169424743 on Build GRCh38
rs7655924 Chr.4: 169424742-169424742 on Build GRCh38
rs747236398 Chr.4: 169424804-169424804 on Build GRCh38
rs142337223 Chr.4: 169424721-169424721 on Build GRCh38
rs748055814 Chr.4: 169424683-169424683 on Build GRCh38
rs769781253 Chr.4: 169424690-169424690 on Build GRCh38
rs367836027 Chr.4: 169424650-169424650 on Build GRCh38
rs775805551 Chr.4: 169424707-169424707 on Build GRCh38
rs760811823 Chr.4: 169424682-169424682 on Build GRCh38
rs776651638 Chr.4: 169424616-169424616 on Build GRCh38
rs745952330 Chr.4: 169424698-169424698 on Build GRCh38
rs865859911 Chr.4: 169424797-169424797 on Build GRCh38
rs886059228 Chr.4: 169424776-169424776 on Build GRCh38
rs200619983 Chr.4: 169424728-169424728 on Build GRCh38
rs777703171 Chr.4: 169424790-169424790 on Build GRCh38
rs763310804 Chr.4: 169424783-169424784 on Build GRCh38
rs199947197 Chr.4: 169424668-169424668 on Build GRCh38
rs780952942 Chr.4: 169424768-169424768 on Build GRCh38
rs751192051 Chr.4: 169424758-169424758 on Build GRCh38
rs1013407857 Chr.4: 169424630-169424630 on Build GRCh38
rs764463497 Chr.4: 169424644-169424644 on Build GRCh38
rs886059227 Chr.4: 169424700-169424700 on Build GRCh38
rs374598155 Chr.4: 169424649-169424649 on Build GRCh38
rs552200309 Chr.4: 169424730-169424730 on Build GRCh38
rs754634844 Chr.4: 169424759-169424759 on Build GRCh38
rs764822486 Chr.4: 169424646-169424646 on Build GRCh38

View Full Product Details