Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 124319596-124319823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR8D2
Entrez Gene ID
283160
Gene Name
olfactory receptor family 8 subfamily D member 2 (gene/pseudogene)
Gene Aliases
JCG2
UniGene
Hs.504212
Gene Chromosome Location
Chr.11: 124319262-124320197 on Build GRCh38
Transcripts
NM_001002918.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
297828 c.382C>T p.R128C Chr.11: 124319816-124319816 on Build GRCh38
5948831 c.595C>T p.L199L Chr.11: 124319603-124319603 on Build GRCh38
6868220 c.482C>T p.T161I Chr.11: 124319716-124319716 on Build GRCh38
3375707 c.507T>C p.C169C Chr.11: 124319691-124319691 on Build GRCh38
3375706 c.540T>G p.D180E Chr.11: 124319658-124319658 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772231561 Chr.11: 124319689-124319689 on Build GRCh38
rs927794468 Chr.11: 124319675-124319675 on Build GRCh38
rs560961386 Chr.11: 124319727-124319727 on Build GRCh38
rs200914193 Chr.11: 124319767-124319767 on Build GRCh38
rs749271846 Chr.11: 124319688-124319688 on Build GRCh38
rs776771740 Chr.11: 124319713-124319713 on Build GRCh38
rs759027625 Chr.11: 124319721-124319721 on Build GRCh38
rs111696603 Chr.11: 124319800-124319800 on Build GRCh38
rs753738230 Chr.11: 124319763-124319763 on Build GRCh38
rs371657294 Chr.11: 124319631-124319631 on Build GRCh38
rs762379495 Chr.11: 124319651-124319651 on Build GRCh38
rs575308677 Chr.11: 124319678-124319678 on Build GRCh38
rs201214294 Chr.11: 124319731-124319731 on Build GRCh38
rs143397967 Chr.11: 124319643-124319643 on Build GRCh38
rs750939397 Chr.11: 124319649-124319649 on Build GRCh38
rs150419269 Chr.11: 124319690-124319690 on Build GRCh38
rs760981484 Chr.11: 124319628-124319628 on Build GRCh38
rs142664111 Chr.11: 124319641-124319641 on Build GRCh38
rs754635766 Chr.11: 124319663-124319663 on Build GRCh38
rs374212663 Chr.11: 124319719-124319719 on Build GRCh38
rs918081937 Chr.11: 124319743-124319743 on Build GRCh38
rs777735632 Chr.11: 124319753-124319753 on Build GRCh38
rs768914648 Chr.11: 124319704-124319704 on Build GRCh38
rs560131977 Chr.11: 124319780-124319780 on Build GRCh38
rs556507903 Chr.11: 124319739-124319739 on Build GRCh38
rs778786235 Chr.11: 124319774-124319774 on Build GRCh38
rs751121226 Chr.11: 124319658-124319658 on Build GRCh38
rs151287996 Chr.11: 124319680-124319680 on Build GRCh38
rs765903401 Chr.11: 124319657-124319657 on Build GRCh38
rs780776143 Chr.11: 124319674-124319674 on Build GRCh38
rs542225246 Chr.11: 124319715-124319715 on Build GRCh38
rs368365573 Chr.11: 124319645-124319645 on Build GRCh38
rs145025229 Chr.11: 124319789-124319789 on Build GRCh38
rs781405818 Chr.11: 124319799-124319799 on Build GRCh38
rs766843978 Chr.11: 124319718-124319718 on Build GRCh38
rs780163805 Chr.11: 124319691-124319691 on Build GRCh38
rs748397485 Chr.11: 124319801-124319801 on Build GRCh38
rs140562028 Chr.11: 124319683-124319683 on Build GRCh38
rs939116181 Chr.11: 124319679-124319679 on Build GRCh38
rs759214639 Chr.11: 124319671-124319674 on Build GRCh38
rs763470996 Chr.11: 124319716-124319716 on Build GRCh38
rs142738979 Chr.11: 124319751-124319751 on Build GRCh38
rs368108920 Chr.11: 124319726-124319726 on Build GRCh38
rs747102054 Chr.11: 124319699-124319699 on Build GRCh38

View Full Product Details