Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 71854178-71854419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO9A
Entrez Gene ID
4649
Gene Name
myosin IXA
Gene Aliases
-
UniGene
Hs.513000
Gene Chromosome Location
Chr.15: 71822290-72118577 on Build GRCh38
Transcripts
NM_006901.3 XM_006720539.2 XM_011521613.2 XM_011521614.2 XM_011521615.2 XM_011521616.2 XM_011521617.2 XM_011521618.2 XM_011521619.2 XM_011521620.2 XM_011521621.2 XM_011521622.2 XM_011521623.2 XM_017022228.1 XM_017022229.1 XM_017022230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4872531 c.6519G>A p.S2173S Chr.15: 71854417-71854417 on Build GRCh38
3887401 c.6305C>T p.S2102L Chr.15: 71854418-71854418 on Build GRCh38
6077817 c.6341A>G p.D2114G Chr.15: 71854382-71854382 on Build GRCh38
4862942 c.6556A>T p.T2186S Chr.15: 71854380-71854380 on Build GRCh38
6077818 c.6554A>G p.D2185G Chr.15: 71854382-71854382 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774065269 Chr.15: 71854317-71854318 on Build GRCh38
rs934995652 Chr.15: 71854217-71854217 on Build GRCh38
rs762511123 Chr.15: 71854216-71854216 on Build GRCh38
rs1052140890 Chr.15: 71854220-71854221 on Build GRCh38
rs1004881351 Chr.15: 71854292-71854292 on Build GRCh38
rs749487255 Chr.15: 71854393-71854393 on Build GRCh38
rs565507322 Chr.15: 71854218-71854218 on Build GRCh38
rs1033985840 Chr.15: 71854242-71854242 on Build GRCh38
rs535830873 Chr.15: 71854248-71854248 on Build GRCh38
rs1029214314 Chr.15: 71854338-71854338 on Build GRCh38
rs958387234 Chr.15: 71854244-71854244 on Build GRCh38
rs758183669 Chr.15: 71854329-71854329 on Build GRCh38
rs975482892 Chr.15: 71854390-71854390 on Build GRCh38
rs977247040 Chr.15: 71854309-71854309 on Build GRCh38
rs1024263466 Chr.15: 71854285-71854285 on Build GRCh38
rs34791308 Chr.15: 71854281-71854282 on Build GRCh38
rs745892917 Chr.15: 71854386-71854386 on Build GRCh38
rs765830775 Chr.15: 71854271-71854271 on Build GRCh38
rs946488512 Chr.15: 71854236-71854236 on Build GRCh38
rs145718155 Chr.15: 71854387-71854387 on Build GRCh38
rs148839084 Chr.15: 71854221-71854222 on Build GRCh38
rs557313234 Chr.15: 71854243-71854244 on Build GRCh38
rs994593511 Chr.15: 71854213-71854213 on Build GRCh38
rs372093588 Chr.15: 71854362-71854362 on Build GRCh38
rs777690323 Chr.15: 71854343-71854343 on Build GRCh38
rs967607432 Chr.15: 71854300-71854300 on Build GRCh38
rs911832207 Chr.15: 71854213-71854214 on Build GRCh38
rs757169963 Chr.15: 71854366-71854366 on Build GRCh38
rs1014935941 Chr.15: 71854298-71854298 on Build GRCh38
rs115844073 Chr.15: 71854320-71854320 on Build GRCh38
rs781005199 Chr.15: 71854383-71854383 on Build GRCh38
rs1035927506 Chr.15: 71854255-71854255 on Build GRCh38

View Full Product Details